Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102359376-102359909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CINP
Entrez Gene ID
51550
Gene Name
cyclin dependent kinase 2 interacting protein
Gene Aliases
-
UniGene
Hs.129634
Gene Chromosome Location
Chr.14: 102341102-102362916 on Build GRCh38
Transcripts
NM_001320046.1 NM_032630.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
953548 c.64A>C p.K22Q Chr.14: 102359531-102359531 on Build GRCh38
4672461 c.42T>C p.P14P Chr.14: 102359553-102359553 on Build GRCh38
3753787 c.159C>G p.N53K Chr.14: 102359436-102359436 on Build GRCh38
279906 c.155C>T p.A52V Chr.14: 102359440-102359440 on Build GRCh38
5980674 c.56G>C p.S19T Chr.14: 102359539-102359539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759078422 Chr.14: 102359643-102359643 on Build GRCh38
rs369073138 Chr.14: 102359456-102359456 on Build GRCh38
rs971529761 Chr.14: 102359667-102359667 on Build GRCh38
rs140348090 Chr.14: 102359852-102359852 on Build GRCh38
rs773739015 Chr.14: 102359599-102359599 on Build GRCh38
rs766717863 Chr.14: 102359750-102359750 on Build GRCh38
rs767820135 Chr.14: 102359612-102359614 on Build GRCh38
rs11629090 Chr.14: 102359800-102359800 on Build GRCh38
rs996211320 Chr.14: 102359826-102359826 on Build GRCh38
rs946256753 Chr.14: 102359656-102359656 on Build GRCh38
rs1050771578 Chr.14: 102359570-102359570 on Build GRCh38
rs145802438 Chr.14: 102359475-102359475 on Build GRCh38
rs755617740 Chr.14: 102359439-102359439 on Build GRCh38
rs753382808 Chr.14: 102359446-102359446 on Build GRCh38
rs777845643 Chr.14: 102359464-102359464 on Build GRCh38
rs772403351 Chr.14: 102359595-102359595 on Build GRCh38
rs376024315 Chr.14: 102359515-102359515 on Build GRCh38
rs138637384 Chr.14: 102359493-102359493 on Build GRCh38
rs921076365 Chr.14: 102359843-102359843 on Build GRCh38
rs770487318 Chr.14: 102359628-102359628 on Build GRCh38
rs1041881444 Chr.14: 102359660-102359660 on Build GRCh38
rs890172638 Chr.14: 102359863-102359863 on Build GRCh38
rs147428140 Chr.14: 102359535-102359535 on Build GRCh38
rs562361918 Chr.14: 102359519-102359519 on Build GRCh38
rs776061264 Chr.14: 102359630-102359630 on Build GRCh38
rs564105226 Chr.14: 102359768-102359768 on Build GRCh38
rs753532232 Chr.14: 102359520-102359520 on Build GRCh38
rs542293766 Chr.14: 102359624-102359624 on Build GRCh38
rs759890397 Chr.14: 102359596-102359597 on Build GRCh38
rs1010789144 Chr.14: 102359445-102359445 on Build GRCh38
rs768820933 Chr.14: 102359566-102359566 on Build GRCh38
rs777902898 Chr.14: 102359543-102359543 on Build GRCh38
rs144884749 Chr.14: 102359492-102359492 on Build GRCh38
rs917280226 Chr.14: 102359633-102359633 on Build GRCh38
rs904680301 Chr.14: 102359680-102359680 on Build GRCh38
rs772633817 Chr.14: 102359514-102359514 on Build GRCh38
rs950728834 Chr.14: 102359786-102359786 on Build GRCh38
rs975298657 Chr.14: 102359842-102359842 on Build GRCh38
rs748552839 Chr.14: 102359592-102359592 on Build GRCh38
rs751497706 Chr.14: 102359551-102359551 on Build GRCh38
rs941269467 Chr.14: 102359833-102359833 on Build GRCh38
rs11544426 Chr.14: 102359436-102359436 on Build GRCh38
rs1000545288 Chr.14: 102359469-102359469 on Build GRCh38
rs779192439 Chr.14: 102359569-102359569 on Build GRCh38
rs766060539 Chr.14: 102359441-102359441 on Build GRCh38
rs909843644 Chr.14: 102359812-102359812 on Build GRCh38
rs774765787 Chr.14: 102359754-102359754 on Build GRCh38
rs749433358 Chr.14: 102359478-102359478 on Build GRCh38
rs373406995 Chr.14: 102359467-102359467 on Build GRCh38
rs756297430 Chr.14: 102359563-102359563 on Build GRCh38
rs767030250 Chr.14: 102359400-102359400 on Build GRCh38
rs749541105 Chr.14: 102359565-102359565 on Build GRCh38
rs936263171 Chr.14: 102359765-102359765 on Build GRCh38
rs764927652 Chr.14: 102359526-102359526 on Build GRCh38
rs199840100 Chr.14: 102359473-102359473 on Build GRCh38
rs973090009 Chr.14: 102359603-102359604 on Build GRCh38
rs200858256 Chr.14: 102359539-102359539 on Build GRCh38
rs1041974009 Chr.14: 102359455-102359455 on Build GRCh38
rs763501720 Chr.14: 102359544-102359544 on Build GRCh38
rs779934364 Chr.14: 102359564-102359564 on Build GRCh38
rs992931892 Chr.14: 102359593-102359593 on Build GRCh38
rs769071939 Chr.14: 102359486-102359486 on Build GRCh38
rs368991800 Chr.14: 102359600-102359600 on Build GRCh38
rs754831981 Chr.14: 102359453-102359453 on Build GRCh38
rs572825595 Chr.14: 102359572-102359572 on Build GRCh38
rs758464250 Chr.14: 102359457-102359457 on Build GRCh38
rs1050843268 Chr.14: 102359875-102359875 on Build GRCh38

View Full Product Details