Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103338656-103339161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5
Entrez Gene ID
1983
Gene Name
eukaryotic translation initiation factor 5
Gene Aliases
EIF-5, EIF-5A
UniGene
Hs.433702
Gene Chromosome Location
Chr.14: 103334002-103345025 on Build GRCh38
Transcripts
NM_001969.4 NM_183004.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
697001 c.625G>T p.E209* Chr.14: 103338774-103338774 on Build GRCh38
7245790 c.604G>C p.D202H Chr.14: 103338753-103338753 on Build GRCh38
4759661 c.659G>T p.S220I Chr.14: 103338808-103338808 on Build GRCh38
5362332 c.704delC p.T235fs*38 Chr.14: 103338853-103338853 on Build GRCh38
5743679 c.641G>A p.R214H Chr.14: 103338790-103338790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145996139 Chr.14: 103339097-103339097 on Build GRCh38
rs373721891 Chr.14: 103338729-103338729 on Build GRCh38
rs766937738 Chr.14: 103338875-103338875 on Build GRCh38
rs745608982 Chr.14: 103338903-103338906 on Build GRCh38
rs116234673 Chr.14: 103339033-103339033 on Build GRCh38
rs751664078 Chr.14: 103338692-103338701 on Build GRCh38
rs537681270 Chr.14: 103338716-103338716 on Build GRCh38
rs949531711 Chr.14: 103338970-103338970 on Build GRCh38
rs776694142 Chr.14: 103338684-103338684 on Build GRCh38
rs955338737 Chr.14: 103338946-103338946 on Build GRCh38
rs186685915 Chr.14: 103339009-103339009 on Build GRCh38
rs544451648 Chr.14: 103339004-103339004 on Build GRCh38
rs559021623 Chr.14: 103338992-103338992 on Build GRCh38
rs575908251 Chr.14: 103339002-103339002 on Build GRCh38
rs1027220565 Chr.14: 103339086-103339086 on Build GRCh38
rs191481387 Chr.14: 103338961-103338961 on Build GRCh38
rs920190592 Chr.14: 103338956-103338956 on Build GRCh38
rs775776236 Chr.14: 103338698-103338698 on Build GRCh38
rs749466812 Chr.14: 103339121-103339121 on Build GRCh38
rs532040550 Chr.14: 103339036-103339036 on Build GRCh38
rs996633996 Chr.14: 103339017-103339017 on Build GRCh38
rs755013187 Chr.14: 103338703-103338705 on Build GRCh38
rs753724160 Chr.14: 103338906-103338906 on Build GRCh38
rs778640706 Chr.14: 103338902-103338902 on Build GRCh38
rs370173098 Chr.14: 103338953-103338953 on Build GRCh38
rs143595557 Chr.14: 103338855-103338855 on Build GRCh38
rs1137359 Chr.14: 103338757-103338757 on Build GRCh38
rs183913820 Chr.14: 103338966-103338966 on Build GRCh38
rs115282451 Chr.14: 103338991-103338991 on Build GRCh38
rs755758368 Chr.14: 103338719-103338719 on Build GRCh38
rs939608545 Chr.14: 103339030-103339030 on Build GRCh38
rs1049583142 Chr.14: 103339018-103339018 on Build GRCh38
rs752748168 Chr.14: 103338951-103338951 on Build GRCh38
rs751124726 Chr.14: 103338706-103338706 on Build GRCh38
rs188671824 Chr.14: 103338938-103338938 on Build GRCh38
rs768929388 Chr.14: 103339135-103339135 on Build GRCh38
rs201881623 Chr.14: 103338775-103338775 on Build GRCh38
rs778720875 Chr.14: 103338911-103338911 on Build GRCh38
rs781266081 Chr.14: 103338783-103338783 on Build GRCh38
rs763275534 Chr.14: 103338834-103338834 on Build GRCh38
rs754798567 Chr.14: 103338909-103338909 on Build GRCh38
rs747160865 Chr.14: 103338773-103338773 on Build GRCh38
rs780235735 Chr.14: 103338935-103338935 on Build GRCh38
rs371697061 Chr.14: 103338929-103338929 on Build GRCh38
rs573318644 Chr.14: 103338967-103338967 on Build GRCh38
rs765590186 Chr.14: 103338696-103338696 on Build GRCh38
rs749888858 Chr.14: 103338878-103338878 on Build GRCh38
rs548383514 Chr.14: 103338717-103338717 on Build GRCh38
rs934405166 Chr.14: 103338686-103338686 on Build GRCh38
rs770948575 Chr.14: 103338781-103338781 on Build GRCh38
rs961306316 Chr.14: 103339050-103339050 on Build GRCh38
rs764583013 Chr.14: 103338703-103338703 on Build GRCh38
rs755030744 Chr.14: 103338749-103338749 on Build GRCh38
rs539980408 Chr.14: 103339027-103339027 on Build GRCh38
rs752581668 Chr.14: 103338914-103338914 on Build GRCh38
rs756236739 Chr.14: 103338955-103338955 on Build GRCh38
rs567713328 Chr.14: 103338740-103338740 on Build GRCh38
rs1025112834 Chr.14: 103339129-103339129 on Build GRCh38
rs778683941 Chr.14: 103338755-103338755 on Build GRCh38
rs924285139 Chr.14: 103338980-103338980 on Build GRCh38
rs774883300 Chr.14: 103338684-103338686 on Build GRCh38
rs749273358 Chr.14: 103338741-103338743 on Build GRCh38
rs898330731 Chr.14: 103339061-103339061 on Build GRCh38
rs756702834 Chr.14: 103338709-103338709 on Build GRCh38
rs991008910 Chr.14: 103338965-103338965 on Build GRCh38
rs370211286 Chr.14: 103338928-103338928 on Build GRCh38
rs961077534 Chr.14: 103338954-103338954 on Build GRCh38
rs544654901 Chr.14: 103338767-103338767 on Build GRCh38
rs775902979 Chr.14: 103338812-103338812 on Build GRCh38
rs995198736 Chr.14: 103339068-103339068 on Build GRCh38
rs766124326 Chr.14: 103338902-103338902 on Build GRCh38
rs760458639 Chr.14: 103338887-103338887 on Build GRCh38
rs936703195 Chr.14: 103338993-103338993 on Build GRCh38
rs781219443 Chr.14: 103338713-103338714 on Build GRCh38
rs774746517 Chr.14: 103338866-103338866 on Build GRCh38
rs775231576 Chr.14: 103338933-103338934 on Build GRCh38
rs755158929 Chr.14: 103338746-103338746 on Build GRCh38
rs142695713 Chr.14: 103339011-103339011 on Build GRCh38
rs771635560 Chr.14: 103338931-103338932 on Build GRCh38
rs1038450156 Chr.14: 103339056-103339057 on Build GRCh38
rs763511551 Chr.14: 103338699-103338699 on Build GRCh38
rs887127605 Chr.14: 103339034-103339034 on Build GRCh38
rs371375525 Chr.14: 103338689-103338689 on Build GRCh38
rs758298876 Chr.14: 103338922-103338922 on Build GRCh38
rs762337983 Chr.14: 103338870-103338870 on Build GRCh38
rs867959385 Chr.14: 103338783-103338784 on Build GRCh38
rs896980627 Chr.14: 103339112-103339112 on Build GRCh38
rs757206797 Chr.14: 103338895-103338896 on Build GRCh38
rs746094041 Chr.14: 103338790-103338790 on Build GRCh38
rs562609310 Chr.14: 103339101-103339101 on Build GRCh38
rs1007886791 Chr.14: 103338947-103338947 on Build GRCh38
rs1056871414 Chr.14: 103338856-103338856 on Build GRCh38
rs138136527 Chr.14: 103338807-103338807 on Build GRCh38

View Full Product Details