Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105491150-105491652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf80
Entrez Gene ID
283643
Gene Name
chromosome 14 open reading frame 80
Gene Aliases
-
UniGene
Hs.720306
Gene Chromosome Location
Chr.14: 105489855-105499248 on Build GRCh38
Transcripts
NM_001134875.1 NM_001134876.1 NM_001134877.1 NM_001198983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428306 c.28C>A p.P10T Chr.14: 105491403-105491403 on Build GRCh38
6015649 c.54C>G p.L18L Chr.14: 105491645-105491645 on Build GRCh38
6431669 c.86C>T p.S29L Chr.14: 105491461-105491461 on Build GRCh38
4409723 c.178G>T p.V60L Chr.14: 105491652-105491652 on Build GRCh38
6015650 c.171C>G p.L57L Chr.14: 105491645-105491645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782524145 Chr.14: 105491262-105491262 on Build GRCh38
rs587594638 Chr.14: 105491458-105491458 on Build GRCh38
rs587745330 Chr.14: 105491600-105491600 on Build GRCh38
rs782677666 Chr.14: 105491617-105491617 on Build GRCh38
rs781876508 Chr.14: 105491580-105491580 on Build GRCh38
rs587758610 Chr.14: 105491314-105491314 on Build GRCh38
rs979290216 Chr.14: 105491292-105491292 on Build GRCh38
rs896219472 Chr.14: 105491220-105491220 on Build GRCh38
rs782732626 Chr.14: 105491336-105491336 on Build GRCh38
rs1009266463 Chr.14: 105491226-105491226 on Build GRCh38
rs587657980 Chr.14: 105491324-105491324 on Build GRCh38
rs587678565 Chr.14: 105491535-105491535 on Build GRCh38
rs929363210 Chr.14: 105491361-105491361 on Build GRCh38
rs997328066 Chr.14: 105491592-105491592 on Build GRCh38
rs1057480978 Chr.14: 105491477-105491477 on Build GRCh38
rs34183800 Chr.14: 105491563-105491564 on Build GRCh38
rs115896717 Chr.14: 105491175-105491175 on Build GRCh38
rs964475353 Chr.14: 105491319-105491319 on Build GRCh38
rs923408730 Chr.14: 105491330-105491330 on Build GRCh38
rs1020632849 Chr.14: 105491531-105491531 on Build GRCh38
rs115066186 Chr.14: 105491554-105491554 on Build GRCh38
rs1009710023 Chr.14: 105491510-105491510 on Build GRCh38
rs942266793 Chr.14: 105491410-105491410 on Build GRCh38
rs1044626528 Chr.14: 105491433-105491433 on Build GRCh38
rs905995484 Chr.14: 105491171-105491171 on Build GRCh38
rs587737632 Chr.14: 105491396-105491396 on Build GRCh38
rs587659699 Chr.14: 105491586-105491586 on Build GRCh38
rs1000740184 Chr.14: 105491550-105491550 on Build GRCh38
rs1030554188 Chr.14: 105491593-105491593 on Build GRCh38
rs896261752 Chr.14: 105491493-105491493 on Build GRCh38
rs781897415 Chr.14: 105491229-105491229 on Build GRCh38
rs983489160 Chr.14: 105491389-105491389 on Build GRCh38
rs1001954313 Chr.14: 105491172-105491172 on Build GRCh38
rs781787303 Chr.14: 105491631-105491631 on Build GRCh38
rs950842105 Chr.14: 105491630-105491630 on Build GRCh38
rs938866683 Chr.14: 105491466-105491466 on Build GRCh38
rs587735601 Chr.14: 105491538-105491538 on Build GRCh38
rs968010777 Chr.14: 105491240-105491240 on Build GRCh38
rs964585989 Chr.14: 105491572-105491572 on Build GRCh38

View Full Product Details