Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105492009-105492504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf80
Entrez Gene ID
283643
Gene Name
chromosome 14 open reading frame 80
Gene Aliases
-
UniGene
Hs.720306
Gene Chromosome Location
Chr.14: 105489855-105499248 on Build GRCh38
Transcripts
NM_001134875.1 NM_001134876.1 NM_001134877.1 NM_001198983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274015 c.238C>A p.R80S Chr.14: 105492118-105492118 on Build GRCh38
4403542 c.395G>A p.R132Q Chr.14: 105492275-105492275 on Build GRCh38
4405826 c.397G>A p.V133M Chr.14: 105492277-105492277 on Build GRCh38
6249459 c.253G>T p.A85S Chr.14: 105492133-105492133 on Build GRCh38
432760 c.358C>G p.R120G Chr.14: 105492238-105492238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782308934 Chr.14: 105492291-105492291 on Build GRCh38
rs200813738 Chr.14: 105492277-105492277 on Build GRCh38
rs781993666 Chr.14: 105492219-105492219 on Build GRCh38
rs781863269 Chr.14: 105492164-105492164 on Build GRCh38
rs755895663 Chr.14: 105492138-105492138 on Build GRCh38
rs782529386 Chr.14: 105492184-105492184 on Build GRCh38
rs781900336 Chr.14: 105492111-105492111 on Build GRCh38
rs145100366 Chr.14: 105492056-105492056 on Build GRCh38
rs587673531 Chr.14: 105492081-105492081 on Build GRCh38
rs782155658 Chr.14: 105492163-105492163 on Build GRCh38
rs782735860 Chr.14: 105492117-105492117 on Build GRCh38
rs1037554523 Chr.14: 105492388-105492388 on Build GRCh38
rs782462134 Chr.14: 105492267-105492267 on Build GRCh38
rs782008482 Chr.14: 105492160-105492160 on Build GRCh38
rs1014701129 Chr.14: 105492455-105492455 on Build GRCh38
rs1020260039 Chr.14: 105492457-105492457 on Build GRCh38
rs200405621 Chr.14: 105492333-105492333 on Build GRCh38
rs587618908 Chr.14: 105492116-105492116 on Build GRCh38
rs782202834 Chr.14: 105492354-105492354 on Build GRCh38
rs375248543 Chr.14: 105492313-105492313 on Build GRCh38
rs886381048 Chr.14: 105492336-105492336 on Build GRCh38
rs782240144 Chr.14: 105492087-105492087 on Build GRCh38
rs782614506 Chr.14: 105492274-105492274 on Build GRCh38
rs587708518 Chr.14: 105492275-105492275 on Build GRCh38
rs1052151841 Chr.14: 105492064-105492064 on Build GRCh38
rs373315186 Chr.14: 105492253-105492253 on Build GRCh38
rs375685931 Chr.14: 105492225-105492225 on Build GRCh38
rs782084018 Chr.14: 105492293-105492293 on Build GRCh38
rs587720400 Chr.14: 105492357-105492357 on Build GRCh38
rs782248319 Chr.14: 105492078-105492078 on Build GRCh38
rs199951748 Chr.14: 105492320-105492320 on Build GRCh38
rs587605781 Chr.14: 105492407-105492407 on Build GRCh38
rs782107920 Chr.14: 105492179-105492179 on Build GRCh38
rs181709041 Chr.14: 105492119-105492119 on Build GRCh38
rs782140667 Chr.14: 105492237-105492237 on Build GRCh38
rs587661811 Chr.14: 105492460-105492460 on Build GRCh38
rs781817985 Chr.14: 105492238-105492238 on Build GRCh38
rs374016169 Chr.14: 105492340-105492340 on Build GRCh38
rs1005150347 Chr.14: 105492080-105492080 on Build GRCh38
rs190336306 Chr.14: 105492199-105492199 on Build GRCh38
rs782606235 Chr.14: 105492359-105492359 on Build GRCh38
rs368575867 Chr.14: 105492191-105492191 on Build GRCh38
rs781993743 Chr.14: 105492376-105492376 on Build GRCh38
rs370719810 Chr.14: 105492290-105492290 on Build GRCh38
rs782132996 Chr.14: 105492312-105492312 on Build GRCh38
rs186092145 Chr.14: 105492397-105492397 on Build GRCh38
rs886349109 Chr.14: 105492065-105492065 on Build GRCh38
rs587723635 Chr.14: 105492180-105492180 on Build GRCh38
rs587677061 Chr.14: 105492200-105492200 on Build GRCh38
rs180744711 Chr.14: 105492203-105492203 on Build GRCh38
rs587683388 Chr.14: 105492202-105492202 on Build GRCh38
rs782053307 Chr.14: 105492239-105492239 on Build GRCh38
rs377507500 Chr.14: 105492281-105492281 on Build GRCh38
rs782701617 Chr.14: 105492317-105492317 on Build GRCh38
rs201690661 Chr.14: 105492241-105492241 on Build GRCh38
rs959989840 Chr.14: 105492166-105492166 on Build GRCh38
rs781965220 Chr.14: 105492155-105492155 on Build GRCh38
rs782276134 Chr.14: 105492120-105492120 on Build GRCh38
rs199739141 Chr.14: 105492153-105492153 on Build GRCh38
rs781784997 Chr.14: 105492265-105492265 on Build GRCh38
rs782528643 Chr.14: 105492259-105492259 on Build GRCh38
rs587595528 Chr.14: 105492370-105492370 on Build GRCh38
rs185620086 Chr.14: 105492178-105492178 on Build GRCh38
rs372925511 Chr.14: 105492216-105492216 on Build GRCh38
rs781894445 Chr.14: 105492342-105492342 on Build GRCh38
rs782018230 Chr.14: 105492082-105492082 on Build GRCh38
rs1034473777 Chr.14: 105492415-105492415 on Build GRCh38
rs782078773 Chr.14: 105492097-105492097 on Build GRCh38
rs782787250 Chr.14: 105492109-105492109 on Build GRCh38
rs781933301 Chr.14: 105492092-105492092 on Build GRCh38
rs781912905 Chr.14: 105492204-105492204 on Build GRCh38
rs781854466 Chr.14: 105492102-105492102 on Build GRCh38
rs895894175 Chr.14: 105492429-105492429 on Build GRCh38
rs781981411 Chr.14: 105492301-105492301 on Build GRCh38
rs782633199 Chr.14: 105492077-105492077 on Build GRCh38
rs781818686 Chr.14: 105492248-105492248 on Build GRCh38
rs587676881 Chr.14: 105492030-105492031 on Build GRCh38
rs587729025 Chr.14: 105492103-105492103 on Build GRCh38
rs782186927 Chr.14: 105492205-105492205 on Build GRCh38
rs782737665 Chr.14: 105492100-105492100 on Build GRCh38
rs782819389 Chr.14: 105492185-105492185 on Build GRCh38
rs781900892 Chr.14: 105492314-105492314 on Build GRCh38
rs34119753 Chr.14: 105492414-105492414 on Build GRCh38
rs782674304 Chr.14: 105492286-105492286 on Build GRCh38
rs782169968 Chr.14: 105492086-105492086 on Build GRCh38
rs377676869 Chr.14: 105492345-105492345 on Build GRCh38
rs782599615 Chr.14: 105492192-105492192 on Build GRCh38
rs375461639 Chr.14: 105492136-105492136 on Build GRCh38
rs1001592981 Chr.14: 105492135-105492135 on Build GRCh38
rs112465101 Chr.14: 105492035-105492036 on Build GRCh38
rs782351985 Chr.14: 105492481-105492481 on Build GRCh38
rs782354765 Chr.14: 105492297-105492297 on Build GRCh38
rs781879165 Chr.14: 105492182-105492182 on Build GRCh38
rs782297610 Chr.14: 105492287-105492287 on Build GRCh38
rs782695146 Chr.14: 105492175-105492175 on Build GRCh38
rs587672374 Chr.14: 105492391-105492391 on Build GRCh38

View Full Product Details