Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105493755-105494256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf80
Entrez Gene ID
283643
Gene Name
chromosome 14 open reading frame 80
Gene Aliases
-
UniGene
Hs.720306
Gene Chromosome Location
Chr.14: 105489855-105499248 on Build GRCh38
Transcripts
NM_001134875.1 NM_001134876.1 NM_001134877.1 NM_001198983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4049578 c.586-1G>C p.? Chr.14: 105493834-105493834 on Build GRCh38
6300988 c.123G>A p.M41I Chr.14: 105493906-105493906 on Build GRCh38
1477406 c.104C>T p.S35F Chr.14: 105493887-105493887 on Build GRCh38
7347966 c.601C>T p.R201C Chr.14: 105493850-105493850 on Build GRCh38
6300987 c.657G>A p.M219I Chr.14: 105493906-105493906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782391461 Chr.14: 105493962-105493962 on Build GRCh38
rs201083148 Chr.14: 105493956-105493956 on Build GRCh38
rs59080216 Chr.14: 105493851-105493851 on Build GRCh38
rs781880234 Chr.14: 105494166-105494166 on Build GRCh38
rs781827809 Chr.14: 105494119-105494119 on Build GRCh38
rs77656590 Chr.14: 105494167-105494167 on Build GRCh38
rs782034453 Chr.14: 105493966-105493966 on Build GRCh38
rs782813113 Chr.14: 105493929-105493929 on Build GRCh38
rs587730200 Chr.14: 105493816-105493816 on Build GRCh38
rs947309915 Chr.14: 105493907-105493907 on Build GRCh38
rs943940867 Chr.14: 105494175-105494175 on Build GRCh38
rs781789340 Chr.14: 105493825-105493825 on Build GRCh38
rs782198994 Chr.14: 105493969-105493969 on Build GRCh38
rs782472590 Chr.14: 105493959-105493959 on Build GRCh38
rs373964506 Chr.14: 105494044-105494044 on Build GRCh38
rs148446003 Chr.14: 105493845-105493845 on Build GRCh38
rs782106207 Chr.14: 105493808-105493808 on Build GRCh38
rs782154880 Chr.14: 105493973-105493974 on Build GRCh38
rs201752678 Chr.14: 105493925-105493925 on Build GRCh38
rs782759694 Chr.14: 105493873-105493873 on Build GRCh38
rs1024337535 Chr.14: 105494085-105494085 on Build GRCh38
rs1012686732 Chr.14: 105494224-105494224 on Build GRCh38
rs59013219 Chr.14: 105493922-105493922 on Build GRCh38
rs587701408 Chr.14: 105493968-105493968 on Build GRCh38
rs914314499 Chr.14: 105494160-105494160 on Build GRCh38
rs934420773 Chr.14: 105494147-105494147 on Build GRCh38
rs587648068 Chr.14: 105494081-105494081 on Build GRCh38
rs934389740 Chr.14: 105493867-105493867 on Build GRCh38
rs587620649 Chr.14: 105494218-105494218 on Build GRCh38
rs1050140279 Chr.14: 105493912-105493912 on Build GRCh38
rs144656487 Chr.14: 105493862-105493862 on Build GRCh38
rs781819407 Chr.14: 105493872-105493872 on Build GRCh38
rs80086621 Chr.14: 105493905-105493905 on Build GRCh38
rs782046320 Chr.14: 105493801-105493801 on Build GRCh38
rs782719867 Chr.14: 105493803-105493803 on Build GRCh38
rs587724791 Chr.14: 105493974-105493974 on Build GRCh38
rs371651798 Chr.14: 105493941-105493941 on Build GRCh38
rs781963677 Chr.14: 105493806-105493806 on Build GRCh38
rs782481403 Chr.14: 105493938-105493938 on Build GRCh38
rs782631414 Chr.14: 105493895-105493895 on Build GRCh38
rs782795011 Chr.14: 105493819-105493819 on Build GRCh38
rs140812013 Chr.14: 105493853-105493853 on Build GRCh38
rs371562409 Chr.14: 105494188-105494188 on Build GRCh38
rs982708834 Chr.14: 105494110-105494110 on Build GRCh38
rs782463724 Chr.14: 105494158-105494158 on Build GRCh38
rs782570786 Chr.14: 105493880-105493880 on Build GRCh38
rs782574677 Chr.14: 105493954-105493955 on Build GRCh38
rs370630683 Chr.14: 105493798-105493798 on Build GRCh38
rs147865275 Chr.14: 105493865-105493865 on Build GRCh38
rs782246489 Chr.14: 105493896-105493896 on Build GRCh38
rs782758382 Chr.14: 105493812-105493812 on Build GRCh38
rs782226694 Chr.14: 105493836-105493836 on Build GRCh38
rs782563406 Chr.14: 105493958-105493958 on Build GRCh38
rs782671488 Chr.14: 105493840-105493840 on Build GRCh38
rs896950565 Chr.14: 105494184-105494184 on Build GRCh38
rs199611581 Chr.14: 105493981-105493981 on Build GRCh38
rs988482049 Chr.14: 105494159-105494159 on Build GRCh38
rs782625454 Chr.14: 105493960-105493960 on Build GRCh38
rs778616457 Chr.14: 105494045-105494045 on Build GRCh38
rs782764432 Chr.14: 105493840-105493840 on Build GRCh38
rs782305933 Chr.14: 105493911-105493911 on Build GRCh38
rs111773029 Chr.14: 105493983-105493983 on Build GRCh38
rs587682527 Chr.14: 105494102-105494102 on Build GRCh38
rs781932897 Chr.14: 105493913-105493913 on Build GRCh38
rs201701957 Chr.14: 105493942-105493942 on Build GRCh38
rs192498325 Chr.14: 105494171-105494171 on Build GRCh38
rs200525924 Chr.14: 105493977-105493977 on Build GRCh38
rs587638468 Chr.14: 105493973-105493973 on Build GRCh38
rs781787226 Chr.14: 105493955-105493956 on Build GRCh38
rs782524294 Chr.14: 105493837-105493837 on Build GRCh38
rs371801390 Chr.14: 105493904-105493904 on Build GRCh38
rs781804769 Chr.14: 105493885-105493885 on Build GRCh38
rs782407779 Chr.14: 105493800-105493800 on Build GRCh38
rs75729586 Chr.14: 105494015-105494016 on Build GRCh38
rs782645455 Chr.14: 105493955-105493955 on Build GRCh38
rs782291389 Chr.14: 105493910-105493910 on Build GRCh38
rs377631365 Chr.14: 105493940-105493940 on Build GRCh38
rs781894955 Chr.14: 105493878-105493878 on Build GRCh38
rs782078661 Chr.14: 105494163-105494163 on Build GRCh38
rs61641418 Chr.14: 105493982-105493982 on Build GRCh38
rs782726555 Chr.14: 105493870-105493870 on Build GRCh38
rs990812720 Chr.14: 105494064-105494064 on Build GRCh38
rs187863153 Chr.14: 105493780-105493780 on Build GRCh38
rs587664633 Chr.14: 105494041-105494041 on Build GRCh38
rs782446315 Chr.14: 105493827-105493827 on Build GRCh38
rs782292459 Chr.14: 105493841-105493841 on Build GRCh38
rs587759556 Chr.14: 105493969-105493969 on Build GRCh38
rs150648235 Chr.14: 105493852-105493852 on Build GRCh38
rs782601510 Chr.14: 105493832-105493832 on Build GRCh38
rs782184388 Chr.14: 105493967-105493967 on Build GRCh38
rs782036378 Chr.14: 105493971-105493972 on Build GRCh38
rs781991381 Chr.14: 105493974-105493975 on Build GRCh38
rs781792222 Chr.14: 105493938-105493938 on Build GRCh38
rs368147486 Chr.14: 105494020-105494021 on Build GRCh38
rs782743629 Chr.14: 105494092-105494092 on Build GRCh38
rs782470937 Chr.14: 105493893-105493893 on Build GRCh38
rs782517511 Chr.14: 105493946-105493946 on Build GRCh38
rs952730235 Chr.14: 105494056-105494056 on Build GRCh38
rs112054580 Chr.14: 105494183-105494183 on Build GRCh38
rs919896877 Chr.14: 105493849-105493849 on Build GRCh38
rs1027107282 Chr.14: 105494206-105494206 on Build GRCh38
rs111318600 Chr.14: 105494020-105494020 on Build GRCh38
rs782080808 Chr.14: 105493916-105493916 on Build GRCh38
rs375393536 Chr.14: 105493813-105493813 on Build GRCh38

View Full Product Details