Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 24675304-24675795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAP1
Entrez Gene ID
23742
Gene Name
nuclear pore associated protein 1
Gene Aliases
C15orf2
UniGene
Hs.649663
Gene Chromosome Location
Chr.15: 24675394-24683446 on Build GRCh38
Transcripts
NM_018958.2

SNP Detail

SNP ID Chromosome Location
rs113890317 Chr.15: 24675754-24675754 on Build GRCh38
rs977255602 Chr.15: 24675397-24675397 on Build GRCh38
rs932321687 Chr.15: 24675450-24675450 on Build GRCh38
rs1049302169 Chr.15: 24675757-24675757 on Build GRCh38
rs970792532 Chr.15: 24675441-24675441 on Build GRCh38
rs1050842019 Chr.15: 24675467-24675467 on Build GRCh38
rs894454946 Chr.15: 24675547-24675547 on Build GRCh38
rs998640770 Chr.15: 24675611-24675611 on Build GRCh38
rs148084832 Chr.15: 24675423-24675423 on Build GRCh38
rs943029537 Chr.15: 24675343-24675343 on Build GRCh38
rs939713265 Chr.15: 24675721-24675721 on Build GRCh38
rs912337878 Chr.15: 24675481-24675481 on Build GRCh38
rs745524000 Chr.15: 24675532-24675532 on Build GRCh38
rs542377967 Chr.15: 24675514-24675514 on Build GRCh38
rs1028676098 Chr.15: 24675647-24675647 on Build GRCh38
rs1024333006 Chr.15: 24675569-24675569 on Build GRCh38
rs369326025 Chr.15: 24675447-24675447 on Build GRCh38
rs972485446 Chr.15: 24675739-24675739 on Build GRCh38
rs987390418 Chr.15: 24675499-24675499 on Build GRCh38
rs372430606 Chr.15: 24675745-24675745 on Build GRCh38
rs2602618 Chr.15: 24675448-24675448 on Build GRCh38
rs538611941 Chr.15: 24675580-24675580 on Build GRCh38
rs990053075 Chr.15: 24675641-24675641 on Build GRCh38
rs1013359242 Chr.15: 24675566-24675566 on Build GRCh38
rs919396555 Chr.15: 24675750-24675750 on Build GRCh38
rs548045885 Chr.15: 24675346-24675346 on Build GRCh38
rs953719328 Chr.15: 24675651-24675651 on Build GRCh38
rs887296928 Chr.15: 24675568-24675568 on Build GRCh38
rs894750160 Chr.15: 24675764-24675764 on Build GRCh38
rs779666816 Chr.15: 24675624-24675624 on Build GRCh38
rs201671866 Chr.15: 24675535-24675536 on Build GRCh38
rs998287806 Chr.15: 24675707-24675722 on Build GRCh38
rs547407585 Chr.15: 24675700-24675700 on Build GRCh38
rs989496417 Chr.15: 24675471-24675471 on Build GRCh38
rs1025258986 Chr.15: 24675577-24675577 on Build GRCh38
rs1033650946 Chr.15: 24675462-24675462 on Build GRCh38
rs957207503 Chr.15: 24675637-24675637 on Build GRCh38
rs375985591 Chr.15: 24675361-24675361 on Build GRCh38
rs966552263 Chr.15: 24675753-24675753 on Build GRCh38
rs750770887 Chr.15: 24675720-24675720 on Build GRCh38
rs930390803 Chr.15: 24675537-24675537 on Build GRCh38
rs1036573281 Chr.15: 24675484-24675484 on Build GRCh38
rs527981328 Chr.15: 24675395-24675395 on Build GRCh38
rs1031426339 Chr.15: 24675635-24675635 on Build GRCh38
rs571440434 Chr.15: 24675510-24675510 on Build GRCh38
rs778620149 Chr.15: 24675508-24675508 on Build GRCh38
rs1047497354 Chr.15: 24675550-24675550 on Build GRCh38
rs957321759 Chr.15: 24675430-24675430 on Build GRCh38
rs139203195 Chr.15: 24675385-24675386 on Build GRCh38
rs769339308 Chr.15: 24675536-24675538 on Build GRCh38
rs771776266 Chr.15: 24675553-24675553 on Build GRCh38
rs181196875 Chr.15: 24675391-24675391 on Build GRCh38
rs924522342 Chr.15: 24675409-24675409 on Build GRCh38
rs760949250 Chr.15: 24675347-24675347 on Build GRCh38
rs116401581 Chr.15: 24675349-24675349 on Build GRCh38
rs986898822 Chr.15: 24675668-24675668 on Build GRCh38
rs943268117 Chr.15: 24675513-24675513 on Build GRCh38
rs1004495255 Chr.15: 24675416-24675416 on Build GRCh38
rs1049791083 Chr.15: 24675535-24675535 on Build GRCh38
rs112214851 Chr.15: 24675482-24675482 on Build GRCh38

View Full Product Details