Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 27480460-27480967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG3
Entrez Gene ID
2567
Gene Name
gamma-aminobutyric acid type A receptor gamma3 subunit
Gene Aliases
-
UniGene
Hs.569475
Gene Chromosome Location
Chr.15: 26971282-27537112 on Build GRCh38
Transcripts
NM_001270873.1 NM_033223.4 XM_011521430.2 XM_011521431.2 XM_017022058.1 XM_017022059.1 XM_017022060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608210 c.698T>A p.V233E Chr.15: 27480773-27480773 on Build GRCh38
1253010 c.617A>G p.N206S Chr.15: 27480692-27480692 on Build GRCh38
6554817 c.664G>T p.D222Y Chr.15: 27480739-27480739 on Build GRCh38
6554816 c.625G>T p.E209* Chr.15: 27480700-27480700 on Build GRCh38
7288849 c.603T>C p.Y201Y Chr.15: 27480678-27480678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774782890 Chr.15: 27480874-27480874 on Build GRCh38
rs1022736279 Chr.15: 27480528-27480528 on Build GRCh38
rs190554781 Chr.15: 27480922-27480922 on Build GRCh38
rs753122646 Chr.15: 27480613-27480613 on Build GRCh38
rs751777104 Chr.15: 27480631-27480631 on Build GRCh38
rs747827094 Chr.15: 27480841-27480841 on Build GRCh38
rs756176068 Chr.15: 27480811-27480811 on Build GRCh38
rs368856088 Chr.15: 27480791-27480791 on Build GRCh38
rs766342945 Chr.15: 27480814-27480814 on Build GRCh38
rs77771286 Chr.15: 27480771-27480771 on Build GRCh38
rs760147809 Chr.15: 27480601-27480601 on Build GRCh38
rs1005568534 Chr.15: 27480812-27480812 on Build GRCh38
rs753751678 Chr.15: 27480828-27480828 on Build GRCh38
rs78907752 Chr.15: 27480731-27480731 on Build GRCh38
rs754822558 Chr.15: 27480829-27480829 on Build GRCh38
rs750464172 Chr.15: 27480703-27480703 on Build GRCh38
rs763270961 Chr.15: 27480785-27480785 on Build GRCh38
rs373457978 Chr.15: 27480798-27480798 on Build GRCh38
rs762127317 Chr.15: 27480799-27480799 on Build GRCh38
rs760201044 Chr.15: 27480779-27480779 on Build GRCh38
rs990791140 Chr.15: 27480540-27480540 on Build GRCh38
rs758080208 Chr.15: 27480846-27480846 on Build GRCh38
rs140676 Chr.15: 27480527-27480527 on Build GRCh38
rs750517024 Chr.15: 27480807-27480807 on Build GRCh38
rs768465048 Chr.15: 27480755-27480755 on Build GRCh38
rs558188065 Chr.15: 27480780-27480780 on Build GRCh38
rs778477419 Chr.15: 27480762-27480762 on Build GRCh38
rs559247266 Chr.15: 27480870-27480870 on Build GRCh38
rs79486115 Chr.15: 27480739-27480739 on Build GRCh38
rs903910801 Chr.15: 27480545-27480545 on Build GRCh38
rs781358782 Chr.15: 27480680-27480680 on Build GRCh38
rs774918879 Chr.15: 27480798-27480799 on Build GRCh38
rs56174531 Chr.15: 27480934-27480934 on Build GRCh38
rs535880505 Chr.15: 27480597-27480597 on Build GRCh38
rs764293810 Chr.15: 27480628-27480628 on Build GRCh38
rs201602655 Chr.15: 27480772-27480772 on Build GRCh38
rs960527833 Chr.15: 27480638-27480638 on Build GRCh38
rs749828005 Chr.15: 27480866-27480866 on Build GRCh38
rs753884665 Chr.15: 27480882-27480882 on Build GRCh38
rs766621565 Chr.15: 27480792-27480792 on Build GRCh38
rs77287112 Chr.15: 27480724-27480724 on Build GRCh38
rs150368441 Chr.15: 27480621-27480621 on Build GRCh38
rs375229216 Chr.15: 27480675-27480675 on Build GRCh38
rs749140479 Chr.15: 27480746-27480746 on Build GRCh38
rs181117003 Chr.15: 27480833-27480833 on Build GRCh38
rs761499095 Chr.15: 27480590-27480590 on Build GRCh38
rs765768282 Chr.15: 27480606-27480606 on Build GRCh38
rs186401684 Chr.15: 27480884-27480884 on Build GRCh38
rs767660237 Chr.15: 27480805-27480805 on Build GRCh38
rs776191140 Chr.15: 27480865-27480865 on Build GRCh38
rs777551520 Chr.15: 27480851-27480851 on Build GRCh38
rs946411703 Chr.15: 27480611-27480611 on Build GRCh38
rs567157233 Chr.15: 27480534-27480534 on Build GRCh38
rs371881362 Chr.15: 27480602-27480602 on Build GRCh38
rs992436525 Chr.15: 27480651-27480651 on Build GRCh38
rs771536585 Chr.15: 27480616-27480616 on Build GRCh38
rs770536915 Chr.15: 27480864-27480864 on Build GRCh38
rs779910417 Chr.15: 27480708-27480708 on Build GRCh38
rs746581485 Chr.15: 27480857-27480857 on Build GRCh38
rs1023540026 Chr.15: 27480713-27480713 on Build GRCh38
rs756049787 Chr.15: 27480707-27480707 on Build GRCh38
rs570958383 Chr.15: 27480522-27480522 on Build GRCh38
rs368580825 Chr.15: 27480769-27480769 on Build GRCh38

View Full Product Details