Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30635166-30635676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11B
Entrez Gene ID
89839
Gene Name
Rho GTPase activating protein 11B
Gene Aliases
B'-T, FAM7B1
UniGene
Hs.659621
Gene Chromosome Location
Chr.15: 30626676-30638810 on Build GRCh38
Transcripts
NM_001039841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5980703 c.660+1G>C p.? Chr.15: 30635189-30635189 on Build GRCh38
2084201 c.661-7C>T p.? Chr.15: 30635480-30635480 on Build GRCh38
6030198 c.718T>G p.S240A Chr.15: 30635544-30635544 on Build GRCh38
5350250 c.683G>T p.W228L Chr.15: 30635509-30635509 on Build GRCh38
5461718 c.660+5G>A p.? Chr.15: 30635193-30635193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765065838 Chr.15: 30635465-30635465 on Build GRCh38
rs549506180 Chr.15: 30635396-30635396 on Build GRCh38
rs150590279 Chr.15: 30635212-30635213 on Build GRCh38
rs781346482 Chr.15: 30635291-30635292 on Build GRCh38
rs780843342 Chr.15: 30635635-30635635 on Build GRCh38
rs748401458 Chr.15: 30635224-30635224 on Build GRCh38
rs773026649 Chr.15: 30635527-30635527 on Build GRCh38
rs986005347 Chr.15: 30635259-30635259 on Build GRCh38
rs879332822 Chr.15: 30635468-30635468 on Build GRCh38
rs113953826 Chr.15: 30635481-30635481 on Build GRCh38
rs1001707761 Chr.15: 30635650-30635650 on Build GRCh38
rs766278322 Chr.15: 30635475-30635475 on Build GRCh38
rs374363097 Chr.15: 30635223-30635223 on Build GRCh38
rs750902247 Chr.15: 30635624-30635624 on Build GRCh38
rs1009236251 Chr.15: 30635229-30635229 on Build GRCh38
rs754109042 Chr.15: 30635599-30635599 on Build GRCh38
rs962673588 Chr.15: 30635646-30635646 on Build GRCh38
rs982737591 Chr.15: 30635373-30635373 on Build GRCh38
rs907482024 Chr.15: 30635645-30635645 on Build GRCh38
rs560934228 Chr.15: 30635365-30635365 on Build GRCh38
rs778197945 Chr.15: 30635489-30635489 on Build GRCh38
rs368543744 Chr.15: 30635225-30635225 on Build GRCh38
rs749657802 Chr.15: 30635490-30635490 on Build GRCh38
rs28444520 Chr.15: 30635314-30635314 on Build GRCh38
rs372915298 Chr.15: 30635233-30635233 on Build GRCh38
rs779236156 Chr.15: 30635500-30635500 on Build GRCh38
rs368349923 Chr.15: 30635467-30635467 on Build GRCh38
rs759030465 Chr.15: 30635625-30635625 on Build GRCh38
rs966012543 Chr.15: 30635525-30635525 on Build GRCh38
rs372005915 Chr.15: 30635578-30635578 on Build GRCh38
rs950583116 Chr.15: 30635402-30635402 on Build GRCh38
rs767574365 Chr.15: 30635564-30635564 on Build GRCh38
rs761562571 Chr.15: 30635460-30635460 on Build GRCh38
rs146907163 Chr.15: 30635609-30635609 on Build GRCh38
rs56230817 Chr.15: 30635428-30635428 on Build GRCh38
rs963157002 Chr.15: 30635371-30635371 on Build GRCh38
rs1001989886 Chr.15: 30635342-30635342 on Build GRCh38
rs369518739 Chr.15: 30635595-30635595 on Build GRCh38
rs199799461 Chr.15: 30635648-30635648 on Build GRCh38
rs757757462 Chr.15: 30635491-30635491 on Build GRCh38
rs768210811 Chr.15: 30635504-30635504 on Build GRCh38
rs765151317 Chr.15: 30635189-30635189 on Build GRCh38
rs755172659 Chr.15: 30635222-30635222 on Build GRCh38
rs752862558 Chr.15: 30635488-30635488 on Build GRCh38
rs767588718 Chr.15: 30635486-30635486 on Build GRCh38
rs776378363 Chr.15: 30635459-30635459 on Build GRCh38
rs975017695 Chr.15: 30635474-30635474 on Build GRCh38
rs377077753 Chr.15: 30635440-30635440 on Build GRCh38
rs982807770 Chr.15: 30635605-30635605 on Build GRCh38
rs759484298 Chr.15: 30635555-30635555 on Build GRCh38
rs373402272 Chr.15: 30635213-30635214 on Build GRCh38
rs371324595 Chr.15: 30635198-30635198 on Build GRCh38
rs1045262808 Chr.15: 30635401-30635401 on Build GRCh38
rs909851533 Chr.15: 30635389-30635389 on Build GRCh38
rs774091219 Chr.15: 30635545-30635545 on Build GRCh38
rs879196590 Chr.15: 30635534-30635534 on Build GRCh38
rs779335510 Chr.15: 30635614-30635614 on Build GRCh38
rs572627526 Chr.15: 30635267-30635267 on Build GRCh38
rs529739004 Chr.15: 30635392-30635392 on Build GRCh38
rs878886037 Chr.15: 30635332-30635332 on Build GRCh38
rs1054089794 Chr.15: 30635244-30635244 on Build GRCh38
rs751539539 Chr.15: 30635480-30635480 on Build GRCh38
rs780863495 Chr.15: 30635513-30635513 on Build GRCh38
rs966240126 Chr.15: 30635313-30635313 on Build GRCh38
rs974049026 Chr.15: 30635379-30635379 on Build GRCh38
rs762608471 Chr.15: 30635535-30635535 on Build GRCh38
rs766467386 Chr.15: 30635210-30635210 on Build GRCh38
rs747788745 Chr.15: 30635519-30635519 on Build GRCh38
rs1017922301 Chr.15: 30635503-30635503 on Build GRCh38
rs565559996 Chr.15: 30635471-30635471 on Build GRCh38
rs755011374 Chr.15: 30635483-30635483 on Build GRCh38
rs528237415 Chr.15: 30635638-30635638 on Build GRCh38
rs1024400211 Chr.15: 30635309-30635309 on Build GRCh38
rs746707373 Chr.15: 30635217-30635218 on Build GRCh38
rs770792015 Chr.15: 30635541-30635541 on Build GRCh38
rs768344412 Chr.15: 30635462-30635465 on Build GRCh38
rs775032103 Chr.15: 30635234-30635234 on Build GRCh38
rs769370376 Chr.15: 30635641-30635641 on Build GRCh38
rs376751790 Chr.15: 30635582-30635582 on Build GRCh38
rs769635666 Chr.15: 30635522-30635522 on Build GRCh38
rs748975355 Chr.15: 30635644-30635644 on Build GRCh38
rs900135475 Chr.15: 30635209-30635209 on Build GRCh38
rs1009288471 Chr.15: 30635444-30635444 on Build GRCh38
rs892204188 Chr.15: 30635443-30635443 on Build GRCh38
rs750307934 Chr.15: 30635193-30635193 on Build GRCh38
rs892028797 Chr.15: 30635265-30635265 on Build GRCh38
rs776094018 Chr.15: 30635497-30635498 on Build GRCh38
rs532312919 Chr.15: 30635438-30635438 on Build GRCh38
rs777252515 Chr.15: 30635642-30635642 on Build GRCh38
rs922659969 Chr.15: 30635351-30635351 on Build GRCh38

View Full Product Details