Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40411181-40411690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IVD
Entrez Gene ID
3712
Gene Name
isovaleryl-CoA dehydrogenase
Gene Aliases
ACAD2
UniGene
Hs.513646
Gene Chromosome Location
Chr.15: 40405485-40435948 on Build GRCh38
Transcripts
NM_001159508.1 NM_002225.3 XM_005254350.2 XM_006720492.2 XM_006720495.2 XM_017022149.1 XM_017022150.1 XM_017022151.1 XM_017022152.1 XM_017022153.1 XM_017022154.1 XM_017022155.1 XM_017022156.1 XM_017022157.1 XM_017022158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3886909 c.550G>A p.E184K Chr.15: 40411344-40411344 on Build GRCh38
6901272 c.680C>T p.A227V Chr.15: 40411675-40411675 on Build GRCh38
961197 c.620T>C p.V207A Chr.15: 40411615-40411615 on Build GRCh38
6142243 c.584A>G p.N195S Chr.15: 40411579-40411579 on Build GRCh38
6196461 c.647_655delTGGCTGCTG p.A217_V219delAAV Chr.15: 40411642-40411650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377147994 Chr.15: 40411354-40411354 on Build GRCh38
rs372448489 Chr.15: 40411371-40411371 on Build GRCh38
rs758187120 Chr.15: 40411280-40411280 on Build GRCh38
rs974139256 Chr.15: 40411347-40411347 on Build GRCh38
rs772915954 Chr.15: 40411424-40411424 on Build GRCh38
rs1033930341 Chr.15: 40411277-40411277 on Build GRCh38
rs757778820 Chr.15: 40411607-40411607 on Build GRCh38
rs756203719 Chr.15: 40411513-40411513 on Build GRCh38
rs766187968 Chr.15: 40411558-40411558 on Build GRCh38
rs557693409 Chr.15: 40411278-40411278 on Build GRCh38
rs762840448 Chr.15: 40411550-40411550 on Build GRCh38
rs372974651 Chr.15: 40411262-40411262 on Build GRCh38
rs767652612 Chr.15: 40411378-40411378 on Build GRCh38
rs574434706 Chr.15: 40411293-40411293 on Build GRCh38
rs142064071 Chr.15: 40411537-40411537 on Build GRCh38
rs559732306 Chr.15: 40411342-40411342 on Build GRCh38
rs36050927 Chr.15: 40411442-40411443 on Build GRCh38
rs1057517043 Chr.15: 40411554-40411554 on Build GRCh38
rs148601951 Chr.15: 40411527-40411527 on Build GRCh38
rs773402809 Chr.15: 40411245-40411245 on Build GRCh38
rs760362637 Chr.15: 40411338-40411338 on Build GRCh38
rs778330267 Chr.15: 40411212-40411212 on Build GRCh38
rs567853455 Chr.15: 40411504-40411504 on Build GRCh38
rs786204427 Chr.15: 40411257-40411258 on Build GRCh38
rs781630355 Chr.15: 40411621-40411621 on Build GRCh38
rs757727506 Chr.15: 40411520-40411520 on Build GRCh38
rs758559650 Chr.15: 40411210-40411210 on Build GRCh38
rs1034640945 Chr.15: 40411320-40411320 on Build GRCh38
rs376471632 Chr.15: 40411396-40411396 on Build GRCh38
rs398123684 Chr.15: 40411301-40411301 on Build GRCh38
rs1010351026 Chr.15: 40411266-40411267 on Build GRCh38
rs765156604 Chr.15: 40411349-40411349 on Build GRCh38
rs752939663 Chr.15: 40411579-40411579 on Build GRCh38
rs1033020481 Chr.15: 40411221-40411221 on Build GRCh38
rs543297907 Chr.15: 40411304-40411304 on Build GRCh38
rs754418427 Chr.15: 40411601-40411601 on Build GRCh38
rs761605034 Chr.15: 40411275-40411275 on Build GRCh38
rs1054846771 Chr.15: 40411486-40411486 on Build GRCh38
rs1011144878 Chr.15: 40411638-40411638 on Build GRCh38
rs759995695 Chr.15: 40411666-40411666 on Build GRCh38
rs759264185 Chr.15: 40411257-40411257 on Build GRCh38
rs755094829 Chr.15: 40411289-40411289 on Build GRCh38
rs780758214 Chr.15: 40411631-40411631 on Build GRCh38
rs4924466 Chr.15: 40411410-40411410 on Build GRCh38
rs752783697 Chr.15: 40411398-40411398 on Build GRCh38
rs756742372 Chr.15: 40411500-40411500 on Build GRCh38
rs532775974 Chr.15: 40411461-40411461 on Build GRCh38
rs762298810 Chr.15: 40411348-40411361 on Build GRCh38
rs563452741 Chr.15: 40411613-40411613 on Build GRCh38
rs749778050 Chr.15: 40411515-40411515 on Build GRCh38
rs966893680 Chr.15: 40411662-40411662 on Build GRCh38
rs773174399 Chr.15: 40411541-40411541 on Build GRCh38
rs8039298 Chr.15: 40411491-40411491 on Build GRCh38
rs140182461 Chr.15: 40411267-40411267 on Build GRCh38
rs944850552 Chr.15: 40411416-40411416 on Build GRCh38
rs749726739 Chr.15: 40411306-40411306 on Build GRCh38
rs745748728 Chr.15: 40411532-40411532 on Build GRCh38
rs751815296 Chr.15: 40411369-40411369 on Build GRCh38
rs768709744 Chr.15: 40411240-40411240 on Build GRCh38
rs1024133823 Chr.15: 40411283-40411283 on Build GRCh38
rs906650254 Chr.15: 40411514-40411514 on Build GRCh38
rs1021245627 Chr.15: 40411656-40411656 on Build GRCh38
rs551911271 Chr.15: 40411560-40411560 on Build GRCh38
rs748713834 Chr.15: 40411295-40411295 on Build GRCh38
rs751760095 Chr.15: 40411286-40411286 on Build GRCh38
rs369759141 Chr.15: 40411211-40411211 on Build GRCh38
rs771395584 Chr.15: 40411323-40411323 on Build GRCh38
rs775944806 Chr.15: 40411343-40411343 on Build GRCh38
rs962667792 Chr.15: 40411325-40411325 on Build GRCh38
rs747807491 Chr.15: 40411635-40411635 on Build GRCh38
rs995458380 Chr.15: 40411605-40411605 on Build GRCh38
rs767687961 Chr.15: 40411566-40411566 on Build GRCh38
rs1039203303 Chr.15: 40411418-40411418 on Build GRCh38
rs121434285 Chr.15: 40411600-40411600 on Build GRCh38
rs747158425 Chr.15: 40411228-40411228 on Build GRCh38
rs765782600 Chr.15: 40411507-40411542 on Build GRCh38
rs74011152 Chr.15: 40411496-40411496 on Build GRCh38
rs758778071 Chr.15: 40411627-40411627 on Build GRCh38
rs558189416 Chr.15: 40411614-40411614 on Build GRCh38
rs369030793 Chr.15: 40411356-40411356 on Build GRCh38
rs771914739 Chr.15: 40411258-40411258 on Build GRCh38
rs142289644 Chr.15: 40411412-40411412 on Build GRCh38
rs767644358 Chr.15: 40411395-40411395 on Build GRCh38
rs796051982 Chr.15: 40411272-40411273 on Build GRCh38
rs768125931 Chr.15: 40411531-40411531 on Build GRCh38
rs761815406 Chr.15: 40411348-40411348 on Build GRCh38
rs963770828 Chr.15: 40411664-40411664 on Build GRCh38
rs951022339 Chr.15: 40411616-40411616 on Build GRCh38
rs748577522 Chr.15: 40411243-40411243 on Build GRCh38
rs779371595 Chr.15: 40411522-40411522 on Build GRCh38
rs769221723 Chr.15: 40411637-40411637 on Build GRCh38
rs528252990 Chr.15: 40411540-40411540 on Build GRCh38
rs9635325 Chr.15: 40411402-40411402 on Build GRCh38
rs146360134 Chr.15: 40411436-40411436 on Build GRCh38
rs543060558 Chr.15: 40411474-40411474 on Build GRCh38
rs770227976 Chr.15: 40411244-40411244 on Build GRCh38
rs756184085 Chr.15: 40411580-40411580 on Build GRCh38
rs763665120 Chr.15: 40411260-40411260 on Build GRCh38
rs764191165 Chr.15: 40411598-40411598 on Build GRCh38
rs760307260 Chr.15: 40411259-40411259 on Build GRCh38
rs780313307 Chr.15: 40411282-40411282 on Build GRCh38
rs775318094 Chr.15: 40411337-40411337 on Build GRCh38

View Full Product Details