Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50354177-50354676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ10038
Entrez Gene ID
55056
Gene Name
uncharacterized protein FLJ10038
Gene Aliases
-
UniGene
Hs.511316
Gene Chromosome Location
Chr.15: 50348936-50354879 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

Gene Symbol
GABPB1
Entrez Gene ID
2553
Gene Name
GA binding protein transcription factor beta subunit 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
UniGene
Hs.654350
Gene Chromosome Location
Chr.15: 50277192-50355429 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

Gene Symbol
GABPB1-AS1
Entrez Gene ID
100129387
Gene Name
GABPB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659360
Gene Chromosome Location
Chr.15: 50354174-50358312 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

SNP Detail

SNP ID Chromosome Location
rs757358904 Chr.15: 50354428-50354428 on Build GRCh38
rs967409150 Chr.15: 50354599-50354599 on Build GRCh38
rs768420561 Chr.15: 50354415-50354415 on Build GRCh38
rs1018369306 Chr.15: 50354295-50354295 on Build GRCh38
rs577762378 Chr.15: 50354495-50354495 on Build GRCh38
rs542769059 Chr.15: 50354457-50354457 on Build GRCh38
rs754335588 Chr.15: 50354545-50354545 on Build GRCh38
rs749396173 Chr.15: 50354488-50354488 on Build GRCh38
rs945880443 Chr.15: 50354305-50354305 on Build GRCh38
rs533554918 Chr.15: 50354229-50354229 on Build GRCh38
rs1005868004 Chr.15: 50354472-50354472 on Build GRCh38
rs959080469 Chr.15: 50354540-50354540 on Build GRCh38
rs918859921 Chr.15: 50354314-50354314 on Build GRCh38
rs945991287 Chr.15: 50354426-50354426 on Build GRCh38
rs556424290 Chr.15: 50354463-50354463 on Build GRCh38
rs11537688 Chr.15: 50354578-50354578 on Build GRCh38
rs963315142 Chr.15: 50354357-50354357 on Build GRCh38
rs761274276 Chr.15: 50354563-50354563 on Build GRCh38
rs985502954 Chr.15: 50354287-50354287 on Build GRCh38
rs771012878 Chr.15: 50354496-50354496 on Build GRCh38
rs914489715 Chr.15: 50354419-50354419 on Build GRCh38
rs750110967 Chr.15: 50354588-50354588 on Build GRCh38
rs772567322 Chr.15: 50354528-50354528 on Build GRCh38
rs758215730 Chr.15: 50354365-50354365 on Build GRCh38
rs923070854 Chr.15: 50354428-50354428 on Build GRCh38
rs774090012 Chr.15: 50354620-50354620 on Build GRCh38
rs754931968 Chr.15: 50354299-50354300 on Build GRCh38
rs369291578 Chr.15: 50354557-50354557 on Build GRCh38
rs949243484 Chr.15: 50354402-50354402 on Build GRCh38
rs573824340 Chr.15: 50354635-50354635 on Build GRCh38
rs754798936 Chr.15: 50354622-50354622 on Build GRCh38
rs572259026 Chr.15: 50354538-50354538 on Build GRCh38
rs1054833941 Chr.15: 50354386-50354386 on Build GRCh38
rs888759985 Chr.15: 50354344-50354344 on Build GRCh38
rs965571034 Chr.15: 50354311-50354311 on Build GRCh38
rs941695288 Chr.15: 50354380-50354380 on Build GRCh38
rs753960782 Chr.15: 50354345-50354353 on Build GRCh38
rs1018399876 Chr.15: 50354502-50354502 on Build GRCh38
rs781032024 Chr.15: 50354623-50354623 on Build GRCh38
rs544296975 Chr.15: 50354654-50354654 on Build GRCh38
rs866045772 Chr.15: 50354361-50354361 on Build GRCh38
rs916347764 Chr.15: 50354596-50354596 on Build GRCh38
rs965435497 Chr.15: 50354516-50354516 on Build GRCh38
rs997190575 Chr.15: 50354329-50354329 on Build GRCh38
rs763670272 Chr.15: 50354298-50354298 on Build GRCh38
rs1017173633 Chr.15: 50354353-50354353 on Build GRCh38
rs981531693 Chr.15: 50354234-50354234 on Build GRCh38
rs1046206021 Chr.15: 50354416-50354416 on Build GRCh38
rs933175110 Chr.15: 50354445-50354445 on Build GRCh38
rs902441741 Chr.15: 50354438-50354438 on Build GRCh38
rs1037811786 Chr.15: 50354485-50354485 on Build GRCh38
rs376761536 Chr.15: 50354616-50354616 on Build GRCh38
rs976938802 Chr.15: 50354523-50354523 on Build GRCh38
rs976905926 Chr.15: 50354312-50354312 on Build GRCh38
rs537925296 Chr.15: 50354462-50354462 on Build GRCh38
rs930222034 Chr.15: 50354326-50354326 on Build GRCh38
rs756783365 Chr.15: 50354322-50354322 on Build GRCh38
rs779840805 Chr.15: 50354613-50354613 on Build GRCh38
rs561730769 Chr.15: 50354600-50354600 on Build GRCh38
rs981500528 Chr.15: 50354392-50354392 on Build GRCh38
rs567387615 Chr.15: 50354370-50354370 on Build GRCh38
rs3825796 Chr.15: 50354358-50354358 on Build GRCh38
rs937356133 Chr.15: 50354251-50354251 on Build GRCh38
rs770946754 Chr.15: 50354638-50354638 on Build GRCh38
rs901596189 Chr.15: 50354327-50354327 on Build GRCh38
rs764573929 Chr.15: 50354544-50354544 on Build GRCh38
rs1013363693 Chr.15: 50354198-50354198 on Build GRCh38
rs553793422 Chr.15: 50354529-50354529 on Build GRCh38
rs772965670 Chr.15: 50354618-50354618 on Build GRCh38
rs548970070 Chr.15: 50354283-50354283 on Build GRCh38
rs774736676 Chr.15: 50354507-50354507 on Build GRCh38
rs941727865 Chr.15: 50354566-50354566 on Build GRCh38
rs1025919611 Chr.15: 50354521-50354521 on Build GRCh38
rs753344019 Chr.15: 50354307-50354307 on Build GRCh38
rs746955878 Chr.15: 50354371-50354371 on Build GRCh38
rs1006889011 Chr.15: 50354497-50354497 on Build GRCh38
rs894883070 Chr.15: 50354401-50354401 on Build GRCh38
rs893715554 Chr.15: 50354656-50354656 on Build GRCh38
rs1024874716 Chr.15: 50354202-50354202 on Build GRCh38
rs552027960 Chr.15: 50354322-50354322 on Build GRCh38
rs1046667150 Chr.15: 50354601-50354601 on Build GRCh38
rs745501432 Chr.15: 50354350-50354350 on Build GRCh38
rs971558173 Chr.15: 50354389-50354389 on Build GRCh38
rs778467125 Chr.15: 50354334-50354334 on Build GRCh38
rs958370395 Chr.15: 50354259-50354259 on Build GRCh38
rs538964833 Chr.15: 50354524-50354524 on Build GRCh38
rs753286060 Chr.15: 50354241-50354241 on Build GRCh38
rs999414464 Chr.15: 50354442-50354442 on Build GRCh38
rs756510827 Chr.15: 50354275-50354275 on Build GRCh38
rs918657277 Chr.15: 50354199-50354199 on Build GRCh38
rs8552 Chr.15: 50354586-50354586 on Build GRCh38
rs958719069 Chr.15: 50354414-50354414 on Build GRCh38
rs933142654 Chr.15: 50354602-50354602 on Build GRCh38
rs926850441 Chr.15: 50354383-50354383 on Build GRCh38
rs747972297 Chr.15: 50354249-50354249 on Build GRCh38
rs746099945 Chr.15: 50354659-50354659 on Build GRCh38
rs1054349029 Chr.15: 50354294-50354294 on Build GRCh38
rs3743043 Chr.15: 50354214-50354214 on Build GRCh38

View Full Product Details