Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50354564-50355101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ10038
Entrez Gene ID
55056
Gene Name
uncharacterized protein FLJ10038
Gene Aliases
-
UniGene
Hs.511316
Gene Chromosome Location
Chr.15: 50348936-50354879 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

Gene Symbol
GABPB1
Entrez Gene ID
2553
Gene Name
GA binding protein transcription factor beta subunit 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2
UniGene
Hs.654350
Gene Chromosome Location
Chr.15: 50277192-50355429 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

Gene Symbol
GABPB1-AS1
Entrez Gene ID
100129387
Gene Name
GABPB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659360
Gene Chromosome Location
Chr.15: 50354174-50358312 on Build GRCh38
Transcripts
NR_026891.1 NM_001320910.1 NM_001320915.1 NM_002041.4 NM_005254.5 NM_016654.4 NM_016655.4 NM_181427.3 XM_011521426.2 XM_017022053.1 XM_017022054.1 NR_024490.1

SNP Detail

SNP ID Chromosome Location
rs989343453 Chr.15: 50354928-50354928 on Build GRCh38
rs967409150 Chr.15: 50354599-50354599 on Build GRCh38
rs942598772 Chr.15: 50354967-50354967 on Build GRCh38
rs919040373 Chr.15: 50354813-50354813 on Build GRCh38
rs1047480893 Chr.15: 50354981-50354981 on Build GRCh38
rs1012037515 Chr.15: 50354785-50354785 on Build GRCh38
rs1043271061 Chr.15: 50355036-50355036 on Build GRCh38
rs113692002 Chr.15: 50354703-50354703 on Build GRCh38
rs1013082016 Chr.15: 50354854-50354854 on Build GRCh38
rs750110967 Chr.15: 50354588-50354588 on Build GRCh38
rs975369253 Chr.15: 50354970-50354970 on Build GRCh38
rs533367993 Chr.15: 50355009-50355009 on Build GRCh38
rs1021731235 Chr.15: 50354786-50354786 on Build GRCh38
rs774090012 Chr.15: 50354620-50354620 on Build GRCh38
rs935207982 Chr.15: 50354763-50354763 on Build GRCh38
rs765328944 Chr.15: 50354839-50354839 on Build GRCh38
rs1024542455 Chr.15: 50355042-50355042 on Build GRCh38
rs758910796 Chr.15: 50354879-50354879 on Build GRCh38
rs1013029776 Chr.15: 50355041-50355041 on Build GRCh38
rs573824340 Chr.15: 50354635-50354635 on Build GRCh38
rs915045253 Chr.15: 50354933-50354933 on Build GRCh38
rs754798936 Chr.15: 50354622-50354622 on Build GRCh38
rs998810819 Chr.15: 50354857-50354857 on Build GRCh38
rs934029096 Chr.15: 50354975-50354975 on Build GRCh38
rs942567371 Chr.15: 50354783-50354783 on Build GRCh38
rs781032024 Chr.15: 50354623-50354623 on Build GRCh38
rs544296975 Chr.15: 50354654-50354654 on Build GRCh38
rs36062201 Chr.15: 50355061-50355061 on Build GRCh38
rs967756041 Chr.15: 50354790-50354790 on Build GRCh38
rs916347764 Chr.15: 50354596-50354596 on Build GRCh38
rs1003037656 Chr.15: 50354712-50354712 on Build GRCh38
rs1031057110 Chr.15: 50355059-50355059 on Build GRCh38
rs773967341 Chr.15: 50354829-50354829 on Build GRCh38
rs1047169358 Chr.15: 50354799-50354799 on Build GRCh38
rs759460080 Chr.15: 50354763-50354764 on Build GRCh38
rs1034963028 Chr.15: 50354764-50354764 on Build GRCh38
rs887211265 Chr.15: 50354997-50354997 on Build GRCh38
rs907444462 Chr.15: 50354832-50354832 on Build GRCh38
rs956637415 Chr.15: 50354923-50354923 on Build GRCh38
rs376761536 Chr.15: 50354616-50354616 on Build GRCh38
rs938878120 Chr.15: 50354833-50354833 on Build GRCh38
rs1038504505 Chr.15: 50355020-50355020 on Build GRCh38
rs1011760725 Chr.15: 50354852-50354852 on Build GRCh38
rs747239747 Chr.15: 50354678-50354678 on Build GRCh38
rs1017162652 Chr.15: 50354696-50354696 on Build GRCh38
rs981923107 Chr.15: 50354740-50354740 on Build GRCh38
rs993175840 Chr.15: 50354683-50354683 on Build GRCh38
rs901137101 Chr.15: 50354789-50354789 on Build GRCh38
rs958912176 Chr.15: 50354767-50354767 on Build GRCh38
rs779840805 Chr.15: 50354613-50354613 on Build GRCh38
rs1037458755 Chr.15: 50354676-50354676 on Build GRCh38
rs1054011002 Chr.15: 50354765-50354765 on Build GRCh38
rs561730769 Chr.15: 50354600-50354600 on Build GRCh38
rs894614049 Chr.15: 50354851-50354851 on Build GRCh38
rs989192179 Chr.15: 50355076-50355076 on Build GRCh38
rs772477231 Chr.15: 50354677-50354677 on Build GRCh38
rs951610550 Chr.15: 50354692-50354692 on Build GRCh38
rs770946754 Chr.15: 50354638-50354638 on Build GRCh38
rs760981711 Chr.15: 50354693-50354693 on Build GRCh38
rs963601109 Chr.15: 50354798-50354799 on Build GRCh38
rs918997398 Chr.15: 50354977-50354977 on Build GRCh38
rs1039735832 Chr.15: 50354784-50354784 on Build GRCh38
rs775243591 Chr.15: 50355033-50355033 on Build GRCh38
rs894757600 Chr.15: 50355028-50355028 on Build GRCh38
rs772965670 Chr.15: 50354618-50354618 on Build GRCh38
rs779059378 Chr.15: 50354713-50354714 on Build GRCh38
rs770766895 Chr.15: 50354805-50354805 on Build GRCh38
rs973946381 Chr.15: 50354811-50354820 on Build GRCh38
rs991562179 Chr.15: 50354803-50354803 on Build GRCh38
rs958922597 Chr.15: 50354697-50354697 on Build GRCh38
rs767203026 Chr.15: 50355000-50355000 on Build GRCh38
rs903268127 Chr.15: 50355047-50355047 on Build GRCh38
rs954718421 Chr.15: 50354882-50354882 on Build GRCh38
rs776189026 Chr.15: 50355053-50355053 on Build GRCh38
rs986429415 Chr.15: 50354914-50354914 on Build GRCh38
rs1017027252 Chr.15: 50354818-50354818 on Build GRCh38
rs777029845 Chr.15: 50354738-50354738 on Build GRCh38
rs1026238202 Chr.15: 50354689-50354689 on Build GRCh38
rs901272790 Chr.15: 50354681-50354681 on Build GRCh38
rs956491989 Chr.15: 50355072-50355072 on Build GRCh38
rs893715554 Chr.15: 50354656-50354656 on Build GRCh38
rs916212184 Chr.15: 50354709-50354709 on Build GRCh38
rs958955159 Chr.15: 50354828-50354828 on Build GRCh38
rs551902082 Chr.15: 50355015-50355015 on Build GRCh38
rs907431027 Chr.15: 50355045-50355045 on Build GRCh38
rs927576593 Chr.15: 50355004-50355004 on Build GRCh38
rs982172562 Chr.15: 50354883-50354883 on Build GRCh38
rs865775910 Chr.15: 50354791-50354791 on Build GRCh38
rs998592890 Chr.15: 50354798-50354798 on Build GRCh38
rs1024509857 Chr.15: 50354856-50354856 on Build GRCh38
rs1046667150 Chr.15: 50354601-50354601 on Build GRCh38
rs1056423601 Chr.15: 50355027-50355027 on Build GRCh38
rs928936030 Chr.15: 50354817-50354817 on Build GRCh38
rs1006919963 Chr.15: 50354663-50354663 on Build GRCh38
rs1056056670 Chr.15: 50354842-50354842 on Build GRCh38
rs1022117074 Chr.15: 50354853-50354853 on Build GRCh38
rs887170210 Chr.15: 50354800-50354800 on Build GRCh38
rs1039704496 Chr.15: 50354673-50354673 on Build GRCh38
rs915181254 Chr.15: 50354770-50354770 on Build GRCh38
rs922498564 Chr.15: 50354972-50354972 on Build GRCh38
rs1003230338 Chr.15: 50355057-50355057 on Build GRCh38
rs35689237 Chr.15: 50354949-50354950 on Build GRCh38
rs776902158 Chr.15: 50354760-50354760 on Build GRCh38
rs923737571 Chr.15: 50354895-50354895 on Build GRCh38
rs970453915 Chr.15: 50354724-50354724 on Build GRCh38
rs8552 Chr.15: 50354586-50354586 on Build GRCh38
rs775847453 Chr.15: 50354687-50354687 on Build GRCh38
rs933142654 Chr.15: 50354602-50354602 on Build GRCh38
rs923679344 Chr.15: 50354754-50354754 on Build GRCh38
rs1030391311 Chr.15: 50355060-50355060 on Build GRCh38
rs894686679 Chr.15: 50355035-50355035 on Build GRCh38
rs746099945 Chr.15: 50354659-50354659 on Build GRCh38

View Full Product Details