Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52121532-52122074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB5
Entrez Gene ID
10681
Gene Name
G protein subunit beta 5
Gene Aliases
GB5
UniGene
Hs.155090
Gene Chromosome Location
Chr.15: 52120926-52191368 on Build GRCh38
Transcripts
NM_006578.3 NM_016194.3 XM_011521162.2 XM_011521163.2 XM_017021867.1

SNP Detail

SNP ID Chromosome Location
rs985729744 Chr.15: 52121856-52121856 on Build GRCh38
rs758537144 Chr.15: 52121999-52121999 on Build GRCh38
rs562823284 Chr.15: 52121665-52121665 on Build GRCh38
rs992274672 Chr.15: 52121696-52121696 on Build GRCh38
rs192448111 Chr.15: 52121704-52121704 on Build GRCh38
rs564219135 Chr.15: 52121610-52121611 on Build GRCh38
rs1026415413 Chr.15: 52121793-52121793 on Build GRCh38
rs148106165 Chr.15: 52121883-52121883 on Build GRCh38
rs188981291 Chr.15: 52121872-52121872 on Build GRCh38
rs531105373 Chr.15: 52121672-52121672 on Build GRCh38
rs911063508 Chr.15: 52121869-52121869 on Build GRCh38
rs972192721 Chr.15: 52121643-52121643 on Build GRCh38
rs539217852 Chr.15: 52121742-52121742 on Build GRCh38
rs982420253 Chr.15: 52121681-52121681 on Build GRCh38
rs557395013 Chr.15: 52121765-52121765 on Build GRCh38
rs543993425 Chr.15: 52122019-52122019 on Build GRCh38
rs975395682 Chr.15: 52121823-52121823 on Build GRCh38
rs920031900 Chr.15: 52121832-52121832 on Build GRCh38
rs557667640 Chr.15: 52121839-52121839 on Build GRCh38
rs964071901 Chr.15: 52121699-52121699 on Build GRCh38
rs370758308 Chr.15: 52121728-52121728 on Build GRCh38
rs533874836 Chr.15: 52121878-52121878 on Build GRCh38
rs925056347 Chr.15: 52121844-52121844 on Build GRCh38
rs918957090 Chr.15: 52121649-52121649 on Build GRCh38
rs776773129 Chr.15: 52121881-52121881 on Build GRCh38
rs554950687 Chr.15: 52121927-52121927 on Build GRCh38
rs71130132 Chr.15: 52121625-52121626 on Build GRCh38
rs563745212 Chr.15: 52121615-52121615 on Build GRCh38
rs901959937 Chr.15: 52121639-52121639 on Build GRCh38
rs8024767 Chr.15: 52121984-52121984 on Build GRCh38
rs3205246 Chr.15: 52121778-52121778 on Build GRCh38
rs936429026 Chr.15: 52121846-52121846 on Build GRCh38
rs147515144 Chr.15: 52121760-52121760 on Build GRCh38
rs984949081 Chr.15: 52121671-52121671 on Build GRCh38
rs1025157989 Chr.15: 52121834-52121834 on Build GRCh38
rs1042847833 Chr.15: 52121739-52121739 on Build GRCh38
rs929322137 Chr.15: 52121949-52121949 on Build GRCh38
rs539602431 Chr.15: 52121806-52121806 on Build GRCh38
rs1033850765 Chr.15: 52121814-52121814 on Build GRCh38
rs889738996 Chr.15: 52121732-52121732 on Build GRCh38
rs926256074 Chr.15: 52121992-52121992 on Build GRCh38
rs1016012319 Chr.15: 52121630-52121630 on Build GRCh38
rs985934344 Chr.15: 52121604-52121608 on Build GRCh38
rs1041004035 Chr.15: 52121884-52121884 on Build GRCh38
rs938259664 Chr.15: 52122003-52122003 on Build GRCh38
rs1016061635 Chr.15: 52121849-52121849 on Build GRCh38
rs959632497 Chr.15: 52121693-52121693 on Build GRCh38
rs17540546 Chr.15: 52122038-52122038 on Build GRCh38
rs959923954 Chr.15: 52121817-52121817 on Build GRCh38
rs1049561555 Chr.15: 52121729-52121729 on Build GRCh38
rs971861636 Chr.15: 52121893-52121893 on Build GRCh38
rs1007994234 Chr.15: 52121610-52121610 on Build GRCh38
rs970470902 Chr.15: 52121983-52121983 on Build GRCh38
rs575916318 Chr.15: 52121764-52121764 on Build GRCh38
rs1047745010 Chr.15: 52121979-52121979 on Build GRCh38
rs531046902 Chr.15: 52122036-52122036 on Build GRCh38
rs184664169 Chr.15: 52121847-52121847 on Build GRCh38
rs547014456 Chr.15: 52121740-52121740 on Build GRCh38
rs3208908 Chr.15: 52121779-52121779 on Build GRCh38
rs752120007 Chr.15: 52121998-52122000 on Build GRCh38
rs192930127 Chr.15: 52122024-52122024 on Build GRCh38
rs1026365078 Chr.15: 52121666-52121666 on Build GRCh38
rs1030201595 Chr.15: 52121557-52121560 on Build GRCh38
rs551161957 Chr.15: 52121744-52121744 on Build GRCh38
rs3208909 Chr.15: 52121777-52121777 on Build GRCh38
rs989718971 Chr.15: 52122027-52122027 on Build GRCh38
rs552932103 Chr.15: 52121703-52121703 on Build GRCh38
rs992430937 Chr.15: 52121827-52121827 on Build GRCh38
rs966361582 Chr.15: 52121835-52121835 on Build GRCh38
rs187882116 Chr.15: 52121570-52121570 on Build GRCh38
rs578075817 Chr.15: 52121669-52121669 on Build GRCh38
rs889695901 Chr.15: 52121593-52121593 on Build GRCh38
rs566186163 Chr.15: 52121805-52121805 on Build GRCh38
rs993476483 Chr.15: 52121772-52121772 on Build GRCh38
rs977828646 Chr.15: 52121721-52121721 on Build GRCh38
rs925004015 Chr.15: 52121726-52121726 on Build GRCh38

View Full Product Details