Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52121916-52122457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB5
Entrez Gene ID
10681
Gene Name
G protein subunit beta 5
Gene Aliases
GB5
UniGene
Hs.155090
Gene Chromosome Location
Chr.15: 52120926-52191368 on Build GRCh38
Transcripts
NM_006578.3 NM_016194.3 XM_011521162.2 XM_011521163.2 XM_017021867.1

SNP Detail

SNP ID Chromosome Location
rs758537144 Chr.15: 52121999-52121999 on Build GRCh38
rs938259664 Chr.15: 52122003-52122003 on Build GRCh38
rs1031996779 Chr.15: 52122202-52122202 on Build GRCh38
rs1049736126 Chr.15: 52122181-52122181 on Build GRCh38
rs895320204 Chr.15: 52122094-52122094 on Build GRCh38
rs957906326 Chr.15: 52122236-52122236 on Build GRCh38
rs966914435 Chr.15: 52122164-52122164 on Build GRCh38
rs910941563 Chr.15: 52122299-52122299 on Build GRCh38
rs569322786 Chr.15: 52122391-52122391 on Build GRCh38
rs17540546 Chr.15: 52122038-52122038 on Build GRCh38
rs970470902 Chr.15: 52121983-52121983 on Build GRCh38
rs1037561611 Chr.15: 52122229-52122230 on Build GRCh38
rs531046902 Chr.15: 52122036-52122036 on Build GRCh38
rs1047745010 Chr.15: 52121979-52121979 on Build GRCh38
rs543993425 Chr.15: 52122019-52122019 on Build GRCh38
rs550826410 Chr.15: 52122380-52122380 on Build GRCh38
rs945535119 Chr.15: 52122065-52122065 on Build GRCh38
rs922685911 Chr.15: 52122163-52122163 on Build GRCh38
rs889753083 Chr.15: 52122208-52122208 on Build GRCh38
rs192930127 Chr.15: 52122024-52122024 on Build GRCh38
rs752120007 Chr.15: 52121998-52122000 on Build GRCh38
rs561957187 Chr.15: 52122351-52122351 on Build GRCh38
rs547275513 Chr.15: 52122152-52122152 on Build GRCh38
rs918532957 Chr.15: 52122373-52122373 on Build GRCh38
rs1033374282 Chr.15: 52122367-52122367 on Build GRCh38
rs79339279 Chr.15: 52122355-52122355 on Build GRCh38
rs1025208955 Chr.15: 52122150-52122150 on Build GRCh38
rs899035342 Chr.15: 52122294-52122294 on Build GRCh38
rs184686820 Chr.15: 52122130-52122130 on Build GRCh38
rs985227224 Chr.15: 52122296-52122296 on Build GRCh38
rs999186293 Chr.15: 52122195-52122195 on Build GRCh38
rs989718971 Chr.15: 52122027-52122027 on Build GRCh38
rs766212828 Chr.15: 52122227-52122227 on Build GRCh38
rs80351019 Chr.15: 52122194-52122194 on Build GRCh38
rs1026109990 Chr.15: 52122341-52122341 on Build GRCh38
rs8024767 Chr.15: 52121984-52121984 on Build GRCh38
rs866136732 Chr.15: 52122122-52122122 on Build GRCh38
rs1047795491 Chr.15: 52122423-52122423 on Build GRCh38
rs1013677710 Chr.15: 52122142-52122142 on Build GRCh38
rs74686287 Chr.15: 52122105-52122105 on Build GRCh38
rs970904232 Chr.15: 52122343-52122343 on Build GRCh38
rs929322137 Chr.15: 52121949-52121949 on Build GRCh38
rs564817503 Chr.15: 52122100-52122100 on Build GRCh38
rs926256074 Chr.15: 52121992-52121992 on Build GRCh38
rs1007458517 Chr.15: 52122216-52122216 on Build GRCh38

View Full Product Details