Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55355112-55355658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCPG1
Entrez Gene ID
9236
Gene Name
cell cycle progression 1
Gene Aliases
CPR8
UniGene
Hs.285051
Gene Chromosome Location
Chr.15: 55355223-55408510 on Build GRCh38
Transcripts
NM_001204450.1 NM_001204451.1 NM_004748.4 NM_020739.3 NR_037923.1 NM_004855.4 XM_005254795.4 XM_011522235.2 XM_011522236.2 XM_011522237.2 XM_017022730.1 XM_017022731.1 XM_017022732.1

Gene Symbol
DYX1C1-CCPG1
Entrez Gene ID
100533483
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.285051
Gene Chromosome Location
Chr.15: 55355223-55498584 on Build GRCh38
Transcripts
NM_001204450.1 NM_001204451.1 NM_004748.4 NM_020739.3 NR_037923.1 NM_004855.4 XM_005254795.4 XM_011522235.2 XM_011522236.2 XM_011522237.2 XM_017022730.1 XM_017022731.1 XM_017022732.1

Gene Symbol
PIGB
Entrez Gene ID
9488
Gene Name
phosphatidylinositol glycan anchor biosynthesis class B
Gene Aliases
GPI-MT-III, PIG-B
UniGene
Hs.612814
Gene Chromosome Location
Chr.15: 55318935-55355648 on Build GRCh38
Transcripts
NM_001204450.1 NM_001204451.1 NM_004748.4 NM_020739.3 NR_037923.1 NM_004855.4 XM_005254795.4 XM_011522235.2 XM_011522236.2 XM_011522237.2 XM_017022730.1 XM_017022731.1 XM_017022732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422569 c.1583T>A p.L528* Chr.15: 55355350-55355350 on Build GRCh38
6213132 c.1637G>T p.G546V Chr.15: 55355404-55355404 on Build GRCh38
6746041 c.1653G>A p.K551K Chr.15: 55355420-55355420 on Build GRCh38
7201900 c.1547A>G p.N516S Chr.15: 55355314-55355314 on Build GRCh38
4935442 c.1588G>A p.E530K Chr.15: 55355355-55355355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982380026 Chr.15: 55355223-55355223 on Build GRCh38
rs2444042 Chr.15: 55355419-55355419 on Build GRCh38
rs756358630 Chr.15: 55355414-55355414 on Build GRCh38
rs367927367 Chr.15: 55355361-55355361 on Build GRCh38
rs553776058 Chr.15: 55355219-55355219 on Build GRCh38
rs770336696 Chr.15: 55355564-55355564 on Build GRCh38
rs765017632 Chr.15: 55355391-55355391 on Build GRCh38
rs774234795 Chr.15: 55355392-55355392 on Build GRCh38
rs745934716 Chr.15: 55355162-55355163 on Build GRCh38
rs753741353 Chr.15: 55355264-55355264 on Build GRCh38
rs776068686 Chr.15: 55355386-55355386 on Build GRCh38
rs769773951 Chr.15: 55355376-55355377 on Build GRCh38
rs973581925 Chr.15: 55355159-55355159 on Build GRCh38
rs1034849650 Chr.15: 55355236-55355236 on Build GRCh38
rs973531223 Chr.15: 55355561-55355561 on Build GRCh38
rs760160969 Chr.15: 55355263-55355263 on Build GRCh38
rs916315006 Chr.15: 55355171-55355171 on Build GRCh38
rs574364354 Chr.15: 55355228-55355228 on Build GRCh38
rs778049348 Chr.15: 55355341-55355341 on Build GRCh38
rs191218911 Chr.15: 55355492-55355492 on Build GRCh38
rs553633768 Chr.15: 55355252-55355252 on Build GRCh38
rs377008200 Chr.15: 55355380-55355380 on Build GRCh38
rs746450890 Chr.15: 55355437-55355437 on Build GRCh38
rs2444041 Chr.15: 55355265-55355265 on Build GRCh38
rs775218163 Chr.15: 55355248-55355248 on Build GRCh38
rs746015165 Chr.15: 55355399-55355399 on Build GRCh38
rs1038765695 Chr.15: 55355507-55355507 on Build GRCh38
rs753034934 Chr.15: 55355297-55355297 on Build GRCh38
rs919526409 Chr.15: 55355385-55355385 on Build GRCh38
rs982270650 Chr.15: 55355631-55355631 on Build GRCh38
rs539834403 Chr.15: 55355170-55355170 on Build GRCh38
rs947837999 Chr.15: 55355233-55355233 on Build GRCh38
rs371775367 Chr.15: 55355376-55355376 on Build GRCh38
rs750144952 Chr.15: 55355289-55355289 on Build GRCh38
rs1010564754 Chr.15: 55355583-55355583 on Build GRCh38
rs1023773442 Chr.15: 55355575-55355575 on Build GRCh38
rs758656192 Chr.15: 55355443-55355443 on Build GRCh38
rs375047051 Chr.15: 55355312-55355312 on Build GRCh38
rs746244095 Chr.15: 55355525-55355525 on Build GRCh38
rs370075431 Chr.15: 55355463-55355463 on Build GRCh38
rs753906457 Chr.15: 55355423-55355423 on Build GRCh38
rs747395128 Chr.15: 55355475-55355475 on Build GRCh38
rs747586835 Chr.15: 55355532-55355532 on Build GRCh38
rs11732 Chr.15: 55355588-55355588 on Build GRCh38
rs565194380 Chr.15: 55355393-55355393 on Build GRCh38
rs879775251 Chr.15: 55355300-55355300 on Build GRCh38
rs772274694 Chr.15: 55355458-55355461 on Build GRCh38
rs757376460 Chr.15: 55355425-55355425 on Build GRCh38
rs991591036 Chr.15: 55355281-55355281 on Build GRCh38
rs374303086 Chr.15: 55355397-55355397 on Build GRCh38
rs369616294 Chr.15: 55355382-55355382 on Build GRCh38
rs775040015 Chr.15: 55355387-55355387 on Build GRCh38
rs761342771 Chr.15: 55355390-55355390 on Build GRCh38
rs749813222 Chr.15: 55355589-55355589 on Build GRCh38
rs369915830 Chr.15: 55355279-55355279 on Build GRCh38
rs746164541 Chr.15: 55355372-55355372 on Build GRCh38
rs1046019122 Chr.15: 55355262-55355262 on Build GRCh38
rs1020602005 Chr.15: 55355586-55355586 on Build GRCh38
rs560980266 Chr.15: 55355435-55355435 on Build GRCh38
rs779483306 Chr.15: 55355362-55355362 on Build GRCh38
rs8043415 Chr.15: 55355294-55355294 on Build GRCh38
rs781259075 Chr.15: 55355295-55355295 on Build GRCh38

View Full Product Details