Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64380866-64381367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0101
Entrez Gene ID
9768
Gene Name
KIAA0101
Gene Aliases
L5, NS5ATP9, OEATC, OEATC-1, OEATC1, PAF, PAF15, p15(PAF), p15/PAF, p15PAF
UniGene
Hs.81892
Gene Chromosome Location
Chr.15: 64364994-64387687 on Build GRCh38
Transcripts
NM_001029989.2 NM_014736.5 NR_109934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692968 c.28C>A p.P10T Chr.15: 64381344-64381344 on Build GRCh38
4695133 c.7C>T p.R3W Chr.15: 64381365-64381365 on Build GRCh38
3503049 c.28C>T p.P10S Chr.15: 64381344-64381344 on Build GRCh38
6464014 c.108A>G p.T36T Chr.15: 64380977-64380977 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs188268476 Chr.15: 64381318-64381318 on Build GRCh38
rs540984263 Chr.15: 64381264-64381264 on Build GRCh38
rs751140531 Chr.15: 64380920-64380920 on Build GRCh38
rs553888960 Chr.15: 64381109-64381109 on Build GRCh38
rs760555818 Chr.15: 64381044-64381045 on Build GRCh38
rs754525854 Chr.15: 64381036-64381036 on Build GRCh38
rs895099515 Chr.15: 64381258-64381259 on Build GRCh38
rs987037573 Chr.15: 64381297-64381297 on Build GRCh38
rs1002493153 Chr.15: 64381180-64381180 on Build GRCh38
rs1044161588 Chr.15: 64381320-64381320 on Build GRCh38
rs1034830089 Chr.15: 64381093-64381093 on Build GRCh38
rs1002418839 Chr.15: 64380937-64380937 on Build GRCh38
rs769242466 Chr.15: 64381073-64381073 on Build GRCh38
rs772637057 Chr.15: 64380954-64380954 on Build GRCh38
rs575819041 Chr.15: 64381204-64381204 on Build GRCh38
rs779357833 Chr.15: 64380946-64380946 on Build GRCh38
rs528002964 Chr.15: 64381012-64381012 on Build GRCh38
rs905328012 Chr.15: 64381132-64381132 on Build GRCh38
rs750482897 Chr.15: 64381286-64381286 on Build GRCh38
rs932576154 Chr.15: 64381072-64381072 on Build GRCh38
rs367671592 Chr.15: 64380988-64380988 on Build GRCh38
rs1003442552 Chr.15: 64381081-64381081 on Build GRCh38
rs138161344 Chr.15: 64381051-64381051 on Build GRCh38
rs561818575 Chr.15: 64381239-64381239 on Build GRCh38
rs879126183 Chr.15: 64381293-64381293 on Build GRCh38
rs767046896 Chr.15: 64381037-64381037 on Build GRCh38
rs541184581 Chr.15: 64381277-64381277 on Build GRCh38
rs780755338 Chr.15: 64381063-64381063 on Build GRCh38
rs990543567 Chr.15: 64381101-64381101 on Build GRCh38
rs375121530 Chr.15: 64381341-64381341 on Build GRCh38
rs911354479 Chr.15: 64381136-64381136 on Build GRCh38
rs112539029 Chr.15: 64381031-64381031 on Build GRCh38
rs201419596 Chr.15: 64381274-64381274 on Build GRCh38
rs752279175 Chr.15: 64381230-64381230 on Build GRCh38
rs758835769 Chr.15: 64381338-64381338 on Build GRCh38
rs187410301 Chr.15: 64381066-64381066 on Build GRCh38
rs751479594 Chr.15: 64381033-64381033 on Build GRCh38
rs1041687319 Chr.15: 64381222-64381222 on Build GRCh38
rs921127878 Chr.15: 64381247-64381247 on Build GRCh38
rs146271324 Chr.15: 64381048-64381048 on Build GRCh38
rs190689392 Chr.15: 64381116-64381116 on Build GRCh38
rs28504999 Chr.15: 64381094-64381094 on Build GRCh38
rs943004870 Chr.15: 64381305-64381305 on Build GRCh38
rs770605302 Chr.15: 64381291-64381291 on Build GRCh38
rs1004633661 Chr.15: 64381194-64381194 on Build GRCh38
rs747141479 Chr.15: 64380998-64380998 on Build GRCh38
rs946157792 Chr.15: 64381218-64381218 on Build GRCh38
rs1051443174 Chr.15: 64381087-64381087 on Build GRCh38
rs767322024 Chr.15: 64381099-64381099 on Build GRCh38
rs1022444400 Chr.15: 64380908-64380908 on Build GRCh38
rs552887085 Chr.15: 64381278-64381278 on Build GRCh38
rs534342023 Chr.15: 64381131-64381131 on Build GRCh38
rs750864583 Chr.15: 64381060-64381060 on Build GRCh38
rs892906625 Chr.15: 64381107-64381107 on Build GRCh38
rs746162199 Chr.15: 64380947-64380947 on Build GRCh38
rs1011308546 Chr.15: 64381111-64381111 on Build GRCh38
rs200434561 Chr.15: 64381059-64381059 on Build GRCh38
rs747769298 Chr.15: 64380933-64380933 on Build GRCh38
rs942871324 Chr.15: 64381152-64381152 on Build GRCh38
rs376008563 Chr.15: 64380940-64380940 on Build GRCh38
rs577503846 Chr.15: 64381243-64381243 on Build GRCh38
rs1003390350 Chr.15: 64381249-64381249 on Build GRCh38
rs780593782 Chr.15: 64381340-64381340 on Build GRCh38
rs766165927 Chr.15: 64380917-64380917 on Build GRCh38
rs780306735 Chr.15: 64381289-64381289 on Build GRCh38
rs868538497 Chr.15: 64381021-64381021 on Build GRCh38
rs781442087 Chr.15: 64381080-64381080 on Build GRCh38
rs375863113 Chr.15: 64381256-64381256 on Build GRCh38
rs962769904 Chr.15: 64381231-64381231 on Build GRCh38
rs1056607258 Chr.15: 64381344-64381344 on Build GRCh38
rs202164043 Chr.15: 64381296-64381296 on Build GRCh38
rs372672778 Chr.15: 64380929-64380929 on Build GRCh38
rs369121099 Chr.15: 64381335-64381335 on Build GRCh38
rs543294479 Chr.15: 64381209-64381209 on Build GRCh38
rs960767839 Chr.15: 64381191-64381191 on Build GRCh38
rs182521306 Chr.15: 64381187-64381187 on Build GRCh38
rs773069483 Chr.15: 64381020-64381020 on Build GRCh38
rs372935868 Chr.15: 64381337-64381337 on Build GRCh38
rs559409372 Chr.15: 64381272-64381272 on Build GRCh38
rs564807992 Chr.15: 64381281-64381281 on Build GRCh38
rs889965430 Chr.15: 64381005-64381005 on Build GRCh38
rs1011068030 Chr.15: 64380905-64380905 on Build GRCh38
rs1017090035 Chr.15: 64381105-64381105 on Build GRCh38
rs765324151 Chr.15: 64381013-64381013 on Build GRCh38
rs567110586 Chr.15: 64381006-64381006 on Build GRCh38
rs908606744 Chr.15: 64381041-64381041 on Build GRCh38
rs764067006 Chr.15: 64381059-64381059 on Build GRCh38
rs780875295 Chr.15: 64380930-64380930 on Build GRCh38
rs762655316 Chr.15: 64381027-64381027 on Build GRCh38
rs1012493132 Chr.15: 64381270-64381270 on Build GRCh38
rs201415383 Chr.15: 64381314-64381314 on Build GRCh38
rs770123662 Chr.15: 64381279-64381279 on Build GRCh38
rs758674691 Chr.15: 64381062-64381062 on Build GRCh38
rs759492613 Chr.15: 64381300-64381300 on Build GRCh38
rs141142230 Chr.15: 64381288-64381289 on Build GRCh38
rs201688731 Chr.15: 64380948-64380948 on Build GRCh38
rs774997542 Chr.15: 64381217-64381217 on Build GRCh38
rs760097397 Chr.15: 64381327-64381327 on Build GRCh38
rs866948787 Chr.15: 64381091-64381091 on Build GRCh38
rs768833618 Chr.15: 64381002-64381002 on Build GRCh38

View Full Product Details