Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65454331-65454875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
UniGene
Hs.458609
Gene Chromosome Location
Chr.15: 65442463-65517713 on Build GRCh38
Transcripts
NM_001320875.1 NM_001320876.1 NM_017743.5 NM_130434.4 NM_197960.3 NM_197961.3 NR_135485.1 NR_135486.1 XM_005254505.1 XM_011521730.1 XM_011521731.1 XM_011521732.2 XM_011521733.1 XM_011521734.1 XM_017022373.1 XM_017022374.1 XM_017022375.1 XM_017022376.1 XM_017022377.1 XM_017022378.1 XM_017022379.1 XM_017022380.1 XM_017022381.1 XM_017022382.1 XM_017022383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929565 c.2218C>T p.R740* Chr.15: 65454364-65454364 on Build GRCh38
6649021 c.2185G>A p.D729N Chr.15: 65454397-65454397 on Build GRCh38
6649022 c.2137G>A p.D713N Chr.15: 65454397-65454397 on Build GRCh38
5929566 c.2170C>T p.R724* Chr.15: 65454364-65454364 on Build GRCh38
964062 c.2171G>A p.R724Q Chr.15: 65454363-65454363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919655767 Chr.15: 65454612-65454612 on Build GRCh38
rs560425972 Chr.15: 65454563-65454563 on Build GRCh38
rs1044985769 Chr.15: 65454673-65454673 on Build GRCh38
rs755760929 Chr.15: 65454420-65454421 on Build GRCh38
rs796482240 Chr.15: 65454613-65454613 on Build GRCh38
rs114135120 Chr.15: 65454604-65454604 on Build GRCh38
rs376085142 Chr.15: 65454828-65454828 on Build GRCh38
rs143327271 Chr.15: 65454831-65454832 on Build GRCh38
rs956734848 Chr.15: 65454521-65454521 on Build GRCh38
rs142425317 Chr.15: 65454676-65454676 on Build GRCh38
rs771332122 Chr.15: 65454427-65454427 on Build GRCh38
rs918393177 Chr.15: 65454712-65454712 on Build GRCh38
rs1034297024 Chr.15: 65454768-65454768 on Build GRCh38
rs902679716 Chr.15: 65454840-65454840 on Build GRCh38
rs552453632 Chr.15: 65454647-65454647 on Build GRCh38
rs1055043290 Chr.15: 65454377-65454377 on Build GRCh38
rs959007715 Chr.15: 65454669-65454669 on Build GRCh38
rs189043654 Chr.15: 65454499-65454499 on Build GRCh38
rs910674910 Chr.15: 65454596-65454596 on Build GRCh38
rs1013431539 Chr.15: 65454380-65454380 on Build GRCh38
rs763916455 Chr.15: 65454464-65454464 on Build GRCh38
rs1030715953 Chr.15: 65454735-65454735 on Build GRCh38
rs771842252 Chr.15: 65454445-65454445 on Build GRCh38
rs540718279 Chr.15: 65454549-65454549 on Build GRCh38
rs773708779 Chr.15: 65454359-65454359 on Build GRCh38
rs561242601 Chr.15: 65454490-65454490 on Build GRCh38
rs1030601746 Chr.15: 65454561-65454561 on Build GRCh38
rs1008266676 Chr.15: 65454776-65454776 on Build GRCh38
rs531038777 Chr.15: 65454852-65454852 on Build GRCh38
rs1009537468 Chr.15: 65454734-65454734 on Build GRCh38
rs757169316 Chr.15: 65454694-65454694 on Build GRCh38
rs981847408 Chr.15: 65454648-65454648 on Build GRCh38
rs941263290 Chr.15: 65454692-65454692 on Build GRCh38
rs200291174 Chr.15: 65454465-65454465 on Build GRCh38
rs1032414435 Chr.15: 65454693-65454693 on Build GRCh38
rs748955538 Chr.15: 65454460-65454460 on Build GRCh38
rs890482266 Chr.15: 65454797-65454797 on Build GRCh38
rs763416291 Chr.15: 65454363-65454363 on Build GRCh38
rs1051795954 Chr.15: 65454752-65454752 on Build GRCh38
rs972981310 Chr.15: 65454701-65454702 on Build GRCh38
rs929644843 Chr.15: 65454623-65454623 on Build GRCh38
rs1034720642 Chr.15: 65454655-65454655 on Build GRCh38
rs566909473 Chr.15: 65454821-65454821 on Build GRCh38
rs1000640744 Chr.15: 65454685-65454685 on Build GRCh38
rs764901709 Chr.15: 65454466-65454466 on Build GRCh38
rs951074871 Chr.15: 65454826-65454826 on Build GRCh38
rs986325792 Chr.15: 65454595-65454595 on Build GRCh38
rs929427731 Chr.15: 65454722-65454722 on Build GRCh38
rs548499711 Chr.15: 65454820-65454820 on Build GRCh38
rs182735251 Chr.15: 65454448-65454448 on Build GRCh38
rs770695851 Chr.15: 65454465-65454466 on Build GRCh38
rs1003315892 Chr.15: 65454851-65454851 on Build GRCh38
rs145976889 Chr.15: 65454398-65454398 on Build GRCh38
rs760595723 Chr.15: 65454454-65454454 on Build GRCh38
rs193008753 Chr.15: 65454699-65454699 on Build GRCh38
rs917611762 Chr.15: 65454836-65454836 on Build GRCh38
rs963561118 Chr.15: 65454804-65454804 on Build GRCh38
rs1046733299 Chr.15: 65454641-65454641 on Build GRCh38
rs942211512 Chr.15: 65454603-65454603 on Build GRCh38
rs929354146 Chr.15: 65454492-65454492 on Build GRCh38
rs750149513 Chr.15: 65454811-65454811 on Build GRCh38
rs778351830 Chr.15: 65454391-65454391 on Build GRCh38
rs937942672 Chr.15: 65454356-65454356 on Build GRCh38
rs777462003 Chr.15: 65454456-65454457 on Build GRCh38
rs954989701 Chr.15: 65454772-65454772 on Build GRCh38
rs1023855428 Chr.15: 65454394-65454394 on Build GRCh38
rs1051724953 Chr.15: 65454545-65454545 on Build GRCh38
rs949287474 Chr.15: 65454649-65454649 on Build GRCh38
rs765223347 Chr.15: 65454803-65454803 on Build GRCh38
rs759689505 Chr.15: 65454443-65454443 on Build GRCh38
rs1039008544 Chr.15: 65454810-65454810 on Build GRCh38
rs774528179 Chr.15: 65454434-65454434 on Build GRCh38
rs949073109 Chr.15: 65454845-65454845 on Build GRCh38

View Full Product Details