Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65466843-65467368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
UniGene
Hs.458609
Gene Chromosome Location
Chr.15: 65442463-65517713 on Build GRCh38
Transcripts
NM_001320875.1 NM_001320876.1 NM_017743.5 NM_130434.4 NM_197960.3 NM_197961.3 NR_135485.1 NR_135486.1 XM_005254505.1 XM_011521730.1 XM_011521731.1 XM_011521732.2 XM_011521733.1 XM_011521734.1 XM_017022373.1 XM_017022374.1 XM_017022375.1 XM_017022376.1 XM_017022377.1 XM_017022378.1 XM_017022379.1 XM_017022380.1 XM_017022381.1 XM_017022382.1 XM_017022383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1323886 c.1636G>T p.E546* Chr.15: 65467124-65467124 on Build GRCh38
4652618 c.1634A>G p.D545G Chr.15: 65467174-65467174 on Build GRCh38
2256585 c.1653T>C p.T551T Chr.15: 65467107-65467107 on Build GRCh38
2256583 c.1688A>G p.Q563R Chr.15: 65467072-65467072 on Build GRCh38
3932137 c.1647G>A p.E549E Chr.15: 65467161-65467161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559503285 Chr.15: 65467082-65467082 on Build GRCh38
rs758546266 Chr.15: 65467015-65467015 on Build GRCh38
rs765457590 Chr.15: 65467241-65467241 on Build GRCh38
rs375992766 Chr.15: 65467277-65467277 on Build GRCh38
rs771493218 Chr.15: 65467095-65467095 on Build GRCh38
rs1045323213 Chr.15: 65467336-65467336 on Build GRCh38
rs748946642 Chr.15: 65466869-65466869 on Build GRCh38
rs1045695738 Chr.15: 65467050-65467050 on Build GRCh38
rs763114062 Chr.15: 65467234-65467236 on Build GRCh38
rs745687089 Chr.15: 65467030-65467030 on Build GRCh38
rs1030038496 Chr.15: 65466876-65466876 on Build GRCh38
rs547235511 Chr.15: 65467292-65467292 on Build GRCh38
rs370549371 Chr.15: 65467103-65467103 on Build GRCh38
rs762667349 Chr.15: 65467326-65467328 on Build GRCh38
rs542879302 Chr.15: 65466977-65466977 on Build GRCh38
rs761976530 Chr.15: 65467228-65467228 on Build GRCh38
rs774661670 Chr.15: 65467099-65467099 on Build GRCh38
rs915895498 Chr.15: 65467299-65467299 on Build GRCh38
rs765268257 Chr.15: 65467125-65467125 on Build GRCh38
rs758476300 Chr.15: 65467254-65467254 on Build GRCh38
rs928687868 Chr.15: 65466964-65466964 on Build GRCh38
rs147909400 Chr.15: 65467219-65467219 on Build GRCh38
rs1003290617 Chr.15: 65467271-65467271 on Build GRCh38
rs188375473 Chr.15: 65466883-65466883 on Build GRCh38
rs758254879 Chr.15: 65467132-65467132 on Build GRCh38
rs375352055 Chr.15: 65467154-65467154 on Build GRCh38
rs760756541 Chr.15: 65467217-65467217 on Build GRCh38
rs772390890 Chr.15: 65467201-65467201 on Build GRCh38
rs764054639 Chr.15: 65467113-65467113 on Build GRCh38
rs796920831 Chr.15: 65466985-65466985 on Build GRCh38
rs771781997 Chr.15: 65467035-65467035 on Build GRCh38
rs780061945 Chr.15: 65467136-65467136 on Build GRCh38
rs774856069 Chr.15: 65467182-65467182 on Build GRCh38
rs750354590 Chr.15: 65467253-65467253 on Build GRCh38
rs1032771854 Chr.15: 65467064-65467064 on Build GRCh38
rs74561645 Chr.15: 65467204-65467204 on Build GRCh38
rs573882501 Chr.15: 65466906-65466906 on Build GRCh38
rs1035750588 Chr.15: 65467281-65467281 on Build GRCh38
rs746436941 Chr.15: 65467194-65467194 on Build GRCh38
rs34833283 Chr.15: 65467284-65467285 on Build GRCh38
rs958163514 Chr.15: 65466926-65466926 on Build GRCh38
rs531015904 Chr.15: 65467137-65467137 on Build GRCh38
rs766716862 Chr.15: 65467247-65467248 on Build GRCh38
rs974073991 Chr.15: 65467336-65467337 on Build GRCh38
rs759901599 Chr.15: 65467102-65467102 on Build GRCh38
rs755132636 Chr.15: 65467265-65467265 on Build GRCh38
rs776246737 Chr.15: 65467090-65467090 on Build GRCh38
rs373576240 Chr.15: 65467056-65467056 on Build GRCh38
rs1047084822 Chr.15: 65466929-65466929 on Build GRCh38
rs751632817 Chr.15: 65467148-65467148 on Build GRCh38
rs764405977 Chr.15: 65467218-65467218 on Build GRCh38
rs989355286 Chr.15: 65466927-65466927 on Build GRCh38
rs1038433139 Chr.15: 65466870-65466870 on Build GRCh38
rs967528239 Chr.15: 65466943-65466943 on Build GRCh38
rs144699847 Chr.15: 65467155-65467155 on Build GRCh38
rs754709462 Chr.15: 65467146-65467146 on Build GRCh38
rs991422065 Chr.15: 65467012-65467012 on Build GRCh38
rs565424889 Chr.15: 65467117-65467117 on Build GRCh38
rs71396463 Chr.15: 65467238-65467238 on Build GRCh38
rs73480741 Chr.15: 65466893-65466893 on Build GRCh38
rs377019921 Chr.15: 65467060-65467060 on Build GRCh38
rs374355237 Chr.15: 65466957-65466957 on Build GRCh38
rs916194274 Chr.15: 65467036-65467036 on Build GRCh38
rs1011237672 Chr.15: 65467161-65467161 on Build GRCh38
rs780520983 Chr.15: 65467149-65467149 on Build GRCh38
rs545521561 Chr.15: 65467110-65467110 on Build GRCh38
rs751381994 Chr.15: 65467269-65467269 on Build GRCh38
rs1019970313 Chr.15: 65467107-65467107 on Build GRCh38
rs77365136 Chr.15: 65467282-65467282 on Build GRCh38
rs763349508 Chr.15: 65467091-65467091 on Build GRCh38
rs949391521 Chr.15: 65467323-65467323 on Build GRCh38
rs757101017 Chr.15: 65467122-65467122 on Build GRCh38
rs750184682 Chr.15: 65467130-65467130 on Build GRCh38
rs557104790 Chr.15: 65466901-65466901 on Build GRCh38

View Full Product Details