Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86154340-86154839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848816 c.138A>G p.G46G Chr.15: 86154443-86154443 on Build GRCh38
6298178 c.224G>T p.C75F Chr.15: 86154529-86154529 on Build GRCh38
3504915 c.251C>T p.S84F Chr.15: 86154556-86154556 on Build GRCh38
4974049 c.239G>A p.R80H Chr.15: 86154544-86154544 on Build GRCh38
4957672 c.206G>A p.G69D Chr.15: 86154511-86154511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776060266 Chr.15: 86154527-86154527 on Build GRCh38
rs772906867 Chr.15: 86154439-86154439 on Build GRCh38
rs776473286 Chr.15: 86154431-86154432 on Build GRCh38
rs754991663 Chr.15: 86154420-86154420 on Build GRCh38
rs150261781 Chr.15: 86154444-86154444 on Build GRCh38
rs762071832 Chr.15: 86154546-86154546 on Build GRCh38
rs1002379683 Chr.15: 86154483-86154483 on Build GRCh38
rs752649388 Chr.15: 86154557-86154557 on Build GRCh38
rs138966010 Chr.15: 86154542-86154542 on Build GRCh38
rs1051057616 Chr.15: 86154697-86154697 on Build GRCh38
rs1014163656 Chr.15: 86154813-86154813 on Build GRCh38
rs560232482 Chr.15: 86154434-86154434 on Build GRCh38
rs759985409 Chr.15: 86154448-86154448 on Build GRCh38
rs746632721 Chr.15: 86154517-86154517 on Build GRCh38
rs548207356 Chr.15: 86154433-86154433 on Build GRCh38
rs749202050 Chr.15: 86154416-86154416 on Build GRCh38
rs199905407 Chr.15: 86154575-86154575 on Build GRCh38
rs778991414 Chr.15: 86154501-86154501 on Build GRCh38
rs971411510 Chr.15: 86154493-86154493 on Build GRCh38
rs762265930 Chr.15: 86154596-86154596 on Build GRCh38
rs544335237 Chr.15: 86154504-86154504 on Build GRCh38
rs773547112 Chr.15: 86154610-86154610 on Build GRCh38
rs751323432 Chr.15: 86154563-86154563 on Build GRCh38
rs572299339 Chr.15: 86154675-86154675 on Build GRCh38
rs765745164 Chr.15: 86154450-86154450 on Build GRCh38
rs372376267 Chr.15: 86154558-86154558 on Build GRCh38
rs1046838581 Chr.15: 86154377-86154377 on Build GRCh38
rs772624377 Chr.15: 86154604-86154604 on Build GRCh38
rs776148401 Chr.15: 86154445-86154445 on Build GRCh38
rs563023472 Chr.15: 86154765-86154765 on Build GRCh38
rs765774131 Chr.15: 86154399-86154399 on Build GRCh38
rs757342123 Chr.15: 86154413-86154413 on Build GRCh38
rs777002622 Chr.15: 86154393-86154393 on Build GRCh38
rs1033745429 Chr.15: 86154785-86154785 on Build GRCh38
rs1039531359 Chr.15: 86154654-86154654 on Build GRCh38
rs960893552 Chr.15: 86154801-86154801 on Build GRCh38
rs141400352 Chr.15: 86154717-86154718 on Build GRCh38
rs992445657 Chr.15: 86154569-86154569 on Build GRCh38
rs892499228 Chr.15: 86154458-86154458 on Build GRCh38
rs7161846 Chr.15: 86154365-86154365 on Build GRCh38
rs754898743 Chr.15: 86154497-86154497 on Build GRCh38
rs907286193 Chr.15: 86154799-86154799 on Build GRCh38
rs746689219 Chr.15: 86154442-86154442 on Build GRCh38
rs759854587 Chr.15: 86154397-86154397 on Build GRCh38
rs374388706 Chr.15: 86154524-86154524 on Build GRCh38
rs368702760 Chr.15: 86154579-86154579 on Build GRCh38
rs549745873 Chr.15: 86154554-86154554 on Build GRCh38
rs985917131 Chr.15: 86154617-86154617 on Build GRCh38
rs750546749 Chr.15: 86154489-86154489 on Build GRCh38
rs766419259 Chr.15: 86154556-86154556 on Build GRCh38
rs767658695 Chr.15: 86154471-86154471 on Build GRCh38
rs968083705 Chr.15: 86154476-86154476 on Build GRCh38
rs201845968 Chr.15: 86154737-86154739 on Build GRCh38
rs535428630 Chr.15: 86154657-86154657 on Build GRCh38
rs758468484 Chr.15: 86154408-86154408 on Build GRCh38
rs764420756 Chr.15: 86154466-86154466 on Build GRCh38
rs760826210 Chr.15: 86154555-86154555 on Build GRCh38
rs995102529 Chr.15: 86154378-86154378 on Build GRCh38
rs930549039 Chr.15: 86154679-86154679 on Build GRCh38
rs371439668 Chr.15: 86154670-86154670 on Build GRCh38
rs909641157 Chr.15: 86154612-86154612 on Build GRCh38
rs777468842 Chr.15: 86154513-86154513 on Build GRCh38
rs960965539 Chr.15: 86154566-86154566 on Build GRCh38
rs142973417 Chr.15: 86154749-86154749 on Build GRCh38
rs572068096 Chr.15: 86154730-86154730 on Build GRCh38
rs747649160 Chr.15: 86154386-86154386 on Build GRCh38
rs763415157 Chr.15: 86154543-86154543 on Build GRCh38
rs182802715 Chr.15: 86154544-86154544 on Build GRCh38
rs765889786 Chr.15: 86154384-86154384 on Build GRCh38
rs1051437081 Chr.15: 86154449-86154449 on Build GRCh38
rs780816050 Chr.15: 86154577-86154577 on Build GRCh38
rs553739134 Chr.15: 86154688-86154688 on Build GRCh38
rs191931920 Chr.15: 86154467-86154467 on Build GRCh38
rs1009571128 Chr.15: 86154736-86154736 on Build GRCh38
rs868466402 Chr.15: 86154437-86154437 on Build GRCh38
rs753183150 Chr.15: 86154403-86154403 on Build GRCh38
rs992506660 Chr.15: 86154363-86154363 on Build GRCh38
rs7169490 Chr.15: 86154769-86154769 on Build GRCh38
rs919490336 Chr.15: 86154576-86154576 on Build GRCh38
rs758190679 Chr.15: 86154508-86154508 on Build GRCh38
rs529592156 Chr.15: 86154410-86154410 on Build GRCh38
rs892372544 Chr.15: 86154705-86154705 on Build GRCh38
rs775648368 Chr.15: 86154461-86154461 on Build GRCh38
rs763367858 Chr.15: 86154465-86154465 on Build GRCh38
rs1049376602 Chr.15: 86154703-86154703 on Build GRCh38
rs376033481 Chr.15: 86154381-86154381 on Build GRCh38
rs777575244 Chr.15: 86154561-86154561 on Build GRCh38
rs781466603 Chr.15: 86154415-86154415 on Build GRCh38
rs371403599 Chr.15: 86154586-86154586 on Build GRCh38
rs769060136 Chr.15: 86154590-86154590 on Build GRCh38
rs771829983 Chr.15: 86154438-86154438 on Build GRCh38
rs755943063 Chr.15: 86154676-86154676 on Build GRCh38
rs375958164 Chr.15: 86154725-86154725 on Build GRCh38
rs929613133 Chr.15: 86154372-86154372 on Build GRCh38
rs770592400 Chr.15: 86154525-86154525 on Build GRCh38
rs372684327 Chr.15: 86154719-86154719 on Build GRCh38
rs959056012 Chr.15: 86154382-86154382 on Build GRCh38
rs769267810 Chr.15: 86154585-86154585 on Build GRCh38
rs781210490 Chr.15: 86154635-86154635 on Build GRCh38
rs376493772 Chr.15: 86154665-86154665 on Build GRCh38
rs769357624 Chr.15: 86154540-86154540 on Build GRCh38
rs1022416065 Chr.15: 86154474-86154474 on Build GRCh38
rs368287289 Chr.15: 86154531-86154531 on Build GRCh38
rs1012971538 Chr.15: 86154387-86154387 on Build GRCh38
rs757455858 Chr.15: 86154468-86154468 on Build GRCh38
rs767859259 Chr.15: 86154553-86154553 on Build GRCh38
rs557800233 Chr.15: 86154764-86154764 on Build GRCh38
rs751081785 Chr.15: 86154418-86154418 on Build GRCh38

View Full Product Details