Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86257861-86258391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363054 c.784G>A p.D262N Chr.15: 86257984-86257984 on Build GRCh38
1375166 c.800A>G p.K267R Chr.15: 86258000-86258000 on Build GRCh38
4558390 c.766G>A p.D256N Chr.15: 86257966-86257966 on Build GRCh38
4567684 c.821_822GG>AA p.G274E Chr.15: 86258021-86258022 on Build GRCh38
5176317 c.800A>G p.K267R Chr.15: 86258000-86258000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775153616 Chr.15: 86258056-86258056 on Build GRCh38
rs772055068 Chr.15: 86258036-86258036 on Build GRCh38
rs748746107 Chr.15: 86258163-86258163 on Build GRCh38
rs764653479 Chr.15: 86258066-86258066 on Build GRCh38
rs951828151 Chr.15: 86258286-86258286 on Build GRCh38
rs372187949 Chr.15: 86258073-86258073 on Build GRCh38
rs910234895 Chr.15: 86258144-86258144 on Build GRCh38
rs376926549 Chr.15: 86257921-86257921 on Build GRCh38
rs552802247 Chr.15: 86258026-86258026 on Build GRCh38
rs915814219 Chr.15: 86258170-86258170 on Build GRCh38
rs573037511 Chr.15: 86258185-86258185 on Build GRCh38
rs536331587 Chr.15: 86258158-86258158 on Build GRCh38
rs371923647 Chr.15: 86257984-86257984 on Build GRCh38
rs549906044 Chr.15: 86258200-86258200 on Build GRCh38
rs113770077 Chr.15: 86258169-86258169 on Build GRCh38
rs760505180 Chr.15: 86258044-86258044 on Build GRCh38
rs776392677 Chr.15: 86258049-86258049 on Build GRCh38
rs763302503 Chr.15: 86258062-86258062 on Build GRCh38
rs775629809 Chr.15: 86257970-86257971 on Build GRCh38
rs57877588 Chr.15: 86258283-86258283 on Build GRCh38
rs566704959 Chr.15: 86257995-86257995 on Build GRCh38
rs374213102 Chr.15: 86258007-86258007 on Build GRCh38
rs771198475 Chr.15: 86257931-86257931 on Build GRCh38
rs560330156 Chr.15: 86258291-86258291 on Build GRCh38
rs767325824 Chr.15: 86257983-86257983 on Build GRCh38
rs565138089 Chr.15: 86258215-86258215 on Build GRCh38
rs965920573 Chr.15: 86258146-86258146 on Build GRCh38
rs192090714 Chr.15: 86258030-86258030 on Build GRCh38
rs769832023 Chr.15: 86258022-86258022 on Build GRCh38
rs527846309 Chr.15: 86258304-86258304 on Build GRCh38
rs928471339 Chr.15: 86258333-86258333 on Build GRCh38
rs757625817 Chr.15: 86258012-86258012 on Build GRCh38
rs534244630 Chr.15: 86258002-86258002 on Build GRCh38
rs934745048 Chr.15: 86258191-86258191 on Build GRCh38
rs555747901 Chr.15: 86257895-86257895 on Build GRCh38
rs553495238 Chr.15: 86258347-86258347 on Build GRCh38
rs374703938 Chr.15: 86257948-86257948 on Build GRCh38
rs764881820 Chr.15: 86257992-86257992 on Build GRCh38
rs771254013 Chr.15: 86258039-86258039 on Build GRCh38
rs956007657 Chr.15: 86258307-86258307 on Build GRCh38
rs13380370 Chr.15: 86257907-86257907 on Build GRCh38
rs914570417 Chr.15: 86258201-86258201 on Build GRCh38
rs915763394 Chr.15: 86257947-86257947 on Build GRCh38
rs976000605 Chr.15: 86258161-86258161 on Build GRCh38
rs201119906 Chr.15: 86257924-86257924 on Build GRCh38
rs142535293 Chr.15: 86258162-86258162 on Build GRCh38
rs971804497 Chr.15: 86258159-86258159 on Build GRCh38
rs969919864 Chr.15: 86258178-86258178 on Build GRCh38
rs374974159 Chr.15: 86257916-86257916 on Build GRCh38
rs185403924 Chr.15: 86258116-86258116 on Build GRCh38
rs747823849 Chr.15: 86258240-86258240 on Build GRCh38
rs948524576 Chr.15: 86257998-86257998 on Build GRCh38
rs55987855 Chr.15: 86258366-86258366 on Build GRCh38
rs562690374 Chr.15: 86257936-86257936 on Build GRCh38
rs769916270 Chr.15: 86257944-86257944 on Build GRCh38
rs370635198 Chr.15: 86257939-86257939 on Build GRCh38
rs746134714 Chr.15: 86258021-86258021 on Build GRCh38
rs965706259 Chr.15: 86257890-86257890 on Build GRCh38
rs542241355 Chr.15: 86258197-86258197 on Build GRCh38
rs1005884616 Chr.15: 86258287-86258287 on Build GRCh38
rs756071634 Chr.15: 86258099-86258099 on Build GRCh38
rs955871029 Chr.15: 86258360-86258360 on Build GRCh38
rs140755143 Chr.15: 86258230-86258230 on Build GRCh38
rs548411308 Chr.15: 86257994-86257994 on Build GRCh38
rs759295070 Chr.15: 86258050-86258050 on Build GRCh38
rs368695778 Chr.15: 86258048-86258048 on Build GRCh38
rs571162909 Chr.15: 86258332-86258332 on Build GRCh38
rs544636265 Chr.15: 86258243-86258243 on Build GRCh38
rs757480562 Chr.15: 86257976-86257976 on Build GRCh38
rs747150818 Chr.15: 86257927-86257927 on Build GRCh38
rs60467151 Chr.15: 86258275-86258275 on Build GRCh38
rs773093713 Chr.15: 86258041-86258041 on Build GRCh38
rs530127001 Chr.15: 86257990-86257990 on Build GRCh38
rs1044340660 Chr.15: 86258199-86258199 on Build GRCh38
rs773077463 Chr.15: 86257970-86257970 on Build GRCh38
rs764789862 Chr.15: 86258054-86258054 on Build GRCh38
rs928419479 Chr.15: 86258189-86258189 on Build GRCh38
rs774247398 Chr.15: 86257946-86257946 on Build GRCh38
rs149696422 Chr.15: 86258323-86258323 on Build GRCh38
rs975808547 Chr.15: 86258298-86258298 on Build GRCh38
rs777941296 Chr.15: 86258003-86258003 on Build GRCh38
rs965701606 Chr.15: 86258297-86258297 on Build GRCh38
rs562529572 Chr.15: 86258244-86258244 on Build GRCh38
rs1037681265 Chr.15: 86258237-86258237 on Build GRCh38
rs990366660 Chr.15: 86258337-86258337 on Build GRCh38
rs1040205547 Chr.15: 86257891-86257891 on Build GRCh38
rs1027170484 Chr.15: 86258264-86258264 on Build GRCh38
rs781375714 Chr.15: 86258017-86258017 on Build GRCh38
rs965758499 Chr.15: 86258130-86258130 on Build GRCh38
rs907285815 Chr.15: 86258023-86258023 on Build GRCh38
rs753308441 Chr.15: 86257917-86257917 on Build GRCh38
rs1008331084 Chr.15: 86258113-86258113 on Build GRCh38
rs1012857317 Chr.15: 86257922-86257922 on Build GRCh38

View Full Product Details