Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89175728-89176226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD2
Entrez Gene ID
11057
Gene Name
abhydrolase domain containing 2
Gene Aliases
HS1-2, LABH2, PHPS1-2
UniGene
Hs.122337
Gene Chromosome Location
Chr.15: 89088150-89202360 on Build GRCh38
Transcripts
NM_007011.7 NM_152924.4 XM_005254832.1 XM_005254833.1 XM_011521165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
966278 c.705G>T p.Q235H Chr.15: 89175978-89175978 on Build GRCh38
6590894 c.607G>A p.V203I Chr.15: 89175880-89175880 on Build GRCh38
1375310 c.551A>G p.E184G Chr.15: 89175824-89175824 on Build GRCh38
6428805 c.610G>A p.V204M Chr.15: 89175883-89175883 on Build GRCh38
25208 c.720G>A p.L240L Chr.15: 89175993-89175993 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773174046 Chr.15: 89175799-89175799 on Build GRCh38
rs150079355 Chr.15: 89175970-89175970 on Build GRCh38
rs748236226 Chr.15: 89176008-89176008 on Build GRCh38
rs777461364 Chr.15: 89175852-89175852 on Build GRCh38
rs759224037 Chr.15: 89175835-89175835 on Build GRCh38
rs971857080 Chr.15: 89176152-89176152 on Build GRCh38
rs780427734 Chr.15: 89175991-89175991 on Build GRCh38
rs199669891 Chr.15: 89175939-89175939 on Build GRCh38
rs539541463 Chr.15: 89176104-89176104 on Build GRCh38
rs547064918 Chr.15: 89175870-89175870 on Build GRCh38
rs757775669 Chr.15: 89175969-89175969 on Build GRCh38
rs781435580 Chr.15: 89175774-89175774 on Build GRCh38
rs375201895 Chr.15: 89175955-89175955 on Build GRCh38
rs774435586 Chr.15: 89175908-89175908 on Build GRCh38
rs763642629 Chr.15: 89175963-89175963 on Build GRCh38
rs1005398 Chr.15: 89176144-89176144 on Build GRCh38
rs752233450 Chr.15: 89175843-89175843 on Build GRCh38
rs762721320 Chr.15: 89175961-89175961 on Build GRCh38
rs776473741 Chr.15: 89176021-89176021 on Build GRCh38
rs201540119 Chr.15: 89175972-89175972 on Build GRCh38
rs1032881804 Chr.15: 89176088-89176088 on Build GRCh38
rs766147852 Chr.15: 89175809-89175809 on Build GRCh38
rs1050183204 Chr.15: 89176043-89176043 on Build GRCh38
rs887149056 Chr.15: 89175876-89175876 on Build GRCh38
rs751039237 Chr.15: 89175964-89175964 on Build GRCh38
rs200218946 Chr.15: 89175880-89175880 on Build GRCh38
rs916136851 Chr.15: 89176101-89176101 on Build GRCh38
rs763075619 Chr.15: 89175795-89175795 on Build GRCh38
rs1024359479 Chr.15: 89176150-89176150 on Build GRCh38
rs760560974 Chr.15: 89175806-89175806 on Build GRCh38
rs544589818 Chr.15: 89176100-89176100 on Build GRCh38
rs768682861 Chr.15: 89175885-89175885 on Build GRCh38
rs765041299 Chr.15: 89175839-89175839 on Build GRCh38
rs781555313 Chr.15: 89175857-89175857 on Build GRCh38
rs746904927 Chr.15: 89175935-89175935 on Build GRCh38
rs941634316 Chr.15: 89175998-89175998 on Build GRCh38
rs771290659 Chr.15: 89175767-89175767 on Build GRCh38
rs146397660 Chr.15: 89176009-89176009 on Build GRCh38
rs748569686 Chr.15: 89176158-89176158 on Build GRCh38
rs139776940 Chr.15: 89176056-89176056 on Build GRCh38
rs186981362 Chr.15: 89176086-89176086 on Build GRCh38
rs759300103 Chr.15: 89175950-89175950 on Build GRCh38
rs755205213 Chr.15: 89176004-89176004 on Build GRCh38
rs745850083 Chr.15: 89176024-89176024 on Build GRCh38
rs1036267202 Chr.15: 89176074-89176075 on Build GRCh38
rs756472065 Chr.15: 89175981-89175981 on Build GRCh38
rs932800707 Chr.15: 89176034-89176034 on Build GRCh38
rs752067835 Chr.15: 89175853-89175853 on Build GRCh38
rs746282410 Chr.15: 89175858-89175858 on Build GRCh38
rs1008582270 Chr.15: 89176055-89176055 on Build GRCh38
rs775540935 Chr.15: 89175793-89175793 on Build GRCh38
rs761283479 Chr.15: 89176092-89176092 on Build GRCh38
rs533800783 Chr.15: 89175849-89175849 on Build GRCh38
rs369234475 Chr.15: 89175763-89175763 on Build GRCh38
rs372442772 Chr.15: 89175792-89175792 on Build GRCh38
rs142298746 Chr.15: 89175882-89175882 on Build GRCh38
rs991566252 Chr.15: 89175927-89175927 on Build GRCh38
rs749462886 Chr.15: 89176002-89176002 on Build GRCh38
rs977653197 Chr.15: 89176059-89176059 on Build GRCh38
rs940095954 Chr.15: 89176165-89176165 on Build GRCh38
rs984491786 Chr.15: 89176120-89176120 on Build GRCh38
rs982872732 Chr.15: 89176159-89176159 on Build GRCh38
rs776170737 Chr.15: 89176058-89176058 on Build GRCh38
rs370247405 Chr.15: 89175797-89175797 on Build GRCh38
rs890162854 Chr.15: 89176044-89176044 on Build GRCh38
rs958528757 Chr.15: 89175922-89175922 on Build GRCh38
rs145289422 Chr.15: 89175819-89175819 on Build GRCh38
rs746047601 Chr.15: 89175784-89175784 on Build GRCh38
rs201301214 Chr.15: 89175945-89175945 on Build GRCh38
rs1036221840 Chr.15: 89176186-89176186 on Build GRCh38
rs778474255 Chr.15: 89175761-89175761 on Build GRCh38
rs147649579 Chr.15: 89175856-89175856 on Build GRCh38
rs958540084 Chr.15: 89176110-89176110 on Build GRCh38

View Full Product Details