Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89261741-89262237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
UniGene
Hs.513126
Gene Chromosome Location
Chr.15: 89243954-89317132 on Build GRCh38
Transcripts
NM_001113378.1 NM_018193.2 XM_011521756.2 XM_011521757.2 XM_011521764.2 XM_011521767.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6143170 c.458G>T p.G153V Chr.15: 89261833-89261833 on Build GRCh38
6660484 c.450A>G p.V150V Chr.15: 89261825-89261825 on Build GRCh38
3505236 c.449T>A p.V150E Chr.15: 89261824-89261824 on Build GRCh38
4683367 c.489C>A p.T163T Chr.15: 89261864-89261864 on Build GRCh38
4683366 c.489C>A p.T163T Chr.15: 89261864-89261864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985771344 Chr.15: 89261974-89261976 on Build GRCh38
rs998217496 Chr.15: 89262206-89262206 on Build GRCh38
rs879843471 Chr.15: 89262077-89262077 on Build GRCh38
rs371317348 Chr.15: 89261799-89261799 on Build GRCh38
rs28667701 Chr.15: 89262081-89262081 on Build GRCh38
rs762486364 Chr.15: 89261803-89261803 on Build GRCh38
rs758418421 Chr.15: 89261861-89261861 on Build GRCh38
rs186597857 Chr.15: 89261809-89261809 on Build GRCh38
rs903474883 Chr.15: 89262050-89262050 on Build GRCh38
rs965806124 Chr.15: 89262200-89262200 on Build GRCh38
rs910223342 Chr.15: 89261976-89261976 on Build GRCh38
rs750988926 Chr.15: 89261772-89261772 on Build GRCh38
rs761332032 Chr.15: 89261836-89261836 on Build GRCh38
rs397769335 Chr.15: 89261960-89261961 on Build GRCh38
rs574154974 Chr.15: 89261863-89261863 on Build GRCh38
rs747133340 Chr.15: 89261990-89261990 on Build GRCh38
rs745789593 Chr.15: 89261872-89261872 on Build GRCh38
rs184863079 Chr.15: 89262026-89262026 on Build GRCh38
rs377255054 Chr.15: 89261815-89261815 on Build GRCh38
rs879763864 Chr.15: 89262000-89262000 on Build GRCh38
rs756487442 Chr.15: 89261774-89261774 on Build GRCh38
rs1044339072 Chr.15: 89262111-89262111 on Build GRCh38
rs537136845 Chr.15: 89261830-89261830 on Build GRCh38
rs3075797 Chr.15: 89261959-89261960 on Build GRCh38
rs375678342 Chr.15: 89262163-89262163 on Build GRCh38
rs767388018 Chr.15: 89261849-89261855 on Build GRCh38
rs984731224 Chr.15: 89261867-89261867 on Build GRCh38
rs765463274 Chr.15: 89261850-89261850 on Build GRCh38
rs947592713 Chr.15: 89261982-89261984 on Build GRCh38
rs915398761 Chr.15: 89262032-89262032 on Build GRCh38
rs776675574 Chr.15: 89262083-89262083 on Build GRCh38
rs199841511 Chr.15: 89261926-89261926 on Build GRCh38
rs888277169 Chr.15: 89262180-89262180 on Build GRCh38
rs771706168 Chr.15: 89261903-89261903 on Build GRCh38
rs201645787 Chr.15: 89261789-89261789 on Build GRCh38
rs563270737 Chr.15: 89261956-89261956 on Build GRCh38
rs1044149263 Chr.15: 89262062-89262062 on Build GRCh38
rs761356536 Chr.15: 89261812-89261812 on Build GRCh38
rs112387610 Chr.15: 89261842-89261842 on Build GRCh38
rs769468958 Chr.15: 89261794-89261794 on Build GRCh38
rs749071182 Chr.15: 89261889-89261889 on Build GRCh38
rs530664279 Chr.15: 89261958-89261958 on Build GRCh38
rs775410661 Chr.15: 89261802-89261802 on Build GRCh38
rs778841077 Chr.15: 89261784-89261784 on Build GRCh38
rs761845328 Chr.15: 89261925-89261926 on Build GRCh38
rs369943394 Chr.15: 89262066-89262066 on Build GRCh38
rs768357176 Chr.15: 89261896-89261896 on Build GRCh38
rs761065381 Chr.15: 89261924-89261924 on Build GRCh38
rs374069063 Chr.15: 89261781-89261781 on Build GRCh38
rs79622975 Chr.15: 89262099-89262099 on Build GRCh38
rs947682211 Chr.15: 89261773-89261773 on Build GRCh38
rs989622279 Chr.15: 89262017-89262017 on Build GRCh38
rs768395269 Chr.15: 89261804-89261804 on Build GRCh38
rs200869762 Chr.15: 89261888-89261888 on Build GRCh38
rs768803964 Chr.15: 89262038-89262038 on Build GRCh38
rs192676269 Chr.15: 89261923-89261923 on Build GRCh38
rs747805195 Chr.15: 89261902-89261902 on Build GRCh38
rs192949500 Chr.15: 89262159-89262159 on Build GRCh38
rs3217022 Chr.15: 89261958-89261959 on Build GRCh38
rs754278545 Chr.15: 89261841-89261841 on Build GRCh38
rs748270738 Chr.15: 89261786-89261786 on Build GRCh38
rs766640605 Chr.15: 89261838-89261838 on Build GRCh38
rs768096502 Chr.15: 89261767-89261767 on Build GRCh38
rs1018366495 Chr.15: 89262199-89262199 on Build GRCh38
rs962298483 Chr.15: 89262192-89262192 on Build GRCh38
rs558914752 Chr.15: 89262116-89262116 on Build GRCh38
rs888443839 Chr.15: 89262153-89262153 on Build GRCh38
rs753552933 Chr.15: 89261776-89261776 on Build GRCh38
rs1043307346 Chr.15: 89262011-89262014 on Build GRCh38
rs924221164 Chr.15: 89261937-89261937 on Build GRCh38
rs545051901 Chr.15: 89261954-89261954 on Build GRCh38
rs776717926 Chr.15: 89261924-89261924 on Build GRCh38
rs879314195 Chr.15: 89262078-89262078 on Build GRCh38
rs934829312 Chr.15: 89262053-89262053 on Build GRCh38
rs187618539 Chr.15: 89262122-89262122 on Build GRCh38
rs777039762 Chr.15: 89261928-89261928 on Build GRCh38
rs766384910 Chr.15: 89261934-89261934 on Build GRCh38
rs1007417102 Chr.15: 89262183-89262183 on Build GRCh38
rs1057261148 Chr.15: 89262072-89262072 on Build GRCh38
rs745808001 Chr.15: 89261793-89261793 on Build GRCh38
rs774088324 Chr.15: 89261899-89261899 on Build GRCh38
rs570731414 Chr.15: 89262172-89262172 on Build GRCh38
rs752942226 Chr.15: 89261858-89261858 on Build GRCh38

View Full Product Details