Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90083873-90084369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494433 c.1304C>T p.T435M Chr.15: 90084321-90084321 on Build GRCh38
269346 c.1322T>C p.I441T Chr.15: 90084303-90084303 on Build GRCh38
4691742 c.1324A>G p.K442E Chr.15: 90084301-90084301 on Build GRCh38
5784561 c.1325A>G p.K442R Chr.15: 90084300-90084300 on Build GRCh38
6500191 c.1314C>T p.L438L Chr.15: 90084311-90084311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990164993 Chr.15: 90084220-90084220 on Build GRCh38
rs372655581 Chr.15: 90084236-90084236 on Build GRCh38
rs773670536 Chr.15: 90084228-90084228 on Build GRCh38
rs954587473 Chr.15: 90084256-90084256 on Build GRCh38
rs764864079 Chr.15: 90084320-90084320 on Build GRCh38
rs920191176 Chr.15: 90084347-90084347 on Build GRCh38
rs528326677 Chr.15: 90084044-90084044 on Build GRCh38
rs765526162 Chr.15: 90084215-90084215 on Build GRCh38
rs942753730 Chr.15: 90084052-90084053 on Build GRCh38
rs995162119 Chr.15: 90084095-90084095 on Build GRCh38
rs764173046 Chr.15: 90084274-90084274 on Build GRCh38
rs535303053 Chr.15: 90084243-90084243 on Build GRCh38
rs150574164 Chr.15: 90084323-90084323 on Build GRCh38
rs571080145 Chr.15: 90084193-90084193 on Build GRCh38
rs999442699 Chr.15: 90084200-90084200 on Build GRCh38
rs767032049 Chr.15: 90084265-90084265 on Build GRCh38
rs935360627 Chr.15: 90084153-90084153 on Build GRCh38
rs1017825906 Chr.15: 90084289-90084289 on Build GRCh38
rs769187854 Chr.15: 90083989-90083989 on Build GRCh38
rs993157282 Chr.15: 90084237-90084237 on Build GRCh38
rs778604514 Chr.15: 90084239-90084239 on Build GRCh38
rs148745668 Chr.15: 90084280-90084280 on Build GRCh38
rs368733189 Chr.15: 90084335-90084335 on Build GRCh38
rs760563629 Chr.15: 90084282-90084282 on Build GRCh38
rs757889221 Chr.15: 90084346-90084346 on Build GRCh38
rs750311980 Chr.15: 90084221-90084221 on Build GRCh38
rs1049440726 Chr.15: 90084120-90084120 on Build GRCh38
rs771456529 Chr.15: 90084257-90084257 on Build GRCh38
rs200094442 Chr.15: 90084262-90084262 on Build GRCh38
rs763078146 Chr.15: 90084217-90084217 on Build GRCh38
rs754752564 Chr.15: 90084313-90084313 on Build GRCh38
rs201015211 Chr.15: 90084266-90084266 on Build GRCh38
rs534544502 Chr.15: 90083936-90083936 on Build GRCh38
rs767424579 Chr.15: 90084219-90084219 on Build GRCh38
rs561237245 Chr.15: 90083980-90083980 on Build GRCh38
rs954534670 Chr.15: 90083956-90083956 on Build GRCh38
rs777116882 Chr.15: 90084010-90084010 on Build GRCh38
rs985889811 Chr.15: 90083958-90083958 on Build GRCh38
rs535233898 Chr.15: 90083921-90083922 on Build GRCh38
rs578203629 Chr.15: 90084311-90084311 on Build GRCh38
rs747501657 Chr.15: 90084254-90084254 on Build GRCh38
rs910386641 Chr.15: 90084014-90084014 on Build GRCh38
rs191547948 Chr.15: 90084341-90084341 on Build GRCh38
rs988101235 Chr.15: 90084339-90084339 on Build GRCh38
rs118053940 Chr.15: 90084321-90084321 on Build GRCh38
rs949588153 Chr.15: 90084344-90084344 on Build GRCh38
rs780288726 Chr.15: 90084151-90084151 on Build GRCh38
rs376215927 Chr.15: 90084244-90084244 on Build GRCh38
rs1802920 Chr.15: 90084109-90084109 on Build GRCh38
rs577520416 Chr.15: 90084203-90084203 on Build GRCh38
rs553627406 Chr.15: 90084271-90084271 on Build GRCh38
rs114227266 Chr.15: 90084122-90084122 on Build GRCh38
rs555429283 Chr.15: 90084223-90084223 on Build GRCh38
rs979570697 Chr.15: 90084076-90084076 on Build GRCh38
rs772175496 Chr.15: 90084348-90084349 on Build GRCh38
rs766269621 Chr.15: 90084306-90084306 on Build GRCh38
rs537451132 Chr.15: 90084216-90084216 on Build GRCh38
rs778156657 Chr.15: 90084250-90084250 on Build GRCh38
rs1008022729 Chr.15: 90084182-90084182 on Build GRCh38
rs775400107 Chr.15: 90084201-90084201 on Build GRCh38
rs551836169 Chr.15: 90084198-90084198 on Build GRCh38
rs1052682563 Chr.15: 90084164-90084164 on Build GRCh38
rs1029959105 Chr.15: 90084253-90084253 on Build GRCh38
rs374082123 Chr.15: 90084077-90084077 on Build GRCh38
rs1019874817 Chr.15: 90084190-90084190 on Build GRCh38
rs1046320242 Chr.15: 90083926-90083926 on Build GRCh38
rs781761976 Chr.15: 90084348-90084348 on Build GRCh38
rs762504266 Chr.15: 90084176-90084176 on Build GRCh38
rs753517179 Chr.15: 90084230-90084230 on Build GRCh38
rs970553405 Chr.15: 90084279-90084279 on Build GRCh38
rs1037284467 Chr.15: 90084008-90084008 on Build GRCh38
rs115830969 Chr.15: 90084197-90084197 on Build GRCh38
rs928867570 Chr.15: 90083935-90083935 on Build GRCh38

View Full Product Details