Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90084152-90084656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494433 c.1304C>T p.T435M Chr.15: 90084321-90084321 on Build GRCh38
269346 c.1322T>C p.I441T Chr.15: 90084303-90084303 on Build GRCh38
4691742 c.1324A>G p.K442E Chr.15: 90084301-90084301 on Build GRCh38
5784561 c.1325A>G p.K442R Chr.15: 90084300-90084300 on Build GRCh38
6500191 c.1314C>T p.L438L Chr.15: 90084311-90084311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372655581 Chr.15: 90084236-90084236 on Build GRCh38
rs1051347349 Chr.15: 90084574-90084574 on Build GRCh38
rs990164993 Chr.15: 90084220-90084220 on Build GRCh38
rs954587473 Chr.15: 90084256-90084256 on Build GRCh38
rs773670536 Chr.15: 90084228-90084228 on Build GRCh38
rs764864079 Chr.15: 90084320-90084320 on Build GRCh38
rs920191176 Chr.15: 90084347-90084347 on Build GRCh38
rs560463373 Chr.15: 90084513-90084513 on Build GRCh38
rs1007872814 Chr.15: 90084589-90084589 on Build GRCh38
rs765526162 Chr.15: 90084215-90084215 on Build GRCh38
rs942251460 Chr.15: 90084449-90084449 on Build GRCh38
rs758692398 Chr.15: 90084537-90084537 on Build GRCh38
rs779080885 Chr.15: 90084476-90084476 on Build GRCh38
rs764173046 Chr.15: 90084274-90084274 on Build GRCh38
rs535303053 Chr.15: 90084243-90084243 on Build GRCh38
rs150574164 Chr.15: 90084323-90084323 on Build GRCh38
rs571080145 Chr.15: 90084193-90084193 on Build GRCh38
rs112656331 Chr.15: 90084354-90084354 on Build GRCh38
rs767032049 Chr.15: 90084265-90084265 on Build GRCh38
rs999442699 Chr.15: 90084200-90084200 on Build GRCh38
rs551629636 Chr.15: 90084526-90084526 on Build GRCh38
rs750420733 Chr.15: 90084536-90084536 on Build GRCh38
rs890132028 Chr.15: 90084578-90084578 on Build GRCh38
rs540113594 Chr.15: 90084401-90084401 on Build GRCh38
rs1017825906 Chr.15: 90084289-90084289 on Build GRCh38
rs749473740 Chr.15: 90084380-90084380 on Build GRCh38
rs778604514 Chr.15: 90084239-90084239 on Build GRCh38
rs993157282 Chr.15: 90084237-90084237 on Build GRCh38
rs893748191 Chr.15: 90084525-90084525 on Build GRCh38
rs1040841598 Chr.15: 90084450-90084450 on Build GRCh38
rs148745668 Chr.15: 90084280-90084280 on Build GRCh38
rs112930264 Chr.15: 90084381-90084381 on Build GRCh38
rs368733189 Chr.15: 90084335-90084335 on Build GRCh38
rs760563629 Chr.15: 90084282-90084282 on Build GRCh38
rs999391853 Chr.15: 90084495-90084495 on Build GRCh38
rs757889221 Chr.15: 90084346-90084346 on Build GRCh38
rs750311980 Chr.15: 90084221-90084221 on Build GRCh38
rs542615343 Chr.15: 90084383-90084383 on Build GRCh38
rs771456529 Chr.15: 90084257-90084257 on Build GRCh38
rs200094442 Chr.15: 90084262-90084262 on Build GRCh38
rs182292189 Chr.15: 90084552-90084552 on Build GRCh38
rs763078146 Chr.15: 90084217-90084217 on Build GRCh38
rs754752564 Chr.15: 90084313-90084313 on Build GRCh38
rs201015211 Chr.15: 90084266-90084266 on Build GRCh38
rs926764715 Chr.15: 90084409-90084410 on Build GRCh38
rs767424579 Chr.15: 90084219-90084219 on Build GRCh38
rs118148483 Chr.15: 90084618-90084618 on Build GRCh38
rs77026878 Chr.15: 90084624-90084624 on Build GRCh38
rs761327377 Chr.15: 90084603-90084603 on Build GRCh38
rs765034363 Chr.15: 90084398-90084398 on Build GRCh38
rs578203629 Chr.15: 90084311-90084311 on Build GRCh38
rs747185496 Chr.15: 90084358-90084359 on Build GRCh38
rs747501657 Chr.15: 90084254-90084254 on Build GRCh38
rs191547948 Chr.15: 90084341-90084341 on Build GRCh38
rs118053940 Chr.15: 90084321-90084321 on Build GRCh38
rs988101235 Chr.15: 90084339-90084339 on Build GRCh38
rs60147683 Chr.15: 90084384-90084384 on Build GRCh38
rs949588153 Chr.15: 90084344-90084344 on Build GRCh38
rs776544516 Chr.15: 90084392-90084392 on Build GRCh38
rs376215927 Chr.15: 90084244-90084244 on Build GRCh38
rs1000001798 Chr.15: 90084611-90084612 on Build GRCh38
rs553627406 Chr.15: 90084271-90084271 on Build GRCh38
rs577520416 Chr.15: 90084203-90084203 on Build GRCh38
rs780204079 Chr.15: 90084351-90084352 on Build GRCh38
rs769152803 Chr.15: 90084360-90084360 on Build GRCh38
rs555429283 Chr.15: 90084223-90084223 on Build GRCh38
rs775593059 Chr.15: 90084370-90084370 on Build GRCh38
rs981931785 Chr.15: 90084546-90084546 on Build GRCh38
rs772175496 Chr.15: 90084348-90084349 on Build GRCh38
rs766269621 Chr.15: 90084306-90084306 on Build GRCh38
rs902340220 Chr.15: 90084469-90084469 on Build GRCh38
rs537451132 Chr.15: 90084216-90084216 on Build GRCh38
rs1037849258 Chr.15: 90084465-90084465 on Build GRCh38
rs528389852 Chr.15: 90084394-90084394 on Build GRCh38
rs1008022729 Chr.15: 90084182-90084182 on Build GRCh38
rs35474233 Chr.15: 90084424-90084425 on Build GRCh38
rs778156657 Chr.15: 90084250-90084250 on Build GRCh38
rs564771834 Chr.15: 90084510-90084510 on Build GRCh38
rs775400107 Chr.15: 90084201-90084201 on Build GRCh38
rs551836169 Chr.15: 90084198-90084198 on Build GRCh38
rs1029959105 Chr.15: 90084253-90084253 on Build GRCh38
rs943778685 Chr.15: 90084418-90084418 on Build GRCh38
rs140057157 Chr.15: 90084600-90084600 on Build GRCh38
rs1019874817 Chr.15: 90084190-90084190 on Build GRCh38
rs746303510 Chr.15: 90084361-90084361 on Build GRCh38
rs781761976 Chr.15: 90084348-90084348 on Build GRCh38
rs762504266 Chr.15: 90084176-90084176 on Build GRCh38
rs970553405 Chr.15: 90084279-90084279 on Build GRCh38
rs753517179 Chr.15: 90084230-90084230 on Build GRCh38
rs527632201 Chr.15: 90084518-90084518 on Build GRCh38
rs115830969 Chr.15: 90084197-90084197 on Build GRCh38

View Full Product Details