Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90087356-90087852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH2
Entrez Gene ID
3418
Gene Name
isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
Gene Aliases
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
UniGene
Hs.596461
Gene Chromosome Location
Chr.15: 90083978-90102554 on Build GRCh38
Transcripts
NM_001289910.1 NM_001290114.1 NM_002168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
340426 c.745A>G p.S249G Chr.15: 90087509-90087509 on Build GRCh38
7271582 c.816+5G>A p.? Chr.15: 90087433-90087433 on Build GRCh38
3377698 c.696G>A p.A232A Chr.15: 90087558-90087558 on Build GRCh38
2139734 c.718A>G p.I240V Chr.15: 90087536-90087536 on Build GRCh38
7229430 c.811G>C p.D271H Chr.15: 90087443-90087443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551167191 Chr.15: 90087734-90087734 on Build GRCh38
rs753805390 Chr.15: 90087588-90087591 on Build GRCh38
rs746192613 Chr.15: 90087516-90087516 on Build GRCh38
rs750862424 Chr.15: 90087596-90087596 on Build GRCh38
rs776428776 Chr.15: 90087620-90087620 on Build GRCh38
rs916075513 Chr.15: 90087783-90087783 on Build GRCh38
rs547762277 Chr.15: 90087679-90087679 on Build GRCh38
rs747382357 Chr.15: 90087396-90087396 on Build GRCh38
rs749395621 Chr.15: 90087536-90087536 on Build GRCh38
rs770109177 Chr.15: 90087519-90087519 on Build GRCh38
rs778437352 Chr.15: 90087505-90087505 on Build GRCh38
rs372508630 Chr.15: 90087579-90087579 on Build GRCh38
rs1034158653 Chr.15: 90087778-90087778 on Build GRCh38
rs754701146 Chr.15: 90087494-90087494 on Build GRCh38
rs757725772 Chr.15: 90087508-90087508 on Build GRCh38
rs869221372 Chr.15: 90087807-90087807 on Build GRCh38
rs781648958 Chr.15: 90087512-90087512 on Build GRCh38
rs900252735 Chr.15: 90087703-90087703 on Build GRCh38
rs369432003 Chr.15: 90087780-90087780 on Build GRCh38
rs762925398 Chr.15: 90087424-90087424 on Build GRCh38
rs118101777 Chr.15: 90087472-90087472 on Build GRCh38
rs753466539 Chr.15: 90087484-90087484 on Build GRCh38
rs770959675 Chr.15: 90087619-90087619 on Build GRCh38
rs9972327 Chr.15: 90087660-90087660 on Build GRCh38
rs369238286 Chr.15: 90087578-90087578 on Build GRCh38
rs746060098 Chr.15: 90087406-90087406 on Build GRCh38
rs992306773 Chr.15: 90087668-90087668 on Build GRCh38
rs779006310 Chr.15: 90087612-90087612 on Build GRCh38
rs769872742 Chr.15: 90087408-90087408 on Build GRCh38
rs762573789 Chr.15: 90087583-90087583 on Build GRCh38
rs773159667 Chr.15: 90087549-90087549 on Build GRCh38
rs755875875 Chr.15: 90087391-90087391 on Build GRCh38
rs552868601 Chr.15: 90087672-90087672 on Build GRCh38
rs760479966 Chr.15: 90087466-90087466 on Build GRCh38
rs764775925 Chr.15: 90087495-90087495 on Build GRCh38
rs995233996 Chr.15: 90087634-90087634 on Build GRCh38
rs763991101 Chr.15: 90087392-90087393 on Build GRCh38
rs760567997 Chr.15: 90087555-90087555 on Build GRCh38
rs141606558 Chr.15: 90087724-90087724 on Build GRCh38
rs953461103 Chr.15: 90087674-90087674 on Build GRCh38
rs751080545 Chr.15: 90087585-90087585 on Build GRCh38
rs545968410 Chr.15: 90087628-90087628 on Build GRCh38
rs780131378 Chr.15: 90087524-90087524 on Build GRCh38
rs941758010 Chr.15: 90087404-90087404 on Build GRCh38
rs979919132 Chr.15: 90087656-90087656 on Build GRCh38
rs752232116 Chr.15: 90087582-90087582 on Build GRCh38
rs374859560 Chr.15: 90087500-90087500 on Build GRCh38
rs1051608509 Chr.15: 90087725-90087725 on Build GRCh38
rs373169753 Chr.15: 90087426-90087426 on Build GRCh38
rs777013831 Chr.15: 90087403-90087403 on Build GRCh38
rs147978726 Chr.15: 90087627-90087627 on Build GRCh38
rs1043032321 Chr.15: 90087750-90087750 on Build GRCh38
rs903820154 Chr.15: 90087751-90087751 on Build GRCh38
rs569753415 Chr.15: 90087776-90087776 on Build GRCh38
rs931049841 Chr.15: 90087792-90087792 on Build GRCh38
rs941738910 Chr.15: 90087691-90087691 on Build GRCh38
rs781533725 Chr.15: 90087594-90087594 on Build GRCh38
rs965264504 Chr.15: 90087393-90087395 on Build GRCh38
rs748205361 Chr.15: 90087615-90087615 on Build GRCh38
rs1039159478 Chr.15: 90087695-90087695 on Build GRCh38
rs891660848 Chr.15: 90087727-90087727 on Build GRCh38
rs564635839 Chr.15: 90087629-90087629 on Build GRCh38
rs112284099 Chr.15: 90087437-90087437 on Build GRCh38
rs976570252 Chr.15: 90087394-90087394 on Build GRCh38
rs752310010 Chr.15: 90087498-90087498 on Build GRCh38
rs749466731 Chr.15: 90087611-90087611 on Build GRCh38
rs758227481 Chr.15: 90087804-90087804 on Build GRCh38
rs959766978 Chr.15: 90087661-90087661 on Build GRCh38
rs768462486 Chr.15: 90087506-90087506 on Build GRCh38
rs74743859 Chr.15: 90087780-90087780 on Build GRCh38
rs73469824 Chr.15: 90087647-90087647 on Build GRCh38
rs561924891 Chr.15: 90087671-90087671 on Build GRCh38
rs62019175 Chr.15: 90087805-90087805 on Build GRCh38
rs757290086 Chr.15: 90087780-90087781 on Build GRCh38
rs75551396 Chr.15: 90087804-90087804 on Build GRCh38
rs374285884 Chr.15: 90087610-90087610 on Build GRCh38
rs372473577 Chr.15: 90087690-90087690 on Build GRCh38
rs914590416 Chr.15: 90087449-90087449 on Build GRCh38
rs62019176 Chr.15: 90087807-90087807 on Build GRCh38
rs770825833 Chr.15: 90087558-90087558 on Build GRCh38
rs773071767 Chr.15: 90087439-90087439 on Build GRCh38
rs780339588 Chr.15: 90087606-90087606 on Build GRCh38
rs776539314 Chr.15: 90087572-90087572 on Build GRCh38
rs763654985 Chr.15: 90087584-90087584 on Build GRCh38
rs967411754 Chr.15: 90087723-90087723 on Build GRCh38
rs148671440 Chr.15: 90087542-90087542 on Build GRCh38
rs187234596 Chr.15: 90087602-90087602 on Build GRCh38
rs983692780 Chr.15: 90087673-90087673 on Build GRCh38
rs991650089 Chr.15: 90087779-90087782 on Build GRCh38
rs775684770 Chr.15: 90087420-90087420 on Build GRCh38
rs769048437 Chr.15: 90087806-90087807 on Build GRCh38
rs376293307 Chr.15: 90087603-90087603 on Build GRCh38
rs1048020030 Chr.15: 90087640-90087640 on Build GRCh38
rs753633558 Chr.15: 90087389-90087389 on Build GRCh38
rs538020019 Chr.15: 90087801-90087801 on Build GRCh38
rs77211768 Chr.15: 90087398-90087398 on Build GRCh38

View Full Product Details