Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90231274-90231778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB1
Entrez Gene ID
10519
Gene Name
calcium and integrin binding 1
Gene Aliases
CIB, CIBP, KIP1, PRKDCIP, SIP2-28
UniGene
Hs.715556
Gene Chromosome Location
Chr.15: 90230245-90265759 on Build GRCh38
Transcripts
NM_001277764.1 NM_006384.3 NR_102427.1 NR_102428.1 XM_006720375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235437 c.302C>A p.T101K Chr.15: 90231401-90231401 on Build GRCh38
5587497 c.336C>A p.F112L Chr.15: 90231367-90231367 on Build GRCh38
6341536 c.311C>A p.P104Q Chr.15: 90231392-90231392 on Build GRCh38
4672424 c.238T>C p.S80P Chr.15: 90231465-90231465 on Build GRCh38
3794513 c.318C>T p.I106I Chr.15: 90231385-90231385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975619732 Chr.15: 90231392-90231392 on Build GRCh38
rs756485209 Chr.15: 90231548-90231548 on Build GRCh38
rs2073704 Chr.15: 90231755-90231755 on Build GRCh38
rs562237879 Chr.15: 90231459-90231459 on Build GRCh38
rs139764460 Chr.15: 90231404-90231404 on Build GRCh38
rs1804798 Chr.15: 90231434-90231434 on Build GRCh38
rs754731051 Chr.15: 90231407-90231407 on Build GRCh38
rs368270679 Chr.15: 90231335-90231335 on Build GRCh38
rs769989187 Chr.15: 90231376-90231376 on Build GRCh38
rs574325000 Chr.15: 90231558-90231558 on Build GRCh38
rs757866965 Chr.15: 90231427-90231427 on Build GRCh38
rs771284505 Chr.15: 90231313-90231313 on Build GRCh38
rs1022459767 Chr.15: 90231497-90231497 on Build GRCh38
rs780400910 Chr.15: 90231556-90231556 on Build GRCh38
rs764955332 Chr.15: 90231417-90231417 on Build GRCh38
rs892648982 Chr.15: 90231464-90231464 on Build GRCh38
rs778172578 Chr.15: 90231356-90231356 on Build GRCh38
rs775594329 Chr.15: 90231380-90231380 on Build GRCh38
rs774485052 Chr.15: 90231511-90231511 on Build GRCh38
rs201156295 Chr.15: 90231573-90231574 on Build GRCh38
rs1025979890 Chr.15: 90231664-90231664 on Build GRCh38
rs766485187 Chr.15: 90231419-90231419 on Build GRCh38
rs753545325 Chr.15: 90231338-90231338 on Build GRCh38
rs756617816 Chr.15: 90231724-90231724 on Build GRCh38
rs760594992 Chr.15: 90231397-90231397 on Build GRCh38
rs746208399 Chr.15: 90231462-90231462 on Build GRCh38
rs1139757 Chr.15: 90231461-90231461 on Build GRCh38
rs990248428 Chr.15: 90231617-90231617 on Build GRCh38
rs747471993 Chr.15: 90231435-90231435 on Build GRCh38
rs1055990785 Chr.15: 90231755-90231756 on Build GRCh38
rs1034672392 Chr.15: 90231590-90231590 on Build GRCh38
rs544258960 Chr.15: 90231691-90231691 on Build GRCh38
rs767334293 Chr.15: 90231333-90231333 on Build GRCh38
rs775605318 Chr.15: 90231488-90231488 on Build GRCh38
rs535337679 Chr.15: 90231348-90231348 on Build GRCh38
rs773349164 Chr.15: 90231510-90231512 on Build GRCh38
rs766401205 Chr.15: 90231518-90231518 on Build GRCh38
rs777276144 Chr.15: 90231430-90231430 on Build GRCh38
rs946003599 Chr.15: 90231429-90231429 on Build GRCh38
rs771210225 Chr.15: 90231366-90231366 on Build GRCh38
rs369459169 Chr.15: 90231521-90231521 on Build GRCh38
rs371302257 Chr.15: 90231624-90231624 on Build GRCh38
rs761868095 Chr.15: 90231329-90231329 on Build GRCh38
rs11551249 Chr.15: 90231447-90231447 on Build GRCh38
rs943302957 Chr.15: 90231738-90231738 on Build GRCh38
rs766344209 Chr.15: 90231402-90231402 on Build GRCh38
rs910498462 Chr.15: 90231393-90231393 on Build GRCh38
rs555211318 Chr.15: 90231603-90231603 on Build GRCh38
rs765301247 Chr.15: 90231327-90231327 on Build GRCh38
rs776910503 Chr.15: 90231368-90231368 on Build GRCh38
rs759819987 Chr.15: 90231371-90231371 on Build GRCh38
rs74037065 Chr.15: 90231602-90231602 on Build GRCh38
rs757998537 Chr.15: 90231532-90231532 on Build GRCh38
rs751067207 Chr.15: 90231535-90231535 on Build GRCh38
rs750241867 Chr.15: 90231314-90231314 on Build GRCh38
rs757809091 Chr.15: 90231538-90231538 on Build GRCh38
rs971794373 Chr.15: 90231627-90231627 on Build GRCh38
rs965636782 Chr.15: 90231598-90231598 on Build GRCh38
rs142784235 Chr.15: 90231394-90231394 on Build GRCh38
rs11551250 Chr.15: 90231386-90231386 on Build GRCh38
rs187267727 Chr.15: 90231653-90231653 on Build GRCh38
rs772677858 Chr.15: 90231354-90231354 on Build GRCh38
rs746332906 Chr.15: 90231547-90231547 on Build GRCh38
rs367612323 Chr.15: 90231667-90231667 on Build GRCh38
rs757753639 Chr.15: 90231453-90231453 on Build GRCh38
rs372502181 Chr.15: 90231341-90231341 on Build GRCh38
rs557352577 Chr.15: 90231503-90231503 on Build GRCh38
rs770042283 Chr.15: 90231477-90231477 on Build GRCh38
rs779937416 Chr.15: 90231337-90231337 on Build GRCh38
rs755886403 Chr.15: 90231336-90231336 on Build GRCh38
rs767654753 Chr.15: 90231661-90231661 on Build GRCh38
rs763574675 Chr.15: 90231534-90231534 on Build GRCh38
rs753144431 Chr.15: 90231696-90231696 on Build GRCh38
rs1011455379 Chr.15: 90231466-90231466 on Build GRCh38
rs759438036 Chr.15: 90231522-90231522 on Build GRCh38
rs781640515 Chr.15: 90231542-90231542 on Build GRCh38
rs932085689 Chr.15: 90231615-90231615 on Build GRCh38
rs747497642 Chr.15: 90231365-90231365 on Build GRCh38
rs550044308 Chr.15: 90231326-90231326 on Build GRCh38
rs760591797 Chr.15: 90231514-90231514 on Build GRCh38
rs375148975 Chr.15: 90231349-90231349 on Build GRCh38
rs762909077 Chr.15: 90231387-90231387 on Build GRCh38
rs143773090 Chr.15: 90231489-90231489 on Build GRCh38
rs147405990 Chr.15: 90231395-90231395 on Build GRCh38
rs575974877 Chr.15: 90231531-90231531 on Build GRCh38
rs769409462 Chr.15: 90231469-90231471 on Build GRCh38
rs765073668 Chr.15: 90231529-90231529 on Build GRCh38

View Full Product Details