Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90930842-90931339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC3
Entrez Gene ID
374659
Gene Name
HD domain containing 3
Gene Aliases
(ppGpp)ase, MESH1
UniGene
Hs.349979
Gene Chromosome Location
Chr.15: 90930918-90932569 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
UniGene
Hs.389461
Gene Chromosome Location
Chr.15: 90929980-90954093 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5705987 c.520C>T p.Q174* Chr.15: 90931295-90931295 on Build GRCh38
6860812 c.518A>G p.K173R Chr.15: 90931297-90931297 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113365430 Chr.15: 90931212-90931212 on Build GRCh38
rs1020309451 Chr.15: 90930936-90930936 on Build GRCh38
rs553480439 Chr.15: 90931016-90931016 on Build GRCh38
rs554761360 Chr.15: 90931281-90931282 on Build GRCh38
rs117513973 Chr.15: 90930875-90930875 on Build GRCh38
rs375635949 Chr.15: 90930887-90930887 on Build GRCh38
rs886785319 Chr.15: 90931056-90931056 on Build GRCh38
rs115859518 Chr.15: 90931096-90931096 on Build GRCh38
rs78194647 Chr.15: 90931249-90931249 on Build GRCh38
rs865900648 Chr.15: 90931280-90931281 on Build GRCh38
rs1026484275 Chr.15: 90931227-90931227 on Build GRCh38
rs13252 Chr.15: 90931133-90931133 on Build GRCh38
rs545038439 Chr.15: 90931178-90931178 on Build GRCh38
rs1052528 Chr.15: 90931065-90931065 on Build GRCh38
rs1029210081 Chr.15: 90930917-90930917 on Build GRCh38
rs954703724 Chr.15: 90930919-90930919 on Build GRCh38
rs756850703 Chr.15: 90931272-90931282 on Build GRCh38
rs987475188 Chr.15: 90930929-90930929 on Build GRCh38
rs925786654 Chr.15: 90930996-90930996 on Build GRCh38
rs1016401556 Chr.15: 90930908-90930908 on Build GRCh38
rs991395372 Chr.15: 90931014-90931014 on Build GRCh38
rs536721315 Chr.15: 90931159-90931159 on Build GRCh38
rs766234266 Chr.15: 90930912-90930912 on Build GRCh38
rs958562535 Chr.15: 90931287-90931287 on Build GRCh38
rs935860859 Chr.15: 90931001-90931001 on Build GRCh38
rs560935984 Chr.15: 90931245-90931245 on Build GRCh38
rs987539520 Chr.15: 90931241-90931241 on Build GRCh38
rs899346733 Chr.15: 90930911-90930911 on Build GRCh38
rs1033958409 Chr.15: 90931184-90931184 on Build GRCh38
rs996785220 Chr.15: 90931115-90931115 on Build GRCh38
rs751706004 Chr.15: 90930913-90930913 on Build GRCh38
rs925798831 Chr.15: 90930973-90930973 on Build GRCh38
rs1038528848 Chr.15: 90931076-90931076 on Build GRCh38
rs890733450 Chr.15: 90931201-90931201 on Build GRCh38
rs1008841050 Chr.15: 90931208-90931208 on Build GRCh38
rs556806824 Chr.15: 90931211-90931211 on Build GRCh38
rs1013919774 Chr.15: 90931033-90931033 on Build GRCh38
rs917209688 Chr.15: 90931021-90931021 on Build GRCh38
rs949682307 Chr.15: 90931036-90931036 on Build GRCh38
rs757243903 Chr.15: 90931291-90931291 on Build GRCh38
rs576731844 Chr.15: 90931235-90931235 on Build GRCh38
rs1002885222 Chr.15: 90931142-90931142 on Build GRCh38
rs1046789925 Chr.15: 90931039-90931039 on Build GRCh38
rs1052532 Chr.15: 90931006-90931006 on Build GRCh38
rs1029200371 Chr.15: 90931131-90931131 on Build GRCh38
rs1038922583 Chr.15: 90930931-90930931 on Build GRCh38
rs967518450 Chr.15: 90930977-90930977 on Build GRCh38
rs911894830 Chr.15: 90931248-90931248 on Build GRCh38
rs967802089 Chr.15: 90931230-90931230 on Build GRCh38
rs779859569 Chr.15: 90931265-90931265 on Build GRCh38
rs752756931 Chr.15: 90931111-90931111 on Build GRCh38
rs768366806 Chr.15: 90931292-90931292 on Build GRCh38
rs759667141 Chr.15: 90931127-90931127 on Build GRCh38
rs941094792 Chr.15: 90931057-90931057 on Build GRCh38
rs555682386 Chr.15: 90930873-90930873 on Build GRCh38
rs979268009 Chr.15: 90930978-90930978 on Build GRCh38

View Full Product Details