Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90931239-90931743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC3
Entrez Gene ID
374659
Gene Name
HD domain containing 3
Gene Aliases
(ppGpp)ase, MESH1
UniGene
Hs.349979
Gene Chromosome Location
Chr.15: 90930918-90932569 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
UniGene
Hs.389461
Gene Chromosome Location
Chr.15: 90929980-90954093 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588238 c.418G>A p.E140K Chr.15: 90931397-90931397 on Build GRCh38
6678365 c.415A>C p.I139L Chr.15: 90931551-90931551 on Build GRCh38
5705987 c.520C>T p.Q174* Chr.15: 90931295-90931295 on Build GRCh38
6318878 c.421C>A p.H141N Chr.15: 90931394-90931394 on Build GRCh38
5479020 c.441C>T p.F147F Chr.15: 90931374-90931374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202109806 Chr.15: 90931559-90931559 on Build GRCh38
rs957292376 Chr.15: 90931693-90931693 on Build GRCh38
rs777600971 Chr.15: 90931606-90931606 on Build GRCh38
rs117318472 Chr.15: 90931401-90931401 on Build GRCh38
rs749359481 Chr.15: 90931681-90931681 on Build GRCh38
rs554761360 Chr.15: 90931281-90931282 on Build GRCh38
rs9806656 Chr.15: 90931512-90931512 on Build GRCh38
rs776145083 Chr.15: 90931410-90931411 on Build GRCh38
rs768487452 Chr.15: 90931332-90931332 on Build GRCh38
rs1052535 Chr.15: 90931508-90931508 on Build GRCh38
rs865900648 Chr.15: 90931280-90931281 on Build GRCh38
rs549689339 Chr.15: 90931522-90931522 on Build GRCh38
rs766046997 Chr.15: 90931446-90931446 on Build GRCh38
rs771591823 Chr.15: 90931712-90931712 on Build GRCh38
rs764514278 Chr.15: 90931413-90931413 on Build GRCh38
rs773755660 Chr.15: 90931709-90931709 on Build GRCh38
rs780159584 Chr.15: 90931680-90931680 on Build GRCh38
rs746943969 Chr.15: 90931644-90931644 on Build GRCh38
rs756850703 Chr.15: 90931272-90931282 on Build GRCh38
rs995051168 Chr.15: 90931487-90931487 on Build GRCh38
rs369746305 Chr.15: 90931561-90931561 on Build GRCh38
rs748980570 Chr.15: 90931330-90931330 on Build GRCh38
rs771948355 Chr.15: 90931373-90931373 on Build GRCh38
rs764825110 Chr.15: 90931587-90931587 on Build GRCh38
rs958562535 Chr.15: 90931287-90931287 on Build GRCh38
rs776163027 Chr.15: 90931713-90931713 on Build GRCh38
rs199900582 Chr.15: 90931700-90931700 on Build GRCh38
rs949916887 Chr.15: 90931361-90931361 on Build GRCh38
rs762089808 Chr.15: 90931550-90931550 on Build GRCh38
rs1038116581 Chr.15: 90931416-90931416 on Build GRCh38
rs535250483 Chr.15: 90931599-90931599 on Build GRCh38
rs965144381 Chr.15: 90931620-90931620 on Build GRCh38
rs771171537 Chr.15: 90931347-90931347 on Build GRCh38
rs774617992 Chr.15: 90931414-90931414 on Build GRCh38
rs550617721 Chr.15: 90931449-90931449 on Build GRCh38
rs765959972 Chr.15: 90931570-90931570 on Build GRCh38
rs761258820 Chr.15: 90931630-90931630 on Build GRCh38
rs753043739 Chr.15: 90931398-90931398 on Build GRCh38
rs748092184 Chr.15: 90931370-90931370 on Build GRCh38
rs913032502 Chr.15: 90931362-90931362 on Build GRCh38
rs755701056 Chr.15: 90931444-90931444 on Build GRCh38
rs754606855 Chr.15: 90931580-90931580 on Build GRCh38
rs757243903 Chr.15: 90931291-90931291 on Build GRCh38
rs772563160 Chr.15: 90931703-90931703 on Build GRCh38
rs111567876 Chr.15: 90931409-90931409 on Build GRCh38
rs768170153 Chr.15: 90931433-90931433 on Build GRCh38
rs745887826 Chr.15: 90931511-90931511 on Build GRCh38
rs1033079309 Chr.15: 90931692-90931692 on Build GRCh38
rs1019053188 Chr.15: 90931617-90931617 on Build GRCh38
rs935916081 Chr.15: 90931329-90931329 on Build GRCh38
rs974810102 Chr.15: 90931658-90931658 on Build GRCh38
rs903150688 Chr.15: 90931652-90931652 on Build GRCh38
rs868842776 Chr.15: 90931554-90931554 on Build GRCh38
rs779018863 Chr.15: 90931694-90931694 on Build GRCh38
rs570312956 Chr.15: 90931451-90931451 on Build GRCh38
rs921053482 Chr.15: 90931500-90931500 on Build GRCh38
rs761245831 Chr.15: 90931710-90931710 on Build GRCh38
rs371199456 Chr.15: 90931659-90931659 on Build GRCh38
rs768784013 Chr.15: 90931537-90931537 on Build GRCh38
rs749888237 Chr.15: 90931436-90931436 on Build GRCh38
rs774738103 Chr.15: 90931539-90931539 on Build GRCh38
rs762332074 Chr.15: 90931431-90931431 on Build GRCh38
rs372856910 Chr.15: 90931721-90931721 on Build GRCh38
rs779859569 Chr.15: 90931265-90931265 on Build GRCh38
rs768857344 Chr.15: 90931691-90931691 on Build GRCh38
rs768366806 Chr.15: 90931292-90931292 on Build GRCh38
rs376652457 Chr.15: 90931598-90931598 on Build GRCh38
rs532110423 Chr.15: 90931410-90931410 on Build GRCh38
rs768081934 Chr.15: 90931553-90931553 on Build GRCh38
rs778643169 Chr.15: 90931358-90931358 on Build GRCh38
rs373482128 Chr.15: 90931578-90931578 on Build GRCh38
rs748441338 Chr.15: 90931698-90931698 on Build GRCh38
rs567753381 Chr.15: 90931532-90931532 on Build GRCh38

View Full Product Details