Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90931964-90932456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDDC3
Entrez Gene ID
374659
Gene Name
HD domain containing 3
Gene Aliases
(ppGpp)ase, MESH1
UniGene
Hs.349979
Gene Chromosome Location
Chr.15: 90930918-90932569 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
UniGene
Hs.389461
Gene Chromosome Location
Chr.15: 90929980-90954093 on Build GRCh38
Transcripts
NM_001286451.1 NM_198527.3 NR_104447.1 NM_001039675.1 XM_011521779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7370828 c.160G>A p.V54M Chr.15: 90932063-90932063 on Build GRCh38
7370829 c.160G>A p.V54M Chr.15: 90932063-90932063 on Build GRCh38
7186760 c.155T>C p.I52T Chr.15: 90932068-90932068 on Build GRCh38
7186761 c.155T>C p.I52T Chr.15: 90932068-90932068 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747721755 Chr.15: 90932389-90932389 on Build GRCh38
rs114863118 Chr.15: 90932181-90932181 on Build GRCh38
rs371163282 Chr.15: 90931990-90931990 on Build GRCh38
rs573111430 Chr.15: 90932376-90932376 on Build GRCh38
rs528643436 Chr.15: 90932170-90932170 on Build GRCh38
rs756487914 Chr.15: 90932150-90932150 on Build GRCh38
rs759252883 Chr.15: 90932206-90932206 on Build GRCh38
rs199905818 Chr.15: 90932416-90932416 on Build GRCh38
rs554531112 Chr.15: 90932362-90932362 on Build GRCh38
rs563508946 Chr.15: 90932404-90932404 on Build GRCh38
rs780552478 Chr.15: 90932083-90932083 on Build GRCh38
rs575395909 Chr.15: 90932316-90932316 on Build GRCh38
rs149008342 Chr.15: 90932088-90932088 on Build GRCh38
rs773889911 Chr.15: 90932100-90932100 on Build GRCh38
rs1040553824 Chr.15: 90932339-90932339 on Build GRCh38
rs34697308 Chr.15: 90932307-90932308 on Build GRCh38
rs929651470 Chr.15: 90932236-90932236 on Build GRCh38
rs571363310 Chr.15: 90932217-90932217 on Build GRCh38
rs927082675 Chr.15: 90932165-90932165 on Build GRCh38
rs1010225787 Chr.15: 90932411-90932411 on Build GRCh38
rs778122085 Chr.15: 90932433-90932434 on Build GRCh38
rs974032496 Chr.15: 90932149-90932149 on Build GRCh38
rs558794439 Chr.15: 90932423-90932423 on Build GRCh38
rs757374020 Chr.15: 90932143-90932143 on Build GRCh38
rs567984929 Chr.15: 90932146-90932146 on Build GRCh38
rs776700535 Chr.15: 90932119-90932119 on Build GRCh38
rs894445784 Chr.15: 90932387-90932387 on Build GRCh38
rs751711750 Chr.15: 90932141-90932141 on Build GRCh38
rs752762730 Chr.15: 90932017-90932017 on Build GRCh38
rs111756313 Chr.15: 90932035-90932035 on Build GRCh38
rs369274774 Chr.15: 90932042-90932042 on Build GRCh38
rs549390607 Chr.15: 90932123-90932123 on Build GRCh38
rs771553834 Chr.15: 90932398-90932398 on Build GRCh38
rs780462581 Chr.15: 90932151-90932151 on Build GRCh38
rs767812419 Chr.15: 90932144-90932144 on Build GRCh38
rs556870351 Chr.15: 90932300-90932300 on Build GRCh38
rs973792236 Chr.15: 90932196-90932196 on Build GRCh38
rs924816172 Chr.15: 90932301-90932301 on Build GRCh38
rs34967259 Chr.15: 90932044-90932044 on Build GRCh38
rs754310590 Chr.15: 90932160-90932160 on Build GRCh38
rs761592237 Chr.15: 90931986-90931986 on Build GRCh38
rs978020228 Chr.15: 90932283-90932283 on Build GRCh38
rs746267075 Chr.15: 90932054-90932054 on Build GRCh38
rs912346957 Chr.15: 90932207-90932207 on Build GRCh38
rs781390517 Chr.15: 90932052-90932052 on Build GRCh38
rs953716929 Chr.15: 90932184-90932184 on Build GRCh38
rs752989734 Chr.15: 90932404-90932404 on Build GRCh38
rs747731076 Chr.15: 90932102-90932102 on Build GRCh38
rs986480631 Chr.15: 90932194-90932194 on Build GRCh38
rs778514672 Chr.15: 90932171-90932171 on Build GRCh38
rs375560756 Chr.15: 90931992-90931992 on Build GRCh38
rs772838683 Chr.15: 90932110-90932110 on Build GRCh38
rs752839383 Chr.15: 90932061-90932063 on Build GRCh38
rs1032987575 Chr.15: 90932032-90932032 on Build GRCh38
rs1046752728 Chr.15: 90932256-90932256 on Build GRCh38
rs368232713 Chr.15: 90932008-90932008 on Build GRCh38
rs371686064 Chr.15: 90932148-90932148 on Build GRCh38
rs538760292 Chr.15: 90932222-90932222 on Build GRCh38
rs893206995 Chr.15: 90932385-90932385 on Build GRCh38
rs568910840 Chr.15: 90932346-90932346 on Build GRCh38
rs771775211 Chr.15: 90932108-90932108 on Build GRCh38
rs944857147 Chr.15: 90932214-90932214 on Build GRCh38
rs536683116 Chr.15: 90932358-90932358 on Build GRCh38
rs760519177 Chr.15: 90932115-90932115 on Build GRCh38
rs766367179 Chr.15: 90932010-90932010 on Build GRCh38
rs557074965 Chr.15: 90932296-90932296 on Build GRCh38
rs368111215 Chr.15: 90932112-90932112 on Build GRCh38
rs1054714858 Chr.15: 90932365-90932365 on Build GRCh38
rs996629730 Chr.15: 90931999-90931999 on Build GRCh38
rs758569594 Chr.15: 90932024-90932024 on Build GRCh38
rs766364799 Chr.15: 90932118-90932118 on Build GRCh38
rs1054490583 Chr.15: 90932363-90932363 on Build GRCh38
rs764240110 Chr.15: 90932129-90932129 on Build GRCh38
rs773106026 Chr.15: 90931994-90931994 on Build GRCh38
rs756539452 Chr.15: 90932069-90932069 on Build GRCh38
rs868576419 Chr.15: 90931987-90931987 on Build GRCh38
rs944913105 Chr.15: 90932318-90932318 on Build GRCh38

View Full Product Details