Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92472319-92472829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
-
UniGene
Hs.367879
Gene Chromosome Location
Chr.15: 92465762-92501117 on Build GRCh38
Transcripts
NM_001301106.1 NM_153040.2 XM_011521257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279286 c.364C>A p.L122I Chr.15: 92472512-92472512 on Build GRCh38
5580392 c.238A>T p.K80* Chr.15: 92472386-92472386 on Build GRCh38
335031 c.296C>T p.P99L Chr.15: 92472444-92472444 on Build GRCh38
1246509 c.228A>G p.Q76Q Chr.15: 92472376-92472376 on Build GRCh38
5661228 c.274G>A p.V92M Chr.15: 92472422-92472422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010198478 Chr.15: 92472787-92472787 on Build GRCh38
rs558478969 Chr.15: 92472360-92472360 on Build GRCh38
rs200198289 Chr.15: 92472530-92472530 on Build GRCh38
rs747985033 Chr.15: 92472428-92472428 on Build GRCh38
rs1018825960 Chr.15: 92472638-92472638 on Build GRCh38
rs147905379 Chr.15: 92472460-92472460 on Build GRCh38
rs1004706502 Chr.15: 92472618-92472618 on Build GRCh38
rs772440384 Chr.15: 92472566-92472566 on Build GRCh38
rs375995222 Chr.15: 92472768-92472768 on Build GRCh38
rs139702239 Chr.15: 92472399-92472399 on Build GRCh38
rs546349590 Chr.15: 92472616-92472616 on Build GRCh38
rs200241290 Chr.15: 92472521-92472521 on Build GRCh38
rs115555054 Chr.15: 92472607-92472607 on Build GRCh38
rs975919957 Chr.15: 92472436-92472436 on Build GRCh38
rs560555748 Chr.15: 92472737-92472743 on Build GRCh38
rs901142916 Chr.15: 92472796-92472796 on Build GRCh38
rs566333782 Chr.15: 92472649-92472649 on Build GRCh38
rs1054362248 Chr.15: 92472758-92472758 on Build GRCh38
rs766882666 Chr.15: 92472407-92472407 on Build GRCh38
rs761454920 Chr.15: 92472651-92472651 on Build GRCh38
rs949134379 Chr.15: 92472725-92472725 on Build GRCh38
rs916209143 Chr.15: 92472769-92472769 on Build GRCh38
rs952779001 Chr.15: 92472805-92472805 on Build GRCh38
rs996897530 Chr.15: 92472802-92472802 on Build GRCh38
rs201135368 Chr.15: 92472501-92472501 on Build GRCh38
rs765448572 Chr.15: 92472346-92472346 on Build GRCh38
rs892754627 Chr.15: 92472767-92472767 on Build GRCh38
rs768140065 Chr.15: 92472475-92472475 on Build GRCh38
rs187488934 Chr.15: 92472572-92472572 on Build GRCh38
rs564892980 Chr.15: 92472525-92472525 on Build GRCh38
rs749943796 Chr.15: 92472477-92472477 on Build GRCh38
rs200668582 Chr.15: 92472420-92472420 on Build GRCh38
rs769982817 Chr.15: 92472452-92472452 on Build GRCh38
rs966793454 Chr.15: 92472727-92472727 on Build GRCh38
rs74888661 Chr.15: 92472469-92472469 on Build GRCh38
rs528150105 Chr.15: 92472518-92472518 on Build GRCh38
rs1036037214 Chr.15: 92472701-92472701 on Build GRCh38
rs954083182 Chr.15: 92472708-92472708 on Build GRCh38
rs778798547 Chr.15: 92472425-92472425 on Build GRCh38
rs192410256 Chr.15: 92472713-92472713 on Build GRCh38
rs762299340 Chr.15: 92472563-92472563 on Build GRCh38
rs921733603 Chr.15: 92472443-92472443 on Build GRCh38
rs753498131 Chr.15: 92472499-92472499 on Build GRCh38
rs940600876 Chr.15: 92472687-92472687 on Build GRCh38
rs751972171 Chr.15: 92472391-92472391 on Build GRCh38
rs904933204 Chr.15: 92472744-92472744 on Build GRCh38
rs558827379 Chr.15: 92472710-92472710 on Build GRCh38
rs369303581 Chr.15: 92472547-92472547 on Build GRCh38
rs778512212 Chr.15: 92472502-92472502 on Build GRCh38
rs768930647 Chr.15: 92472555-92472555 on Build GRCh38
rs774794972 Chr.15: 92472467-92472467 on Build GRCh38
rs936849977 Chr.15: 92472757-92472757 on Build GRCh38
rs16974290 Chr.15: 92472789-92472789 on Build GRCh38
rs746013772 Chr.15: 92472449-92472449 on Build GRCh38
rs989585443 Chr.15: 92472482-92472482 on Build GRCh38
rs758070926 Chr.15: 92472795-92472795 on Build GRCh38
rs912672284 Chr.15: 92472427-92472427 on Build GRCh38
rs376764424 Chr.15: 92472571-92472571 on Build GRCh38
rs984576192 Chr.15: 92472664-92472664 on Build GRCh38
rs930360616 Chr.15: 92472587-92472587 on Build GRCh38
rs116841563 Chr.15: 92472403-92472403 on Build GRCh38
rs975427324 Chr.15: 92472627-92472627 on Build GRCh38
rs192750505 Chr.15: 92472361-92472361 on Build GRCh38
rs1032626118 Chr.15: 92472577-92472577 on Build GRCh38
rs77687316 Chr.15: 92472491-92472491 on Build GRCh38
rs370367893 Chr.15: 92472385-92472385 on Build GRCh38
rs763356982 Chr.15: 92472373-92472373 on Build GRCh38
rs1009611936 Chr.15: 92472602-92472602 on Build GRCh38
rs140074819 Chr.15: 92472737-92472737 on Build GRCh38
rs776507506 Chr.15: 92472437-92472437 on Build GRCh38
rs764632670 Chr.15: 92472375-92472375 on Build GRCh38
rs749960219 Chr.15: 92472408-92472408 on Build GRCh38
rs755827744 Chr.15: 92472415-92472415 on Build GRCh38
rs772059164 Chr.15: 92472430-92472430 on Build GRCh38
rs755729767 Chr.15: 92472485-92472485 on Build GRCh38
rs957160647 Chr.15: 92472578-92472578 on Build GRCh38
rs368997262 Chr.15: 92472419-92472419 on Build GRCh38
rs1032362678 Chr.15: 92472414-92472414 on Build GRCh38
rs16974288 Chr.15: 92472674-92472674 on Build GRCh38
rs200522254 Chr.15: 92472548-92472548 on Build GRCh38
rs1028710353 Chr.15: 92472706-92472706 on Build GRCh38
rs774577150 Chr.15: 92472558-92472558 on Build GRCh38
rs753249713 Chr.15: 92472347-92472348 on Build GRCh38
rs1028392284 Chr.15: 92472804-92472804 on Build GRCh38

View Full Product Details