Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92472859-92473401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
-
UniGene
Hs.367879
Gene Chromosome Location
Chr.15: 92465762-92501117 on Build GRCh38
Transcripts
NM_001301106.1 NM_153040.2 XM_011521257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238960 c.458A>G p.H153R Chr.15: 92473036-92473036 on Build GRCh38
7265286 c.442G>A p.G148R Chr.15: 92473020-92473020 on Build GRCh38
5094344 c.489G>T p.Q163H Chr.15: 92473067-92473067 on Build GRCh38
3887810 c.443G>A p.G148E Chr.15: 92473021-92473021 on Build GRCh38
1749382 c.479G>C p.R160T Chr.15: 92473057-92473057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574230545 Chr.15: 92473338-92473338 on Build GRCh38
rs541606680 Chr.15: 92473365-92473365 on Build GRCh38
rs758254850 Chr.15: 92473111-92473112 on Build GRCh38
rs759042133 Chr.15: 92472922-92472922 on Build GRCh38
rs971910722 Chr.15: 92473136-92473136 on Build GRCh38
rs753844446 Chr.15: 92473020-92473020 on Build GRCh38
rs751363048 Chr.15: 92472998-92472998 on Build GRCh38
rs776764157 Chr.15: 92473203-92473203 on Build GRCh38
rs764774045 Chr.15: 92472982-92472982 on Build GRCh38
rs771009922 Chr.15: 92473143-92473143 on Build GRCh38
rs1022285108 Chr.15: 92472911-92472911 on Build GRCh38
rs376091739 Chr.15: 92473123-92473123 on Build GRCh38
rs552621753 Chr.15: 92473284-92473284 on Build GRCh38
rs546539334 Chr.15: 92473158-92473158 on Build GRCh38
rs984454530 Chr.15: 92472993-92472993 on Build GRCh38
rs375505441 Chr.15: 92473118-92473118 on Build GRCh38
rs985050893 Chr.15: 92473272-92473272 on Build GRCh38
rs201944005 Chr.15: 92472977-92472977 on Build GRCh38
rs566200852 Chr.15: 92473055-92473056 on Build GRCh38
rs780044885 Chr.15: 92472954-92472954 on Build GRCh38
rs147844486 Chr.15: 92472962-92472962 on Build GRCh38
rs747443692 Chr.15: 92473093-92473093 on Build GRCh38
rs781168082 Chr.15: 92473019-92473019 on Build GRCh38
rs199987769 Chr.15: 92472980-92472980 on Build GRCh38
rs753410236 Chr.15: 92472913-92472913 on Build GRCh38
rs952293072 Chr.15: 92473269-92473269 on Build GRCh38
rs755129481 Chr.15: 92473026-92473026 on Build GRCh38
rs923030431 Chr.15: 92473369-92473369 on Build GRCh38
rs980416524 Chr.15: 92472928-92472928 on Build GRCh38
rs1028277698 Chr.15: 92473063-92473063 on Build GRCh38
rs902529091 Chr.15: 92473187-92473187 on Build GRCh38
rs375500957 Chr.15: 92473188-92473188 on Build GRCh38
rs756957298 Chr.15: 92472948-92472949 on Build GRCh38
rs368578104 Chr.15: 92473091-92473091 on Build GRCh38
rs529893010 Chr.15: 92473152-92473152 on Build GRCh38
rs1040163083 Chr.15: 92473068-92473068 on Build GRCh38
rs1012410079 Chr.15: 92473254-92473254 on Build GRCh38
rs765005965 Chr.15: 92472924-92472924 on Build GRCh38
rs369616986 Chr.15: 92472965-92472965 on Build GRCh38
rs961722564 Chr.15: 92473290-92473290 on Build GRCh38
rs749454956 Chr.15: 92472956-92472956 on Build GRCh38
rs777243430 Chr.15: 92473113-92473113 on Build GRCh38
rs370317996 Chr.15: 92472939-92472939 on Build GRCh38
rs577559139 Chr.15: 92472885-92472885 on Build GRCh38
rs373062788 Chr.15: 92472929-92472929 on Build GRCh38
rs901419629 Chr.15: 92473004-92473004 on Build GRCh38
rs931757480 Chr.15: 92473370-92473370 on Build GRCh38
rs932896061 Chr.15: 92473328-92473328 on Build GRCh38
rs148911837 Chr.15: 92472978-92472978 on Build GRCh38
rs774489539 Chr.15: 92473173-92473173 on Build GRCh38
rs975768020 Chr.15: 92473362-92473362 on Build GRCh38
rs779247677 Chr.15: 92472958-92472958 on Build GRCh38
rs763820044 Chr.15: 92472995-92472995 on Build GRCh38
rs778886284 Chr.15: 92473027-92473027 on Build GRCh38
rs999567476 Chr.15: 92472933-92472933 on Build GRCh38
rs374107103 Chr.15: 92472941-92472941 on Build GRCh38
rs372809883 Chr.15: 92473112-92473112 on Build GRCh38
rs768751377 Chr.15: 92472957-92472957 on Build GRCh38
rs762537067 Chr.15: 92472983-92472983 on Build GRCh38
rs376227819 Chr.15: 92472935-92472935 on Build GRCh38
rs757185422 Chr.15: 92473016-92473016 on Build GRCh38
rs13380006 Chr.15: 92472898-92472898 on Build GRCh38
rs761203204 Chr.15: 92472968-92472968 on Build GRCh38
rs35488423 Chr.15: 92473176-92473177 on Build GRCh38
rs748349823 Chr.15: 92473037-92473037 on Build GRCh38
rs989608750 Chr.15: 92473204-92473204 on Build GRCh38
rs374399765 Chr.15: 92472963-92472963 on Build GRCh38
rs922895917 Chr.15: 92473318-92473318 on Build GRCh38
rs748426365 Chr.15: 92472959-92472959 on Build GRCh38
rs945563068 Chr.15: 92473277-92473277 on Build GRCh38
rs1049721289 Chr.15: 92473379-92473379 on Build GRCh38
rs375678807 Chr.15: 92472949-92472949 on Build GRCh38
rs761834607 Chr.15: 92473220-92473220 on Build GRCh38
rs1024264093 Chr.15: 92473266-92473266 on Build GRCh38
rs1041198832 Chr.15: 92473285-92473285 on Build GRCh38
rs765935586 Chr.15: 92472909-92472909 on Build GRCh38
rs71412551 Chr.15: 92473168-92473168 on Build GRCh38
rs758704351 Chr.15: 92473089-92473089 on Build GRCh38
rs370284183 Chr.15: 92472960-92472960 on Build GRCh38
rs888564103 Chr.15: 92473111-92473111 on Build GRCh38
rs917604405 Chr.15: 92472893-92472893 on Build GRCh38
rs147273492 Chr.15: 92473202-92473202 on Build GRCh38

View Full Product Details