Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 180290-180802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBQ1
Entrez Gene ID
3049
Gene Name
hemoglobin subunit theta 1
Gene Aliases
HBQ
UniGene
Hs.247921
Gene Chromosome Location
Chr.16: 180334-181179 on Build GRCh38
Transcripts
NM_005331.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291736 c.86C>T p.A29V Chr.16: 180572-180572 on Build GRCh38
1209336 c.179G>A p.G60D Chr.16: 180749-180749 on Build GRCh38
5552449 c.193G>A p.D65N Chr.16: 180763-180763 on Build GRCh38
7370697 c.91G>A p.E31K Chr.16: 180577-180577 on Build GRCh38
5571807 c.211G>A p.V71M Chr.16: 180781-180781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199990776 Chr.16: 180539-180539 on Build GRCh38
rs775317264 Chr.16: 180748-180748 on Build GRCh38
rs780034510 Chr.16: 180592-180592 on Build GRCh38
rs955836532 Chr.16: 180398-180398 on Build GRCh38
rs1043992156 Chr.16: 180614-180614 on Build GRCh38
rs769113173 Chr.16: 180617-180617 on Build GRCh38
rs770545009 Chr.16: 180633-180633 on Build GRCh38
rs767682483 Chr.16: 180674-180674 on Build GRCh38
rs867998291 Chr.16: 180706-180706 on Build GRCh38
rs1018537253 Chr.16: 180637-180637 on Build GRCh38
rs779273965 Chr.16: 180618-180618 on Build GRCh38
rs755072950 Chr.16: 180312-180312 on Build GRCh38
rs1042798209 Chr.16: 180368-180368 on Build GRCh38
rs962037555 Chr.16: 180344-180344 on Build GRCh38
rs3901187 Chr.16: 180318-180318 on Build GRCh38
rs529280017 Chr.16: 180564-180564 on Build GRCh38
rs981274001 Chr.16: 180533-180533 on Build GRCh38
rs939536278 Chr.16: 180556-180556 on Build GRCh38
rs781009419 Chr.16: 180730-180731 on Build GRCh38
rs1029378744 Chr.16: 180462-180462 on Build GRCh38
rs973943467 Chr.16: 180483-180483 on Build GRCh38
rs776138470 Chr.16: 180738-180738 on Build GRCh38
rs372830198 Chr.16: 180522-180522 on Build GRCh38
rs533978818 Chr.16: 180746-180746 on Build GRCh38
rs12935435 Chr.16: 180778-180778 on Build GRCh38
rs763852370 Chr.16: 180774-180774 on Build GRCh38
rs951470419 Chr.16: 180719-180719 on Build GRCh38
rs555350790 Chr.16: 180380-180380 on Build GRCh38
rs920583664 Chr.16: 180355-180355 on Build GRCh38
rs1042860586 Chr.16: 180578-180578 on Build GRCh38
rs779800281 Chr.16: 180447-180447 on Build GRCh38
rs928157251 Chr.16: 180386-180386 on Build GRCh38
rs773029849 Chr.16: 180630-180630 on Build GRCh38
rs781404561 Chr.16: 180555-180555 on Build GRCh38
rs946988830 Chr.16: 180586-180586 on Build GRCh38
rs1011798393 Chr.16: 180366-180366 on Build GRCh38
rs1007598723 Chr.16: 180632-180632 on Build GRCh38
rs144518427 Chr.16: 180708-180708 on Build GRCh38
rs746648542 Chr.16: 180735-180735 on Build GRCh38
rs762758652 Chr.16: 180668-180668 on Build GRCh38
rs1015478640 Chr.16: 180468-180468 on Build GRCh38
rs754757619 Chr.16: 180630-180630 on Build GRCh38
rs890956490 Chr.16: 180465-180465 on Build GRCh38
rs765069266 Chr.16: 180521-180521 on Build GRCh38
rs562598307 Chr.16: 180480-180480 on Build GRCh38
rs181293989 Chr.16: 180640-180640 on Build GRCh38
rs141942919 Chr.16: 180456-180456 on Build GRCh38
rs915772727 Chr.16: 180523-180523 on Build GRCh38
rs12931536 Chr.16: 180769-180769 on Build GRCh38
rs760444390 Chr.16: 180631-180631 on Build GRCh38
rs550015255 Chr.16: 180512-180512 on Build GRCh38
rs899496393 Chr.16: 180451-180451 on Build GRCh38
rs573595881 Chr.16: 180421-180421 on Build GRCh38
rs902904571 Chr.16: 180381-180381 on Build GRCh38
rs771806135 Chr.16: 180628-180628 on Build GRCh38
rs934399582 Chr.16: 180591-180591 on Build GRCh38
rs769721062 Chr.16: 180747-180747 on Build GRCh38
rs772794273 Chr.16: 180495-180495 on Build GRCh38
rs1035529697 Chr.16: 180781-180781 on Build GRCh38
rs539517809 Chr.16: 180690-180690 on Build GRCh38
rs1029937828 Chr.16: 180397-180397 on Build GRCh38
rs369065332 Chr.16: 180736-180736 on Build GRCh38
rs749778892 Chr.16: 180611-180611 on Build GRCh38
rs1004026578 Chr.16: 180466-180466 on Build GRCh38
rs147305378 Chr.16: 180734-180734 on Build GRCh38
rs372406278 Chr.16: 180337-180337 on Build GRCh38
rs759443098 Chr.16: 180511-180511 on Build GRCh38
rs991862699 Chr.16: 180744-180744 on Build GRCh38
rs762914485 Chr.16: 180765-180765 on Build GRCh38
rs11646479 Chr.16: 180321-180321 on Build GRCh38
rs373067961 Chr.16: 180403-180403 on Build GRCh38
rs892701390 Chr.16: 180425-180425 on Build GRCh38
rs980774029 Chr.16: 180762-180762 on Build GRCh38
rs1053121010 Chr.16: 180424-180424 on Build GRCh38
rs1051306330 Chr.16: 180662-180662 on Build GRCh38
rs755458346 Chr.16: 180712-180712 on Build GRCh38
rs1005364 Chr.16: 180482-180482 on Build GRCh38
rs573111348 Chr.16: 180362-180362 on Build GRCh38
rs541775844 Chr.16: 180374-180374 on Build GRCh38
rs1045153798 Chr.16: 180378-180378 on Build GRCh38
rs753777815 Chr.16: 180509-180509 on Build GRCh38
rs776065252 Chr.16: 180634-180634 on Build GRCh38
rs11863726 Chr.16: 180579-180579 on Build GRCh38
rs79277966 Chr.16: 180644-180644 on Build GRCh38
rs772534431 Chr.16: 180732-180732 on Build GRCh38
rs564476176 Chr.16: 180485-180485 on Build GRCh38
rs746828470 Chr.16: 180585-180586 on Build GRCh38
rs1043432842 Chr.16: 180450-180450 on Build GRCh38
rs908427304 Chr.16: 180343-180343 on Build GRCh38
rs973346986 Chr.16: 180351-180351 on Build GRCh38
rs995418856 Chr.16: 180726-180726 on Build GRCh38
rs868520399 Chr.16: 180520-180520 on Build GRCh38
rs748479435 Chr.16: 180627-180627 on Build GRCh38
rs1010591930 Chr.16: 180428-180428 on Build GRCh38
rs563261796 Chr.16: 180666-180666 on Build GRCh38
rs148010777 Chr.16: 180722-180722 on Build GRCh38
rs964250858 Chr.16: 180434-180434 on Build GRCh38
rs969798123 Chr.16: 180743-180743 on Build GRCh38
rs76613236 Chr.16: 180725-180725 on Build GRCh38
rs974608016 Chr.16: 180438-180438 on Build GRCh38
rs948162836 Chr.16: 180375-180375 on Build GRCh38
rs1004356061 Chr.16: 180694-180694 on Build GRCh38
rs995714897 Chr.16: 180669-180669 on Build GRCh38
rs6600165 Chr.16: 180317-180317 on Build GRCh38
rs947276395 Chr.16: 180783-180783 on Build GRCh38
rs760288658 Chr.16: 180497-180497 on Build GRCh38
rs955225609 Chr.16: 180326-180326 on Build GRCh38
rs753870983 Chr.16: 180711-180711 on Build GRCh38
rs748649941 Chr.16: 180338-180338 on Build GRCh38
rs528791639 Chr.16: 180502-180502 on Build GRCh38
rs915862279 Chr.16: 180756-180756 on Build GRCh38
rs756374513 Chr.16: 180698-180698 on Build GRCh38

View Full Product Details