Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 401644-402156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6642134 c.53G>A p.R18H Chr.16: 402016-402016 on Build GRCh38
6642133 c.53G>A p.R18H Chr.16: 402016-402016 on Build GRCh38
6397108 c.46G>A p.A16T Chr.16: 402009-402009 on Build GRCh38
6397109 c.46G>A p.A16T Chr.16: 402009-402009 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891427039 Chr.16: 401838-401838 on Build GRCh38
rs756532333 Chr.16: 401942-401942 on Build GRCh38
rs1015909681 Chr.16: 401671-401671 on Build GRCh38
rs922185761 Chr.16: 401862-401862 on Build GRCh38
rs962153555 Chr.16: 402063-402063 on Build GRCh38
rs968707663 Chr.16: 401855-401855 on Build GRCh38
rs961916266 Chr.16: 402071-402071 on Build GRCh38
rs1001093788 Chr.16: 401832-401832 on Build GRCh38
rs371724576 Chr.16: 401715-401715 on Build GRCh38
rs759401789 Chr.16: 401992-401992 on Build GRCh38
rs530678255 Chr.16: 401835-401835 on Build GRCh38
rs749666907 Chr.16: 401948-401948 on Build GRCh38
rs755397136 Chr.16: 402030-402030 on Build GRCh38
rs1037375874 Chr.16: 401891-401891 on Build GRCh38
rs940473796 Chr.16: 401730-401730 on Build GRCh38
rs1012330625 Chr.16: 401760-401760 on Build GRCh38
rs78920701 Chr.16: 401995-401995 on Build GRCh38
rs576328517 Chr.16: 401691-401691 on Build GRCh38
rs201237938 Chr.16: 401985-401985 on Build GRCh38
rs940444373 Chr.16: 401889-401889 on Build GRCh38
rs767764934 Chr.16: 402014-402014 on Build GRCh38
rs555052741 Chr.16: 402064-402064 on Build GRCh38
rs780630794 Chr.16: 401946-401946 on Build GRCh38
rs904131693 Chr.16: 401802-401802 on Build GRCh38
rs1023888124 Chr.16: 401684-401684 on Build GRCh38
rs754562425 Chr.16: 402072-402072 on Build GRCh38
rs906454169 Chr.16: 401771-401771 on Build GRCh38
rs1023520344 Chr.16: 401761-401761 on Build GRCh38
rs892342917 Chr.16: 401929-401929 on Build GRCh38
rs898876983 Chr.16: 401750-401750 on Build GRCh38
rs917991597 Chr.16: 401777-401777 on Build GRCh38
rs965835724 Chr.16: 401744-401744 on Build GRCh38
rs947616946 Chr.16: 401713-401713 on Build GRCh38
rs867053311 Chr.16: 401991-401991 on Build GRCh38
rs973556036 Chr.16: 402067-402067 on Build GRCh38
rs988307263 Chr.16: 402123-402123 on Build GRCh38
rs956126316 Chr.16: 401704-401704 on Build GRCh38
rs559048058 Chr.16: 401763-401763 on Build GRCh38
rs1012200505 Chr.16: 401680-401680 on Build GRCh38
rs988534656 Chr.16: 401724-401724 on Build GRCh38
rs896636348 Chr.16: 401712-401713 on Build GRCh38
rs770809526 Chr.16: 402076-402076 on Build GRCh38
rs908138914 Chr.16: 402055-402055 on Build GRCh38
rs543353893 Chr.16: 401694-401694 on Build GRCh38
rs531312457 Chr.16: 401914-401914 on Build GRCh38
rs985225766 Chr.16: 401766-401766 on Build GRCh38
rs529722628 Chr.16: 401773-401773 on Build GRCh38
rs988793336 Chr.16: 401707-401707 on Build GRCh38
rs907852835 Chr.16: 401767-401772 on Build GRCh38
rs868438209 Chr.16: 402031-402031 on Build GRCh38
rs1052259681 Chr.16: 401928-401928 on Build GRCh38
rs987586349 Chr.16: 401880-401880 on Build GRCh38
rs768136109 Chr.16: 401954-401954 on Build GRCh38
rs886172066 Chr.16: 401728-401728 on Build GRCh38
rs576165458 Chr.16: 402122-402122 on Build GRCh38
rs566213843 Chr.16: 401986-401986 on Build GRCh38
rs753523219 Chr.16: 402056-402056 on Build GRCh38
rs939246646 Chr.16: 402129-402129 on Build GRCh38
rs758485249 Chr.16: 401900-401900 on Build GRCh38
rs956092110 Chr.16: 401834-401834 on Build GRCh38
rs550218811 Chr.16: 401865-401865 on Build GRCh38
rs771608292 Chr.16: 401969-401969 on Build GRCh38
rs925448205 Chr.16: 401959-401959 on Build GRCh38
rs1030860846 Chr.16: 401794-401794 on Build GRCh38
rs1045216495 Chr.16: 401792-401792 on Build GRCh38
rs1032663387 Chr.16: 401722-401722 on Build GRCh38
rs774070747 Chr.16: 401960-401960 on Build GRCh38
rs757248323 Chr.16: 402125-402126 on Build GRCh38
rs948258531 Chr.16: 401781-401781 on Build GRCh38
rs965785287 Chr.16: 401883-401883 on Build GRCh38
rs780623637 Chr.16: 402023-402023 on Build GRCh38
rs988927853 Chr.16: 401835-401835 on Build GRCh38
rs750959238 Chr.16: 402021-402021 on Build GRCh38
rs778395567 Chr.16: 402035-402035 on Build GRCh38
rs954976581 Chr.16: 402005-402005 on Build GRCh38
rs561145295 Chr.16: 401800-401800 on Build GRCh38
rs951875394 Chr.16: 402051-402051 on Build GRCh38
rs531889480 Chr.16: 401825-401825 on Build GRCh38
rs940766236 Chr.16: 401710-401710 on Build GRCh38
rs781756171 Chr.16: 401933-401933 on Build GRCh38
rs1006103000 Chr.16: 402057-402057 on Build GRCh38
rs3743889 Chr.16: 401719-401719 on Build GRCh38
rs764183443 Chr.16: 401993-401993 on Build GRCh38
rs554790426 Chr.16: 401688-401688 on Build GRCh38
rs892716149 Chr.16: 401988-401988 on Build GRCh38
rs907885452 Chr.16: 401899-401899 on Build GRCh38
rs962052332 Chr.16: 401902-401902 on Build GRCh38
rs933477364 Chr.16: 401720-401720 on Build GRCh38
rs1012507982 Chr.16: 401713-401714 on Build GRCh38
rs576777512 Chr.16: 401716-401716 on Build GRCh38
rs920332374 Chr.16: 402082-402082 on Build GRCh38
rs947817542 Chr.16: 402086-402086 on Build GRCh38
rs747567968 Chr.16: 401963-401963 on Build GRCh38
rs906429270 Chr.16: 401917-401917 on Build GRCh38
rs749566138 Chr.16: 402026-402026 on Build GRCh38
rs1024340857 Chr.16: 401714-401714 on Build GRCh38
rs901620441 Chr.16: 402007-402007 on Build GRCh38
rs933591369 Chr.16: 401875-401875 on Build GRCh38
rs914601949 Chr.16: 401709-401709 on Build GRCh38
rs975027581 Chr.16: 401676-401676 on Build GRCh38
rs1045651028 Chr.16: 401955-401955 on Build GRCh38
rs1037323864 Chr.16: 401747-401747 on Build GRCh38
rs936889523 Chr.16: 402107-402107 on Build GRCh38
rs952069797 Chr.16: 401890-401890 on Build GRCh38
rs1021632267 Chr.16: 401845-401845 on Build GRCh38
rs899076972 Chr.16: 401673-401673 on Build GRCh38
rs975155755 Chr.16: 401857-401857 on Build GRCh38
rs746428082 Chr.16: 401937-401937 on Build GRCh38
rs547791782 Chr.16: 401916-401916 on Build GRCh38
rs549203843 Chr.16: 401795-401795 on Build GRCh38
rs1003896667 Chr.16: 401918-401918 on Build GRCh38
rs1004484299 Chr.16: 401670-401670 on Build GRCh38
rs970549131 Chr.16: 401687-401687 on Build GRCh38
rs757391887 Chr.16: 402010-402010 on Build GRCh38
rs927651313 Chr.16: 402108-402108 on Build GRCh38
rs980839104 Chr.16: 402092-402092 on Build GRCh38
rs920349403 Chr.16: 401901-401901 on Build GRCh38
rs1051901599 Chr.16: 401723-401723 on Build GRCh38
rs747844823 Chr.16: 402042-402042 on Build GRCh38
rs757445116 Chr.16: 401923-401923 on Build GRCh38
rs1044720580 Chr.16: 402088-402088 on Build GRCh38
rs925449325 Chr.16: 402099-402099 on Build GRCh38
rs571878462 Chr.16: 401908-401908 on Build GRCh38
rs773108950 Chr.16: 401977-401977 on Build GRCh38
rs760675937 Chr.16: 401984-401984 on Build GRCh38
rs908084745 Chr.16: 401886-401886 on Build GRCh38
rs771555238 Chr.16: 402043-402043 on Build GRCh38
rs987724864 Chr.16: 402050-402050 on Build GRCh38

View Full Product Details