Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 410775-411272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1377887 c.565A>G p.T189A Chr.16: 410980-410980 on Build GRCh38
5148639 c.583G>A p.E195K Chr.16: 410998-410998 on Build GRCh38
3509651 c.593C>T p.P198L Chr.16: 411008-411008 on Build GRCh38
6204522 c.604C>T p.R202C Chr.16: 411019-411019 on Build GRCh38
5521224 c.611A>G p.N204S Chr.16: 411026-411026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756471986 Chr.16: 410974-410974 on Build GRCh38
rs891245056 Chr.16: 410872-410872 on Build GRCh38
rs776712367 Chr.16: 410801-410801 on Build GRCh38
rs749457989 Chr.16: 411019-411019 on Build GRCh38
rs138357981 Chr.16: 410982-410982 on Build GRCh38
rs952232616 Chr.16: 411037-411037 on Build GRCh38
rs779596992 Chr.16: 411228-411228 on Build GRCh38
rs374085698 Chr.16: 411021-411021 on Build GRCh38
rs764891390 Chr.16: 411053-411053 on Build GRCh38
rs891679548 Chr.16: 411085-411085 on Build GRCh38
rs1023624972 Chr.16: 411181-411181 on Build GRCh38
rs1004209444 Chr.16: 410879-410879 on Build GRCh38
rs774308860 Chr.16: 410799-410799 on Build GRCh38
rs779039524 Chr.16: 411092-411092 on Build GRCh38
rs576730413 Chr.16: 411191-411191 on Build GRCh38
rs769015947 Chr.16: 411020-411020 on Build GRCh38
rs768964385 Chr.16: 411091-411091 on Build GRCh38
rs377441280 Chr.16: 411001-411001 on Build GRCh38
rs768033482 Chr.16: 411034-411034 on Build GRCh38
rs772097736 Chr.16: 411095-411095 on Build GRCh38
rs77751507 Chr.16: 411225-411225 on Build GRCh38
rs1003448419 Chr.16: 411202-411202 on Build GRCh38
rs774579702 Chr.16: 410961-410961 on Build GRCh38
rs61645593 Chr.16: 410830-410830 on Build GRCh38
rs140277658 Chr.16: 411049-411049 on Build GRCh38
rs142463074 Chr.16: 410837-410837 on Build GRCh38
rs777755083 Chr.16: 411067-411067 on Build GRCh38
rs1018878754 Chr.16: 411145-411145 on Build GRCh38
rs767935940 Chr.16: 410966-410966 on Build GRCh38
rs765663302 Chr.16: 410951-410952 on Build GRCh38
rs576664155 Chr.16: 410819-410819 on Build GRCh38
rs765990942 Chr.16: 410975-410975 on Build GRCh38
rs754528043 Chr.16: 410836-410836 on Build GRCh38
rs113423539 Chr.16: 410939-410939 on Build GRCh38
rs901949811 Chr.16: 411162-411162 on Build GRCh38
rs375699056 Chr.16: 411068-411068 on Build GRCh38
rs754566536 Chr.16: 411051-411051 on Build GRCh38
rs746811727 Chr.16: 410933-410933 on Build GRCh38
rs202160157 Chr.16: 410926-410926 on Build GRCh38
rs962247822 Chr.16: 411241-411241 on Build GRCh38
rs745683116 Chr.16: 411082-411082 on Build GRCh38
rs35431910 Chr.16: 410802-410803 on Build GRCh38
rs752172006 Chr.16: 410815-410815 on Build GRCh38
rs752220281 Chr.16: 411060-411060 on Build GRCh38
rs559322911 Chr.16: 411238-411238 on Build GRCh38
rs909576111 Chr.16: 410878-410878 on Build GRCh38
rs1054336300 Chr.16: 411080-411080 on Build GRCh38
rs374737970 Chr.16: 410925-410925 on Build GRCh38
rs749944823 Chr.16: 410957-410960 on Build GRCh38
rs748761351 Chr.16: 410833-410833 on Build GRCh38
rs750770772 Chr.16: 410823-410823 on Build GRCh38
rs759736356 Chr.16: 410802-410802 on Build GRCh38
rs930867676 Chr.16: 410866-410866 on Build GRCh38
rs762327591 Chr.16: 410963-410963 on Build GRCh38
rs1037130609 Chr.16: 411099-411099 on Build GRCh38
rs372631767 Chr.16: 410955-410955 on Build GRCh38
rs563150166 Chr.16: 410962-410962 on Build GRCh38
rs757895871 Chr.16: 411061-411061 on Build GRCh38
rs541871714 Chr.16: 410957-410957 on Build GRCh38
rs1030144882 Chr.16: 410871-410871 on Build GRCh38
rs764398582 Chr.16: 411022-411022 on Build GRCh38
rs773637698 Chr.16: 411038-411038 on Build GRCh38
rs755705464 Chr.16: 411154-411154 on Build GRCh38
rs775853274 Chr.16: 411017-411017 on Build GRCh38
rs532710219 Chr.16: 411175-411175 on Build GRCh38
rs761085304 Chr.16: 411044-411044 on Build GRCh38
rs997380022 Chr.16: 410855-410855 on Build GRCh38
rs115317717 Chr.16: 410796-410796 on Build GRCh38
rs145686298 Chr.16: 410981-410981 on Build GRCh38
rs758307119 Chr.16: 410983-410983 on Build GRCh38
rs755289125 Chr.16: 410920-410920 on Build GRCh38
rs139241707 Chr.16: 410960-410960 on Build GRCh38
rs570280733 Chr.16: 411071-411071 on Build GRCh38
rs751943184 Chr.16: 410793-410793 on Build GRCh38
rs375233826 Chr.16: 410798-410798 on Build GRCh38
rs771958925 Chr.16: 410800-410800 on Build GRCh38
rs765660955 Chr.16: 410914-410914 on Build GRCh38
rs764338169 Chr.16: 410949-410949 on Build GRCh38
rs761101396 Chr.16: 411107-411107 on Build GRCh38
rs947069412 Chr.16: 411003-411003 on Build GRCh38
rs746798406 Chr.16: 410995-410995 on Build GRCh38
rs200015511 Chr.16: 410794-410794 on Build GRCh38
rs181461221 Chr.16: 410797-410797 on Build GRCh38
rs150991342 Chr.16: 411045-411045 on Build GRCh38
rs984966854 Chr.16: 411237-411237 on Build GRCh38
rs748233278 Chr.16: 411093-411093 on Build GRCh38
rs555199808 Chr.16: 410818-410818 on Build GRCh38
rs779707992 Chr.16: 410832-410832 on Build GRCh38
rs972356889 Chr.16: 411047-411047 on Build GRCh38
rs60098912 Chr.16: 410795-410795 on Build GRCh38
rs750756002 Chr.16: 410973-410973 on Build GRCh38
rs757267700 Chr.16: 411070-411070 on Build GRCh38
rs773002038 Chr.16: 410932-410932 on Build GRCh38
rs149169373 Chr.16: 410984-410984 on Build GRCh38
rs754356484 Chr.16: 410793-410794 on Build GRCh38
rs752785156 Chr.16: 410803-410803 on Build GRCh38
rs530131762 Chr.16: 410946-410946 on Build GRCh38
rs770027504 Chr.16: 410937-410937 on Build GRCh38
rs996692261 Chr.16: 411167-411167 on Build GRCh38
rs764677277 Chr.16: 410813-410813 on Build GRCh38
rs559836051 Chr.16: 410940-410940 on Build GRCh38
rs140561403 Chr.16: 410976-410976 on Build GRCh38
rs546270474 Chr.16: 411030-411030 on Build GRCh38
rs757888596 Chr.16: 411039-411039 on Build GRCh38
rs757808522 Chr.16: 410794-410794 on Build GRCh38
rs773762761 Chr.16: 411101-411101 on Build GRCh38

View Full Product Details