Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 635881-636384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL26
Entrez Gene ID
84326
Gene Name
methyltransferase like 26
Gene Aliases
C16orf13, JFP2
UniGene
Hs.239500
Gene Chromosome Location
Chr.16: 634427-636366 on Build GRCh38
Transcripts
NM_001040160.2 NM_001040161.2 NM_001040162.2 NM_001040165.2 NM_001288710.1 NM_032366.4 NR_109976.1 NR_109977.1 NR_109978.1 NR_109979.1 XM_011522713.2 XM_011522714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5385250 c.71C>T p.P24L Chr.16: 636220-636220 on Build GRCh38
5385249 c.71C>T p.P24L Chr.16: 636220-636220 on Build GRCh38
6237458 c.146T>C p.F49S Chr.16: 636145-636145 on Build GRCh38
1324376 c.123C>A p.H41Q Chr.16: 636168-636168 on Build GRCh38
5767591 c.27G>C p.R9R Chr.16: 636264-636264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879008513 Chr.16: 636197-636197 on Build GRCh38
rs747961419 Chr.16: 635981-635981 on Build GRCh38
rs931505312 Chr.16: 635970-635970 on Build GRCh38
rs868001172 Chr.16: 636213-636213 on Build GRCh38
rs771401205 Chr.16: 636299-636301 on Build GRCh38
rs1045015959 Chr.16: 635975-635975 on Build GRCh38
rs1008726674 Chr.16: 636360-636360 on Build GRCh38
rs931451604 Chr.16: 636231-636231 on Build GRCh38
rs779590699 Chr.16: 636290-636290 on Build GRCh38
rs891763530 Chr.16: 636319-636319 on Build GRCh38
rs567780790 Chr.16: 636085-636085 on Build GRCh38
rs953718783 Chr.16: 636257-636257 on Build GRCh38
rs1057144214 Chr.16: 636018-636018 on Build GRCh38
rs769943081 Chr.16: 636052-636052 on Build GRCh38
rs537741456 Chr.16: 636353-636353 on Build GRCh38
rs576875437 Chr.16: 636324-636324 on Build GRCh38
rs1020735826 Chr.16: 635941-635941 on Build GRCh38
rs891836854 Chr.16: 636034-636034 on Build GRCh38
rs950150296 Chr.16: 636313-636313 on Build GRCh38
rs776984903 Chr.16: 636334-636334 on Build GRCh38
rs968927431 Chr.16: 636087-636087 on Build GRCh38
rs776844614 Chr.16: 635988-635988 on Build GRCh38
rs771071496 Chr.16: 635985-635985 on Build GRCh38
rs781290586 Chr.16: 636339-636339 on Build GRCh38
rs182493474 Chr.16: 636168-636168 on Build GRCh38
rs369108594 Chr.16: 636080-636080 on Build GRCh38
rs950182216 Chr.16: 636050-636050 on Build GRCh38
rs554688282 Chr.16: 635967-635967 on Build GRCh38
rs953917642 Chr.16: 636001-636001 on Build GRCh38
rs1054391574 Chr.16: 636321-636321 on Build GRCh38
rs898617980 Chr.16: 635957-635957 on Build GRCh38
rs907543266 Chr.16: 636170-636170 on Build GRCh38
rs750054770 Chr.16: 636252-636252 on Build GRCh38
rs759565536 Chr.16: 636016-636016 on Build GRCh38
rs568769487 Chr.16: 636345-636345 on Build GRCh38
rs906134462 Chr.16: 635976-635976 on Build GRCh38
rs1037111326 Chr.16: 636032-636032 on Build GRCh38
rs996042055 Chr.16: 636121-636121 on Build GRCh38
rs889448267 Chr.16: 636336-636336 on Build GRCh38
rs1009267575 Chr.16: 635934-635934 on Build GRCh38
rs371293187 Chr.16: 636239-636239 on Build GRCh38
rs973525679 Chr.16: 636191-636191 on Build GRCh38
rs374653903 Chr.16: 636177-636177 on Build GRCh38
rs973088901 Chr.16: 636347-636347 on Build GRCh38
rs956230297 Chr.16: 635900-635900 on Build GRCh38
rs984037330 Chr.16: 636276-636276 on Build GRCh38
rs868339921 Chr.16: 636235-636235 on Build GRCh38
rs961293862 Chr.16: 636289-636289 on Build GRCh38
rs753314950 Chr.16: 636291-636291 on Build GRCh38
rs1000384641 Chr.16: 636112-636112 on Build GRCh38
rs775687646 Chr.16: 636135-636135 on Build GRCh38
rs936035499 Chr.16: 636320-636320 on Build GRCh38
rs904587649 Chr.16: 636332-636332 on Build GRCh38
rs754976478 Chr.16: 636308-636308 on Build GRCh38
rs759065242 Chr.16: 636337-636337 on Build GRCh38
rs139372925 Chr.16: 636322-636323 on Build GRCh38
rs75174484 Chr.16: 636230-636230 on Build GRCh38
rs909700351 Chr.16: 636006-636006 on Build GRCh38
rs917088640 Chr.16: 636296-636296 on Build GRCh38
rs755644493 Chr.16: 636277-636277 on Build GRCh38
rs1057418110 Chr.16: 636303-636303 on Build GRCh38
rs1019530844 Chr.16: 636228-636228 on Build GRCh38
rs368149522 Chr.16: 636236-636236 on Build GRCh38
rs920372982 Chr.16: 636212-636212 on Build GRCh38
rs879522650 Chr.16: 636029-636029 on Build GRCh38
rs773562510 Chr.16: 636055-636055 on Build GRCh38
rs763591209 Chr.16: 636148-636148 on Build GRCh38
rs954614856 Chr.16: 636151-636151 on Build GRCh38
rs954581272 Chr.16: 636346-636346 on Build GRCh38
rs984068430 Chr.16: 636002-636002 on Build GRCh38
rs749774299 Chr.16: 636177-636196 on Build GRCh38
rs756872169 Chr.16: 636074-636074 on Build GRCh38
rs771388874 Chr.16: 636327-636327 on Build GRCh38
rs117667029 Chr.16: 636337-636337 on Build GRCh38
rs11557718 Chr.16: 636123-636123 on Build GRCh38
rs1021716279 Chr.16: 636063-636063 on Build GRCh38

View Full Product Details