Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2702273-2702767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD5
Entrez Gene ID
54442
Gene Name
potassium channel tetramerization domain containing 5
Gene Aliases
-
UniGene
Hs.61960
Gene Chromosome Location
Chr.16: 2682494-2709030 on Build GRCh38
Transcripts
NM_018992.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
969270 c.611T>G p.V204G Chr.16: 2702414-2702414 on Build GRCh38
3108949 c.650C>T p.A217V Chr.16: 2702453-2702453 on Build GRCh38
3387346 c.642C>T p.Y214Y Chr.16: 2702445-2702445 on Build GRCh38
145538 c.605T>C p.L202P Chr.16: 2702408-2702408 on Build GRCh38
6143946 c.627G>T p.L209L Chr.16: 2702430-2702430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201376163 Chr.16: 2702510-2702510 on Build GRCh38
rs200979195 Chr.16: 2702307-2702307 on Build GRCh38
rs541485896 Chr.16: 2702682-2702682 on Build GRCh38
rs972993545 Chr.16: 2702530-2702530 on Build GRCh38
rs778282084 Chr.16: 2702452-2702452 on Build GRCh38
rs546984670 Chr.16: 2702577-2702577 on Build GRCh38
rs895179349 Chr.16: 2702592-2702592 on Build GRCh38
rs974364196 Chr.16: 2702710-2702710 on Build GRCh38
rs755043258 Chr.16: 2702445-2702445 on Build GRCh38
rs1035037875 Chr.16: 2702653-2702653 on Build GRCh38
rs763544870 Chr.16: 2702369-2702369 on Build GRCh38
rs773729295 Chr.16: 2702367-2702367 on Build GRCh38
rs1012195282 Chr.16: 2702606-2702606 on Build GRCh38
rs766882747 Chr.16: 2702332-2702332 on Build GRCh38
rs895100408 Chr.16: 2702662-2702662 on Build GRCh38
rs1017002360 Chr.16: 2702698-2702698 on Build GRCh38
rs200209977 Chr.16: 2702347-2702347 on Build GRCh38
rs13337854 Chr.16: 2702687-2702687 on Build GRCh38
rs118149302 Chr.16: 2702364-2702364 on Build GRCh38
rs776176746 Chr.16: 2702490-2702490 on Build GRCh38
rs1051317963 Chr.16: 2702329-2702329 on Build GRCh38
rs760326025 Chr.16: 2702442-2702443 on Build GRCh38
rs771487863 Chr.16: 2702651-2702651 on Build GRCh38
rs530216796 Chr.16: 2702451-2702451 on Build GRCh38
rs761450252 Chr.16: 2702491-2702491 on Build GRCh38
rs201349674 Chr.16: 2702353-2702353 on Build GRCh38
rs747148611 Chr.16: 2702356-2702356 on Build GRCh38
rs781413339 Chr.16: 2702506-2702506 on Build GRCh38
rs770220021 Chr.16: 2702365-2702365 on Build GRCh38
rs753609368 Chr.16: 2702338-2702338 on Build GRCh38
rs781429770 Chr.16: 2702446-2702446 on Build GRCh38
rs536494152 Chr.16: 2702328-2702328 on Build GRCh38
rs761466593 Chr.16: 2702437-2702437 on Build GRCh38
rs571478279 Chr.16: 2702529-2702529 on Build GRCh38
rs75461256 Chr.16: 2702624-2702624 on Build GRCh38
rs914200965 Chr.16: 2702512-2702512 on Build GRCh38
rs745830428 Chr.16: 2702527-2702527 on Build GRCh38
rs972877463 Chr.16: 2702731-2702731 on Build GRCh38
rs200900245 Chr.16: 2702309-2702309 on Build GRCh38
rs1025002144 Chr.16: 2702475-2702475 on Build GRCh38
rs539173875 Chr.16: 2702626-2702626 on Build GRCh38
rs541399652 Chr.16: 2702439-2702439 on Build GRCh38
rs986480902 Chr.16: 2702550-2702550 on Build GRCh38
rs768572338 Chr.16: 2702499-2702499 on Build GRCh38
rs910995680 Chr.16: 2702584-2702584 on Build GRCh38
rs551397880 Chr.16: 2702337-2702337 on Build GRCh38
rs527328247 Chr.16: 2702537-2702537 on Build GRCh38
rs1047678297 Chr.16: 2702403-2702403 on Build GRCh38
rs755289632 Chr.16: 2702412-2702412 on Build GRCh38
rs1054287356 Chr.16: 2702370-2702370 on Build GRCh38
rs559172611 Chr.16: 2702400-2702400 on Build GRCh38
rs202098208 Chr.16: 2702388-2702388 on Build GRCh38
rs941860382 Chr.16: 2702736-2702736 on Build GRCh38
rs764905898 Chr.16: 2702492-2702492 on Build GRCh38
rs1025940937 Chr.16: 2702745-2702745 on Build GRCh38
rs752844864 Chr.16: 2702466-2702466 on Build GRCh38
rs771536000 Chr.16: 2702375-2702375 on Build GRCh38
rs756997736 Chr.16: 2702339-2702339 on Build GRCh38
rs183960699 Chr.16: 2702644-2702644 on Build GRCh38
rs148272021 Chr.16: 2702415-2702415 on Build GRCh38
rs575022146 Chr.16: 2702706-2702706 on Build GRCh38
rs149564820 Chr.16: 2702457-2702457 on Build GRCh38
rs1035555839 Chr.16: 2702488-2702488 on Build GRCh38
rs1018517636 Chr.16: 2702683-2702683 on Build GRCh38
rs750284506 Chr.16: 2702441-2702441 on Build GRCh38
rs962899932 Chr.16: 2702711-2702711 on Build GRCh38
rs918829284 Chr.16: 2702532-2702532 on Build GRCh38
rs375530628 Chr.16: 2702458-2702458 on Build GRCh38
rs754887930 Chr.16: 2702504-2702504 on Build GRCh38
rs752313927 Chr.16: 2702333-2702333 on Build GRCh38
rs749710779 Chr.16: 2702456-2702456 on Build GRCh38
rs983322202 Chr.16: 2702724-2702724 on Build GRCh38
rs367644972 Chr.16: 2702310-2702310 on Build GRCh38
rs748324959 Chr.16: 2702450-2702450 on Build GRCh38
rs561409300 Chr.16: 2702442-2702442 on Build GRCh38
rs1010110275 Chr.16: 2702418-2702418 on Build GRCh38
rs79679889 Chr.16: 2702627-2702627 on Build GRCh38
rs181320131 Chr.16: 2702619-2702619 on Build GRCh38
rs759993139 Chr.16: 2702460-2702460 on Build GRCh38
rs954414931 Chr.16: 2702723-2702723 on Build GRCh38
rs372221749 Chr.16: 2702360-2702360 on Build GRCh38
rs907129164 Chr.16: 2702636-2702636 on Build GRCh38
rs1037604996 Chr.16: 2702588-2702588 on Build GRCh38
rs377197615 Chr.16: 2702496-2702496 on Build GRCh38
rs189273223 Chr.16: 2702499-2702499 on Build GRCh38
rs567049573 Chr.16: 2702578-2702578 on Build GRCh38
rs376746520 Chr.16: 2702341-2702341 on Build GRCh38
rs745747037 Chr.16: 2702345-2702345 on Build GRCh38
rs755719010 Chr.16: 2702336-2702336 on Build GRCh38
rs933129928 Chr.16: 2702378-2702378 on Build GRCh38
rs889524107 Chr.16: 2702340-2702340 on Build GRCh38
rs373383340 Chr.16: 2702471-2702471 on Build GRCh38
rs535924697 Chr.16: 2702585-2702585 on Build GRCh38
rs140133666 Chr.16: 2702493-2702493 on Build GRCh38
rs768735412 Chr.16: 2702358-2702358 on Build GRCh38
rs371740693 Chr.16: 2702323-2702323 on Build GRCh38

View Full Product Details