Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3611449-3611936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1
Entrez Gene ID
1773
Gene Name
deoxyribonuclease I
Gene Aliases
DNL1, DRNI
UniGene
Hs.629638
Gene Chromosome Location
Chr.16: 3611705-3665472 on Build GRCh38
Transcripts
XM_005255148.3 XM_006720854.3 XM_011522393.2 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022997.1 XM_017023001.1 XM_017023004.1 XM_017023005.1 XM_017023007.1 NM_032444.2 XM_011522715.2

Gene Symbol
SLX4
Entrez Gene ID
84464
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
BTBD12, FANCP, MUS312
UniGene
Hs.143681
Gene Chromosome Location
Chr.16: 3581181-3611598 on Build GRCh38
Transcripts
XM_005255148.3 XM_006720854.3 XM_011522393.2 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022997.1 XM_017023001.1 XM_017023004.1 XM_017023005.1 XM_017023007.1 NM_032444.2 XM_011522715.2

SNP Detail

SNP ID Chromosome Location
rs927139671 Chr.16: 3611666-3611666 on Build GRCh38
rs890429359 Chr.16: 3611783-3611783 on Build GRCh38
rs938875275 Chr.16: 3611575-3611575 on Build GRCh38
rs1036044789 Chr.16: 3611857-3611857 on Build GRCh38
rs1007023014 Chr.16: 3611736-3611736 on Build GRCh38
rs988841018 Chr.16: 3611756-3611756 on Build GRCh38
rs557926697 Chr.16: 3611530-3611530 on Build GRCh38
rs886051987 Chr.16: 3611552-3611552 on Build GRCh38
rs369969679 Chr.16: 3611577-3611577 on Build GRCh38
rs1010344544 Chr.16: 3611894-3611894 on Build GRCh38
rs913206577 Chr.16: 3611773-3611773 on Build GRCh38
rs1035972712 Chr.16: 3611704-3611704 on Build GRCh38
rs893871999 Chr.16: 3611583-3611583 on Build GRCh38
rs1048805876 Chr.16: 3611693-3611693 on Build GRCh38
rs937164155 Chr.16: 3611677-3611677 on Build GRCh38
rs1028202540 Chr.16: 3611505-3611505 on Build GRCh38
rs761710170 Chr.16: 3611876-3611876 on Build GRCh38
rs553756754 Chr.16: 3611633-3611633 on Build GRCh38
rs374977442 Chr.16: 3611766-3611767 on Build GRCh38
rs1011629316 Chr.16: 3611584-3611584 on Build GRCh38
rs537941606 Chr.16: 3611643-3611643 on Build GRCh38
rs955424857 Chr.16: 3611509-3611509 on Build GRCh38
rs1004472564 Chr.16: 3611552-3611552 on Build GRCh38
rs907442286 Chr.16: 3611568-3611568 on Build GRCh38
rs773592349 Chr.16: 3611644-3611644 on Build GRCh38
rs1015488187 Chr.16: 3611795-3611795 on Build GRCh38
rs575332149 Chr.16: 3611807-3611807 on Build GRCh38
rs928902753 Chr.16: 3611717-3611717 on Build GRCh38
rs748243241 Chr.16: 3611572-3611572 on Build GRCh38
rs953898116 Chr.16: 3611904-3611904 on Build GRCh38
rs996609926 Chr.16: 3611772-3611772 on Build GRCh38
rs947517715 Chr.16: 3611739-3611739 on Build GRCh38
rs1007096896 Chr.16: 3611882-3611882 on Build GRCh38
rs988809765 Chr.16: 3611609-3611609 on Build GRCh38
rs1033132962 Chr.16: 3611738-3611738 on Build GRCh38
rs529622799 Chr.16: 3611885-3611885 on Build GRCh38
rs985555410 Chr.16: 3611589-3611589 on Build GRCh38
rs998859630 Chr.16: 3611721-3611721 on Build GRCh38
rs1004536912 Chr.16: 3611699-3611699 on Build GRCh38
rs1027339181 Chr.16: 3611543-3611543 on Build GRCh38
rs564688217 Chr.16: 3611838-3611838 on Build GRCh38
rs776911313 Chr.16: 3611836-3611836 on Build GRCh38
rs549536386 Chr.16: 3611887-3611887 on Build GRCh38
rs957360124 Chr.16: 3611605-3611605 on Build GRCh38
rs373397270 Chr.16: 3611870-3611870 on Build GRCh38
rs142297926 Chr.16: 3611826-3611828 on Build GRCh38
rs949995712 Chr.16: 3611637-3611637 on Build GRCh38
rs897426401 Chr.16: 3611708-3611708 on Build GRCh38
rs891677450 Chr.16: 3611478-3611478 on Build GRCh38
rs1008502700 Chr.16: 3611669-3611669 on Build GRCh38
rs928331854 Chr.16: 3611864-3611864 on Build GRCh38
rs948335793 Chr.16: 3611547-3611547 on Build GRCh38
rs191853460 Chr.16: 3611524-3611524 on Build GRCh38
rs1029885705 Chr.16: 3611722-3611722 on Build GRCh38
rs955305182 Chr.16: 3611654-3611654 on Build GRCh38
rs985334475 Chr.16: 3611909-3611909 on Build GRCh38
rs1053270657 Chr.16: 3611580-3611582 on Build GRCh38
rs891939035 Chr.16: 3611581-3611581 on Build GRCh38
rs986644659 Chr.16: 3611516-3611516 on Build GRCh38
rs940424775 Chr.16: 3611855-3611855 on Build GRCh38
rs115932198 Chr.16: 3611782-3611782 on Build GRCh38
rs954362475 Chr.16: 3611727-3611727 on Build GRCh38
rs969813497 Chr.16: 3611815-3611815 on Build GRCh38
rs577799018 Chr.16: 3611539-3611539 on Build GRCh38
rs1053030396 Chr.16: 3611468-3611468 on Build GRCh38
rs886051988 Chr.16: 3611573-3611573 on Build GRCh38
rs534077056 Chr.16: 3611597-3611597 on Build GRCh38
rs1053751137 Chr.16: 3611723-3611723 on Build GRCh38
rs1015455381 Chr.16: 3611674-3611674 on Build GRCh38
rs938962644 Chr.16: 3611718-3611718 on Build GRCh38
rs998392354 Chr.16: 3611570-3611570 on Build GRCh38
rs12924990 Chr.16: 3611814-3611814 on Build GRCh38
rs533388350 Chr.16: 3611858-3611858 on Build GRCh38
rs541783569 Chr.16: 3611719-3611719 on Build GRCh38
rs768417819 Chr.16: 3611695-3611695 on Build GRCh38
rs143214216 Chr.16: 3611901-3611902 on Build GRCh38
rs995338046 Chr.16: 3611810-3611810 on Build GRCh38
rs949954511 Chr.16: 3611520-3611520 on Build GRCh38
rs989024772 Chr.16: 3611867-3611867 on Build GRCh38
rs564851536 Chr.16: 3611903-3611903 on Build GRCh38
rs925993171 Chr.16: 3611602-3611602 on Build GRCh38
rs112135563 Chr.16: 3611794-3611794 on Build GRCh38
rs981429221 Chr.16: 3611907-3611907 on Build GRCh38
rs373745911 Chr.16: 3611895-3611895 on Build GRCh38
rs887460446 Chr.16: 3611549-3611549 on Build GRCh38
rs961919899 Chr.16: 3611569-3611569 on Build GRCh38
rs1043161216 Chr.16: 3611767-3611767 on Build GRCh38
rs901517344 Chr.16: 3611861-3611861 on Build GRCh38
rs138415805 Chr.16: 3611864-3611864 on Build GRCh38
rs954212843 Chr.16: 3611580-3611580 on Build GRCh38
rs866063952 Chr.16: 3611744-3611744 on Build GRCh38
rs1048881237 Chr.16: 3611809-3611809 on Build GRCh38
rs1043088987 Chr.16: 3611625-3611625 on Build GRCh38
rs1001674298 Chr.16: 3611532-3611532 on Build GRCh38
rs544061325 Chr.16: 3611749-3611750 on Build GRCh38
rs779035961 Chr.16: 3611556-3611556 on Build GRCh38
rs747339458 Chr.16: 3611785-3611785 on Build GRCh38
rs900587510 Chr.16: 3611770-3611770 on Build GRCh38
rs905641064 Chr.16: 3611528-3611528 on Build GRCh38

View Full Product Details