Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4508022-4508516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMOX2
Entrez Gene ID
3163
Gene Name
heme oxygenase 2
Gene Aliases
HO-2
UniGene
Hs.284279
Gene Chromosome Location
Chr.16: 4474697-4510347 on Build GRCh38
Transcripts
NM_001127204.1 NM_001127205.1 NM_001127206.2 NM_001286267.1 NM_001286268.1 NM_001286269.1 NM_001286270.1 NM_001286271.1 NM_002134.3 XM_011522473.1 XM_017023196.1 XM_017023197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937069 c.605A>G p.Y202C Chr.16: 4508113-4508113 on Build GRCh38
5792498 c.658G>A p.V220M Chr.16: 4508166-4508166 on Build GRCh38
2920566 c.683A>C p.E228A Chr.16: 4508191-4508191 on Build GRCh38
138071 c.615G>T p.R205S Chr.16: 4508123-4508123 on Build GRCh38
7258379 c.573T>C p.N191N Chr.16: 4508081-4508081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746538008 Chr.16: 4508085-4508085 on Build GRCh38
rs751785391 Chr.16: 4508130-4508130 on Build GRCh38
rs540742826 Chr.16: 4508288-4508288 on Build GRCh38
rs901093070 Chr.16: 4508183-4508183 on Build GRCh38
rs779619149 Chr.16: 4508173-4508173 on Build GRCh38
rs745528281 Chr.16: 4508054-4508054 on Build GRCh38
rs2404579 Chr.16: 4508063-4508063 on Build GRCh38
rs749716286 Chr.16: 4508166-4508166 on Build GRCh38
rs17885831 Chr.16: 4508490-4508490 on Build GRCh38
rs948602116 Chr.16: 4508444-4508444 on Build GRCh38
rs572262225 Chr.16: 4508280-4508280 on Build GRCh38
rs768081169 Chr.16: 4508179-4508179 on Build GRCh38
rs552303061 Chr.16: 4508388-4508388 on Build GRCh38
rs1002620029 Chr.16: 4508180-4508180 on Build GRCh38
rs1004083555 Chr.16: 4508081-4508081 on Build GRCh38
rs563483460 Chr.16: 4508211-4508211 on Build GRCh38
rs998079904 Chr.16: 4508199-4508199 on Build GRCh38
rs542622112 Chr.16: 4508343-4508343 on Build GRCh38
rs762804813 Chr.16: 4508115-4508115 on Build GRCh38
rs968676097 Chr.16: 4508313-4508313 on Build GRCh38
rs773327395 Chr.16: 4508213-4508214 on Build GRCh38
rs1004113775 Chr.16: 4508095-4508095 on Build GRCh38
rs186971432 Chr.16: 4508129-4508129 on Build GRCh38
rs756560446 Chr.16: 4508149-4508149 on Build GRCh38
rs960496128 Chr.16: 4508333-4508344 on Build GRCh38
rs761521776 Chr.16: 4508270-4508270 on Build GRCh38
rs374010778 Chr.16: 4508194-4508194 on Build GRCh38
rs762353294 Chr.16: 4508178-4508178 on Build GRCh38
rs773893843 Chr.16: 4508305-4508305 on Build GRCh38
rs766550816 Chr.16: 4508121-4508121 on Build GRCh38
rs940715981 Chr.16: 4508469-4508469 on Build GRCh38
rs773199343 Chr.16: 4508104-4508104 on Build GRCh38
rs567262048 Chr.16: 4508052-4508052 on Build GRCh38
rs750223780 Chr.16: 4508472-4508472 on Build GRCh38
rs757608081 Chr.16: 4508171-4508171 on Build GRCh38
rs533743252 Chr.16: 4508322-4508322 on Build GRCh38
rs927305021 Chr.16: 4508386-4508386 on Build GRCh38
rs908406338 Chr.16: 4508362-4508362 on Build GRCh38
rs761801798 Chr.16: 4508090-4508090 on Build GRCh38
rs369627803 Chr.16: 4508144-4508144 on Build GRCh38
rs768188799 Chr.16: 4508087-4508087 on Build GRCh38
rs372467895 Chr.16: 4508232-4508232 on Build GRCh38
rs776731134 Chr.16: 4508089-4508089 on Build GRCh38
rs980353242 Chr.16: 4508340-4508340 on Build GRCh38
rs574001201 Chr.16: 4508323-4508323 on Build GRCh38
rs200491627 Chr.16: 4508051-4508051 on Build GRCh38
rs576222272 Chr.16: 4508135-4508135 on Build GRCh38
rs773254390 Chr.16: 4508212-4508212 on Build GRCh38
rs767602543 Chr.16: 4508234-4508234 on Build GRCh38
rs886562318 Chr.16: 4508449-4508449 on Build GRCh38
rs1038232267 Chr.16: 4508113-4508113 on Build GRCh38
rs770712405 Chr.16: 4508217-4508217 on Build GRCh38
rs190293833 Chr.16: 4508213-4508213 on Build GRCh38
rs1044106909 Chr.16: 4508448-4508449 on Build GRCh38
rs773876717 Chr.16: 4508254-4508254 on Build GRCh38
rs538917494 Chr.16: 4508165-4508165 on Build GRCh38
rs1032794073 Chr.16: 4508267-4508267 on Build GRCh38
rs560526718 Chr.16: 4508292-4508292 on Build GRCh38
rs998527399 Chr.16: 4508491-4508491 on Build GRCh38
rs754350119 Chr.16: 4508256-4508256 on Build GRCh38
rs975447449 Chr.16: 4508319-4508319 on Build GRCh38
rs765128900 Chr.16: 4508092-4508092 on Build GRCh38
rs760743555 Chr.16: 4508253-4508253 on Build GRCh38
rs1036873396 Chr.16: 4508479-4508479 on Build GRCh38
rs915516927 Chr.16: 4508294-4508294 on Build GRCh38
rs182054051 Chr.16: 4508441-4508441 on Build GRCh38

View Full Product Details