Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4510579-4511076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDIP1
Entrez Gene ID
29965
Gene Name
cell death-inducing p53 target 1
Gene Aliases
C16orf5, CDIP, I1, LITAFL
UniGene
Hs.654653
Gene Chromosome Location
Chr.16: 4510675-4538815 on Build GRCh38
Transcripts
NM_001199054.1 NM_001199055.1 NM_001199056.1 NM_013399.2

SNP Detail

SNP ID Chromosome Location
rs1016103208 Chr.16: 4510753-4510753 on Build GRCh38
rs929265987 Chr.16: 4510923-4510923 on Build GRCh38
rs7702 Chr.16: 4510928-4510928 on Build GRCh38
rs78481070 Chr.16: 4510655-4510656 on Build GRCh38
rs895209677 Chr.16: 4510699-4510699 on Build GRCh38
rs1048752034 Chr.16: 4510718-4510719 on Build GRCh38
rs1024441514 Chr.16: 4510789-4510789 on Build GRCh38
rs896207539 Chr.16: 4510771-4510772 on Build GRCh38
rs941386894 Chr.16: 4510987-4510987 on Build GRCh38
rs967696806 Chr.16: 4510817-4510817 on Build GRCh38
rs939398371 Chr.16: 4510967-4510967 on Build GRCh38
rs1017386987 Chr.16: 4510872-4510872 on Build GRCh38
rs939453252 Chr.16: 4510669-4510669 on Build GRCh38
rs4786506 Chr.16: 4510859-4510859 on Build GRCh38
rs377311606 Chr.16: 4511037-4511038 on Build GRCh38
rs572771469 Chr.16: 4510754-4510754 on Build GRCh38
rs1040155764 Chr.16: 4510649-4510649 on Build GRCh38
rs17882432 Chr.16: 4511009-4511009 on Build GRCh38
rs964812392 Chr.16: 4510873-4510873 on Build GRCh38
rs977453121 Chr.16: 4510620-4510633 on Build GRCh38
rs754204655 Chr.16: 4510934-4510934 on Build GRCh38
rs1014437618 Chr.16: 4510784-4510784 on Build GRCh38
rs1001982752 Chr.16: 4510765-4510765 on Build GRCh38
rs1037548455 Chr.16: 4510994-4510994 on Build GRCh38
rs573122882 Chr.16: 4510616-4510616 on Build GRCh38
rs906504202 Chr.16: 4510656-4510656 on Build GRCh38
rs756307938 Chr.16: 4510887-4510887 on Build GRCh38
rs1042375191 Chr.16: 4510628-4510628 on Build GRCh38
rs1477123 Chr.16: 4510626-4510626 on Build GRCh38
rs559442460 Chr.16: 4510744-4510744 on Build GRCh38
rs28490369 Chr.16: 4510603-4510603 on Build GRCh38
rs993380176 Chr.16: 4510796-4510796 on Build GRCh38
rs989608951 Chr.16: 4510874-4510874 on Build GRCh38
rs141328854 Chr.16: 4510827-4510827 on Build GRCh38
rs936133302 Chr.16: 4510621-4510622 on Build GRCh38
rs541774143 Chr.16: 4510767-4510767 on Build GRCh38
rs532919445 Chr.16: 4510997-4510997 on Build GRCh38
rs537323181 Chr.16: 4510621-4510621 on Build GRCh38
rs999143276 Chr.16: 4511033-4511033 on Build GRCh38
rs919075118 Chr.16: 4510891-4510891 on Build GRCh38
rs1055529311 Chr.16: 4510969-4510969 on Build GRCh38
rs185879608 Chr.16: 4510861-4510861 on Build GRCh38
rs545925585 Chr.16: 4510742-4510742 on Build GRCh38
rs539660031 Chr.16: 4510886-4510886 on Build GRCh38
rs928190052 Chr.16: 4510942-4510942 on Build GRCh38
rs910410870 Chr.16: 4510622-4510622 on Build GRCh38
rs975926222 Chr.16: 4510888-4510889 on Build GRCh38
rs17878254 Chr.16: 4510705-4510705 on Build GRCh38
rs767516682 Chr.16: 4510707-4510707 on Build GRCh38
rs796916129 Chr.16: 4510828-4510828 on Build GRCh38
rs890360852 Chr.16: 4511028-4511028 on Build GRCh38
rs565803400 Chr.16: 4511022-4511022 on Build GRCh38
rs367976305 Chr.16: 4510890-4510891 on Build GRCh38
rs1008601787 Chr.16: 4511051-4511051 on Build GRCh38
rs895272962 Chr.16: 4510981-4510981 on Build GRCh38
rs181713236 Chr.16: 4510838-4510838 on Build GRCh38
rs577505994 Chr.16: 4510694-4510694 on Build GRCh38
rs1049430027 Chr.16: 4510925-4510925 on Build GRCh38

View Full Product Details