Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 5085306-5085809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG1
Entrez Gene ID
56052
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
CDG1K, HMAT1, HMT-1, HMT1, MT-1, Mat-1, hMat-1
UniGene
Hs.592086
Gene Chromosome Location
Chr.16: 5071809-5087379 on Build GRCh38
Transcripts
NM_019109.4 XM_011522565.1 XM_017023457.1 XM_017023458.1 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

Gene Symbol
EEF2KMT
Entrez Gene ID
196483
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
FAM86A, SB153, eEF2-KMT
UniGene
Hs.406461
Gene Chromosome Location
Chr.16: 5084298-5097820 on Build GRCh38
Transcripts
NM_019109.4 XM_011522565.1 XM_017023457.1 XM_017023458.1 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6263082 c.976C>A p.L326M Chr.16: 5085649-5085649 on Build GRCh38
1378287 c.943T>A p.F315I Chr.16: 5085682-5085682 on Build GRCh38
6281036 c.933G>C p.E311D Chr.16: 5085692-5085692 on Build GRCh38
1708720 c.899C>T p.A300V Chr.16: 5085726-5085726 on Build GRCh38
971367 c.925C>T p.R309C Chr.16: 5085700-5085700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047523 Chr.16: 5085556-5085556 on Build GRCh38
rs1047536 Chr.16: 5085523-5085523 on Build GRCh38
rs556914293 Chr.16: 5085768-5085768 on Build GRCh38
rs190402165 Chr.16: 5085656-5085656 on Build GRCh38
rs1053501736 Chr.16: 5085564-5085564 on Build GRCh38
rs778931362 Chr.16: 5085766-5085766 on Build GRCh38
rs554392935 Chr.16: 5085400-5085400 on Build GRCh38
rs9921840 Chr.16: 5085547-5085547 on Build GRCh38
rs567864126 Chr.16: 5085722-5085722 on Build GRCh38
rs1047545 Chr.16: 5085504-5085504 on Build GRCh38
rs751541341 Chr.16: 5085741-5085742 on Build GRCh38
rs373657874 Chr.16: 5085627-5085627 on Build GRCh38
rs752557770 Chr.16: 5085762-5085764 on Build GRCh38
rs774688938 Chr.16: 5085703-5085703 on Build GRCh38
rs768020685 Chr.16: 5085749-5085749 on Build GRCh38
rs368350974 Chr.16: 5085752-5085752 on Build GRCh38
rs756661216 Chr.16: 5085759-5085759 on Build GRCh38
rs776562609 Chr.16: 5085645-5085645 on Build GRCh38
rs908298276 Chr.16: 5085475-5085475 on Build GRCh38
rs543353647 Chr.16: 5085463-5085463 on Build GRCh38
rs997598228 Chr.16: 5085595-5085595 on Build GRCh38
rs976303888 Chr.16: 5085427-5085427 on Build GRCh38
rs527816882 Chr.16: 5085577-5085577 on Build GRCh38
rs142385356 Chr.16: 5085668-5085668 on Build GRCh38
rs140141371 Chr.16: 5085655-5085655 on Build GRCh38
rs574276334 Chr.16: 5085461-5085461 on Build GRCh38
rs767269843 Chr.16: 5085745-5085745 on Build GRCh38
rs766190615 Chr.16: 5085712-5085712 on Build GRCh38
rs376514038 Chr.16: 5085620-5085620 on Build GRCh38
rs17137613 Chr.16: 5085474-5085474 on Build GRCh38
rs1047502 Chr.16: 5085585-5085585 on Build GRCh38
rs893250822 Chr.16: 5085571-5085571 on Build GRCh38
rs536814642 Chr.16: 5085748-5085748 on Build GRCh38
rs186337138 Chr.16: 5085621-5085621 on Build GRCh38
rs982829657 Chr.16: 5085472-5085472 on Build GRCh38
rs747976218 Chr.16: 5085777-5085777 on Build GRCh38
rs750076157 Chr.16: 5085597-5085597 on Build GRCh38
rs867352336 Chr.16: 5085331-5085331 on Build GRCh38
rs576528783 Chr.16: 5085481-5085481 on Build GRCh38
rs1047566 Chr.16: 5085452-5085452 on Build GRCh38
rs951003072 Chr.16: 5085607-5085607 on Build GRCh38
rs867547136 Chr.16: 5085351-5085351 on Build GRCh38
rs1047534 Chr.16: 5085541-5085541 on Build GRCh38
rs999228998 Chr.16: 5085332-5085332 on Build GRCh38
rs371589415 Chr.16: 5085657-5085657 on Build GRCh38
rs547938013 Chr.16: 5085716-5085716 on Build GRCh38
rs752852537 Chr.16: 5085567-5085567 on Build GRCh38
rs769041738 Chr.16: 5085612-5085612 on Build GRCh38
rs267604573 Chr.16: 5085677-5085677 on Build GRCh38
rs765890148 Chr.16: 5085746-5085746 on Build GRCh38
rs769633492 Chr.16: 5085644-5085645 on Build GRCh38
rs764130039 Chr.16: 5085584-5085584 on Build GRCh38
rs760509902 Chr.16: 5085581-5085581 on Build GRCh38
rs772580812 Chr.16: 5085704-5085704 on Build GRCh38
rs781737396 Chr.16: 5085664-5085664 on Build GRCh38
rs907075710 Chr.16: 5085554-5085554 on Build GRCh38
rs777980781 Chr.16: 5085685-5085685 on Build GRCh38
rs1017716654 Chr.16: 5085399-5085399 on Build GRCh38
rs568457681 Chr.16: 5085637-5085637 on Build GRCh38
rs973810353 Chr.16: 5085652-5085652 on Build GRCh38
rs1047553 Chr.16: 5085478-5085478 on Build GRCh38
rs565122912 Chr.16: 5085558-5085558 on Build GRCh38
rs1011715831 Chr.16: 5085369-5085369 on Build GRCh38
rs372527188 Chr.16: 5085783-5085783 on Build GRCh38
rs762304211 Chr.16: 5085782-5085782 on Build GRCh38
rs529796262 Chr.16: 5085619-5085619 on Build GRCh38
rs762532333 Chr.16: 5085710-5085710 on Build GRCh38
rs1047549 Chr.16: 5085495-5085495 on Build GRCh38
rs1047543 Chr.16: 5085516-5085516 on Build GRCh38
rs151297040 Chr.16: 5085699-5085699 on Build GRCh38
rs748185623 Chr.16: 5085618-5085618 on Build GRCh38
rs1047505 Chr.16: 5085576-5085576 on Build GRCh38
rs755384762 Chr.16: 5085662-5085662 on Build GRCh38
rs534404562 Chr.16: 5085380-5085380 on Build GRCh38
rs756217732 Chr.16: 5085674-5085674 on Build GRCh38
rs780942739 Chr.16: 5085770-5085770 on Build GRCh38
rs757368497 Chr.16: 5085765-5085765 on Build GRCh38
rs1047538 Chr.16: 5085521-5085521 on Build GRCh38
rs6775 Chr.16: 5085379-5085379 on Build GRCh38
rs771051209 Chr.16: 5085700-5085700 on Build GRCh38
rs16843033 Chr.16: 5085462-5085462 on Build GRCh38
rs541160794 Chr.16: 5085605-5085605 on Build GRCh38
rs768441103 Chr.16: 5085724-5085724 on Build GRCh38
rs761523989 Chr.16: 5085614-5085615 on Build GRCh38
rs561174573 Chr.16: 5085611-5085611 on Build GRCh38
rs773216565 Chr.16: 5085779-5085779 on Build GRCh38
rs371074454 Chr.16: 5085624-5085624 on Build GRCh38
rs751858067 Chr.16: 5085659-5085659 on Build GRCh38
rs761761355 Chr.16: 5085725-5085725 on Build GRCh38
rs778492820 Chr.16: 5085763-5085763 on Build GRCh38
rs749311788 Chr.16: 5085764-5085764 on Build GRCh38
rs531718461 Chr.16: 5085393-5085393 on Build GRCh38
rs552184574 Chr.16: 5085643-5085643 on Build GRCh38
rs762583437 Chr.16: 5085730-5085730 on Build GRCh38
rs201345819 Chr.16: 5085658-5085658 on Build GRCh38
rs1047576 Chr.16: 5085417-5085417 on Build GRCh38
rs759443326 Chr.16: 5085740-5085741 on Build GRCh38
rs1047573 Chr.16: 5085423-5085423 on Build GRCh38
rs915743830 Chr.16: 5085527-5085527 on Build GRCh38
rs1047498 Chr.16: 5085594-5085594 on Build GRCh38
rs781779977 Chr.16: 5085614-5085614 on Build GRCh38
rs867696905 Chr.16: 5085673-5085673 on Build GRCh38
rs780301989 Chr.16: 5085606-5085606 on Build GRCh38
rs1038233002 Chr.16: 5085513-5085513 on Build GRCh38
rs1031759797 Chr.16: 5085352-5085352 on Build GRCh38
rs778478979 Chr.16: 5085592-5085592 on Build GRCh38
rs758640759 Chr.16: 5085591-5085591 on Build GRCh38
rs1047514 Chr.16: 5085569-5085569 on Build GRCh38
rs747447071 Chr.16: 5085610-5085610 on Build GRCh38
rs760584474 Chr.16: 5085639-5085639 on Build GRCh38
rs772676442 Chr.16: 5085729-5085729 on Build GRCh38
rs1045658111 Chr.16: 5085552-5085552 on Build GRCh38

View Full Product Details