Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8713619-8714122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_000663.4 NM_020686.5 XM_011522400.2 XM_011522401.2

SNP Detail

SNP ID Chromosome Location
rs763113131 Chr.16: 8713730-8713730 on Build GRCh38
rs550300464 Chr.16: 8713839-8713839 on Build GRCh38
rs899301080 Chr.16: 8713828-8713828 on Build GRCh38
rs1001294125 Chr.16: 8713748-8713748 on Build GRCh38
rs920056531 Chr.16: 8714016-8714016 on Build GRCh38
rs187609685 Chr.16: 8713990-8713990 on Build GRCh38
rs8061339 Chr.16: 8714076-8714076 on Build GRCh38
rs1057146005 Chr.16: 8713750-8713750 on Build GRCh38
rs1025243686 Chr.16: 8713995-8713995 on Build GRCh38
rs765084996 Chr.16: 8713831-8713831 on Build GRCh38
rs1019229532 Chr.16: 8713672-8713674 on Build GRCh38
rs1048699606 Chr.16: 8713662-8713662 on Build GRCh38
rs901682894 Chr.16: 8713686-8713686 on Build GRCh38
rs956460513 Chr.16: 8713876-8713876 on Build GRCh38
rs968078076 Chr.16: 8713721-8713721 on Build GRCh38
rs1053560495 Chr.16: 8713677-8713677 on Build GRCh38
rs12447079 Chr.16: 8713714-8713714 on Build GRCh38
rs1030963667 Chr.16: 8713755-8713755 on Build GRCh38
rs147751577 Chr.16: 8713731-8713731 on Build GRCh38
rs969529379 Chr.16: 8713784-8713784 on Build GRCh38
rs956711603 Chr.16: 8713989-8713989 on Build GRCh38
rs750119251 Chr.16: 8714044-8714044 on Build GRCh38
rs762020030 Chr.16: 8713698-8713698 on Build GRCh38
rs903951432 Chr.16: 8713949-8713950 on Build GRCh38
rs573658821 Chr.16: 8714025-8714025 on Build GRCh38
rs1031457794 Chr.16: 8713963-8713963 on Build GRCh38
rs940768381 Chr.16: 8714004-8714004 on Build GRCh38
rs927047224 Chr.16: 8713809-8713809 on Build GRCh38
rs996788804 Chr.16: 8714062-8714062 on Build GRCh38
rs78607208 Chr.16: 8713841-8713841 on Build GRCh38
rs956399296 Chr.16: 8714097-8714097 on Build GRCh38
rs886416630 Chr.16: 8713999-8713999 on Build GRCh38
rs914981459 Chr.16: 8713908-8713908 on Build GRCh38
rs555303861 Chr.16: 8713996-8713996 on Build GRCh38
rs74898562 Chr.16: 8713772-8713772 on Build GRCh38
rs948008145 Chr.16: 8713696-8713696 on Build GRCh38
rs570893617 Chr.16: 8713691-8713691 on Build GRCh38
rs947903615 Chr.16: 8713955-8713955 on Build GRCh38
rs1040249200 Chr.16: 8713866-8713866 on Build GRCh38
rs140998674 Chr.16: 8713781-8713781 on Build GRCh38
rs778243213 Chr.16: 8714036-8714036 on Build GRCh38
rs552895445 Chr.16: 8713668-8713668 on Build GRCh38
rs1000948482 Chr.16: 8713952-8713952 on Build GRCh38
rs915033226 Chr.16: 8713685-8713685 on Build GRCh38
rs1030635050 Chr.16: 8714092-8714092 on Build GRCh38
rs182667657 Chr.16: 8713842-8713842 on Build GRCh38
rs755505000 Chr.16: 8713883-8713883 on Build GRCh38
rs535894364 Chr.16: 8713770-8713770 on Build GRCh38
rs185000107 Chr.16: 8714098-8714098 on Build GRCh38
rs1009066915 Chr.16: 8714091-8714091 on Build GRCh38
rs535908432 Chr.16: 8713852-8713852 on Build GRCh38
rs531761151 Chr.16: 8713709-8713709 on Build GRCh38
rs541119191 Chr.16: 8714026-8714026 on Build GRCh38
rs995980941 Chr.16: 8713883-8713884 on Build GRCh38
rs1004930802 Chr.16: 8713789-8713789 on Build GRCh38
rs927903362 Chr.16: 8713980-8713980 on Build GRCh38
rs940643583 Chr.16: 8713843-8713843 on Build GRCh38
rs765942833 Chr.16: 8713851-8713851 on Build GRCh38
rs766419278 Chr.16: 8713800-8713800 on Build GRCh38
rs543067946 Chr.16: 8713892-8713892 on Build GRCh38
rs1038220425 Chr.16: 8714035-8714035 on Build GRCh38
rs753196189 Chr.16: 8713974-8713974 on Build GRCh38
rs938703613 Chr.16: 8713734-8713734 on Build GRCh38
rs575906448 Chr.16: 8713875-8713875 on Build GRCh38
rs555362441 Chr.16: 8714077-8714077 on Build GRCh38
rs886356731 Chr.16: 8713665-8713665 on Build GRCh38
rs768053837 Chr.16: 8713958-8713958 on Build GRCh38

View Full Product Details