Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8768101-8768601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_000663.4 NM_001127448.1 NM_020686.5 XM_011522400.2 XM_011522401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196520 c.623A>G p.Y208C Chr.16: 8768212-8768212 on Build GRCh38
180129 c.619G>A p.D207N Chr.16: 8768208-8768208 on Build GRCh38
435910 c.645C>T p.G215G Chr.16: 8768234-8768234 on Build GRCh38
4063479 c.660G>A p.R220R Chr.16: 8768249-8768249 on Build GRCh38
4503254 c.635C>T p.S212F Chr.16: 8768224-8768224 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1031088710 Chr.16: 8768548-8768548 on Build GRCh38
rs892522006 Chr.16: 8768549-8768549 on Build GRCh38
rs757286059 Chr.16: 8768545-8768545 on Build GRCh38
rs932554811 Chr.16: 8768507-8768507 on Build GRCh38
rs752369134 Chr.16: 8768433-8768433 on Build GRCh38
rs982544293 Chr.16: 8768458-8768458 on Build GRCh38
rs1009281467 Chr.16: 8768546-8768546 on Build GRCh38
rs536262347 Chr.16: 8768475-8768475 on Build GRCh38
rs895015601 Chr.16: 8768463-8768463 on Build GRCh38
rs939137461 Chr.16: 8768450-8768450 on Build GRCh38
rs756875305 Chr.16: 8768274-8768274 on Build GRCh38
rs13332215 Chr.16: 8768550-8768550 on Build GRCh38
rs369887885 Chr.16: 8768289-8768289 on Build GRCh38
rs747042892 Chr.16: 8768212-8768212 on Build GRCh38
rs961004192 Chr.16: 8768473-8768473 on Build GRCh38
rs972438537 Chr.16: 8768477-8768477 on Build GRCh38
rs1016829265 Chr.16: 8768470-8768470 on Build GRCh38
rs780199799 Chr.16: 8768564-8768564 on Build GRCh38
rs770138381 Chr.16: 8768297-8768297 on Build GRCh38
rs573277423 Chr.16: 8768127-8768127 on Build GRCh38
rs74815154 Chr.16: 8768558-8768558 on Build GRCh38
rs751426054 Chr.16: 8768152-8768152 on Build GRCh38
rs376594468 Chr.16: 8768148-8768148 on Build GRCh38
rs762742483 Chr.16: 8768375-8768376 on Build GRCh38
rs557781212 Chr.16: 8768325-8768325 on Build GRCh38
rs369668077 Chr.16: 8768158-8768158 on Build GRCh38
rs980462294 Chr.16: 8768438-8768438 on Build GRCh38
rs886052430 Chr.16: 8768264-8768265 on Build GRCh38
rs760072765 Chr.16: 8768265-8768265 on Build GRCh38
rs752253647 Chr.16: 8768175-8768175 on Build GRCh38
rs768799493 Chr.16: 8768568-8768568 on Build GRCh38
rs753309345 Chr.16: 8768271-8768271 on Build GRCh38
rs769070363 Chr.16: 8768288-8768288 on Build GRCh38
rs1139522 Chr.16: 8768234-8768234 on Build GRCh38
rs1028918962 Chr.16: 8768309-8768309 on Build GRCh38
rs1010263326 Chr.16: 8768423-8768423 on Build GRCh38
rs773628864 Chr.16: 8768306-8768306 on Build GRCh38
rs8049072 Chr.16: 8768355-8768355 on Build GRCh38
rs772588617 Chr.16: 8768144-8768144 on Build GRCh38
rs767550217 Chr.16: 8768237-8768237 on Build GRCh38
rs1049934109 Chr.16: 8768322-8768322 on Build GRCh38
rs8057362 Chr.16: 8768439-8768439 on Build GRCh38
rs768826504 Chr.16: 8768225-8768225 on Build GRCh38
rs947705390 Chr.16: 8768137-8768137 on Build GRCh38
rs1031623084 Chr.16: 8768347-8768347 on Build GRCh38
rs546406644 Chr.16: 8768275-8768275 on Build GRCh38
rs181320059 Chr.16: 8768502-8768502 on Build GRCh38
rs908254303 Chr.16: 8768469-8768469 on Build GRCh38
rs1005358602 Chr.16: 8768580-8768580 on Build GRCh38
rs1038256499 Chr.16: 8768268-8768268 on Build GRCh38
rs899771475 Chr.16: 8768503-8768503 on Build GRCh38
rs762050203 Chr.16: 8768232-8768232 on Build GRCh38
rs1029792867 Chr.16: 8768483-8768484 on Build GRCh38
rs753344688 Chr.16: 8768525-8768525 on Build GRCh38
rs779198838 Chr.16: 8768201-8768201 on Build GRCh38
rs1033978532 Chr.16: 8768159-8768159 on Build GRCh38
rs566591045 Chr.16: 8768505-8768505 on Build GRCh38
rs752732104 Chr.16: 8768240-8768240 on Build GRCh38
rs371057736 Chr.16: 8768295-8768295 on Build GRCh38
rs369997983 Chr.16: 8768126-8768126 on Build GRCh38
rs578109728 Chr.16: 8768569-8768569 on Build GRCh38
rs545380653 Chr.16: 8768321-8768321 on Build GRCh38
rs149271402 Chr.16: 8768230-8768230 on Build GRCh38
rs911438259 Chr.16: 8768345-8768345 on Build GRCh38
rs121434578 Chr.16: 8768248-8768248 on Build GRCh38
rs756890572 Chr.16: 8768193-8768193 on Build GRCh38
rs373357773 Chr.16: 8768207-8768207 on Build GRCh38
rs33948976 Chr.16: 8768376-8768376 on Build GRCh38
rs767568755 Chr.16: 8768163-8768163 on Build GRCh38
rs989609493 Chr.16: 8768378-8768378 on Build GRCh38
rs763619234 Chr.16: 8768269-8768269 on Build GRCh38
rs941199667 Chr.16: 8768487-8768487 on Build GRCh38
rs766162227 Chr.16: 8768149-8768149 on Build GRCh38
rs724159990 Chr.16: 8768220-8768220 on Build GRCh38
rs777269086 Chr.16: 8768186-8768186 on Build GRCh38
rs969600620 Chr.16: 8768424-8768424 on Build GRCh38
rs747587543 Chr.16: 8768283-8768283 on Build GRCh38
rs376506551 Chr.16: 8768276-8768276 on Build GRCh38
rs373385788 Chr.16: 8768147-8768147 on Build GRCh38
rs759472888 Chr.16: 8768236-8768236 on Build GRCh38
rs1139523 Chr.16: 8768235-8768235 on Build GRCh38
rs894963157 Chr.16: 8768575-8768575 on Build GRCh38
rs907234474 Chr.16: 8768563-8768563 on Build GRCh38
rs890562013 Chr.16: 8768310-8768310 on Build GRCh38
rs772498753 Chr.16: 8768208-8768208 on Build GRCh38
rs1050899198 Chr.16: 8768524-8768524 on Build GRCh38
rs755708488 Chr.16: 8768177-8768177 on Build GRCh38
rs775488254 Chr.16: 8768145-8768145 on Build GRCh38
rs760745761 Chr.16: 8768146-8768146 on Build GRCh38
rs530804959 Chr.16: 8768160-8768160 on Build GRCh38
rs200255576 Chr.16: 8768376-8768376 on Build GRCh38
rs748783252 Chr.16: 8768261-8768262 on Build GRCh38
rs542575561 Chr.16: 8768187-8768187 on Build GRCh38
rs1033470120 Chr.16: 8768454-8768454 on Build GRCh38
rs561050474 Chr.16: 8768210-8768210 on Build GRCh38
rs8053509 Chr.16: 8768414-8768414 on Build GRCh38

View Full Product Details