Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9118166-9118670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf72
Entrez Gene ID
29035
Gene Name
chromosome 16 open reading frame 72
Gene Aliases
PRO0149
UniGene
Hs.221497
Gene Chromosome Location
Chr.16: 9091329-9119698 on Build GRCh38
Transcripts
NM_014117.2

SNP Detail

SNP ID Chromosome Location
rs893086585 Chr.16: 9118383-9118383 on Build GRCh38
rs138039181 Chr.16: 9118266-9118266 on Build GRCh38
rs564426772 Chr.16: 9118564-9118564 on Build GRCh38
rs962971103 Chr.16: 9118509-9118509 on Build GRCh38
rs573199186 Chr.16: 9118499-9118499 on Build GRCh38
rs557898500 Chr.16: 9118378-9118378 on Build GRCh38
rs1003040153 Chr.16: 9118438-9118438 on Build GRCh38
rs143879237 Chr.16: 9118622-9118622 on Build GRCh38
rs754224763 Chr.16: 9118319-9118319 on Build GRCh38
rs572359372 Chr.16: 9118324-9118324 on Build GRCh38
rs537114015 Chr.16: 9118256-9118256 on Build GRCh38
rs559706246 Chr.16: 9118586-9118586 on Build GRCh38
rs1035607674 Chr.16: 9118274-9118274 on Build GRCh38
rs569881068 Chr.16: 9118229-9118229 on Build GRCh38
rs554444322 Chr.16: 9118322-9118322 on Build GRCh38
rs769072796 Chr.16: 9118218-9118222 on Build GRCh38
rs1038597116 Chr.16: 9118631-9118631 on Build GRCh38
rs73495968 Chr.16: 9118435-9118435 on Build GRCh38
rs1053035246 Chr.16: 9118368-9118368 on Build GRCh38
rs954114527 Chr.16: 9118566-9118566 on Build GRCh38
rs1028749957 Chr.16: 9118542-9118542 on Build GRCh38
rs562079622 Chr.16: 9118483-9118483 on Build GRCh38
rs912401327 Chr.16: 9118325-9118325 on Build GRCh38
rs774470882 Chr.16: 9118195-9118195 on Build GRCh38
rs974505344 Chr.16: 9118317-9118317 on Build GRCh38
rs878996327 Chr.16: 9118456-9118456 on Build GRCh38
rs934540437 Chr.16: 9118354-9118354 on Build GRCh38
rs988665331 Chr.16: 9118607-9118607 on Build GRCh38
rs983015539 Chr.16: 9118304-9118304 on Build GRCh38
rs35494642 Chr.16: 9118464-9118465 on Build GRCh38
rs994424743 Chr.16: 9118644-9118644 on Build GRCh38
rs912368799 Chr.16: 9118594-9118594 on Build GRCh38
rs892824017 Chr.16: 9118231-9118231 on Build GRCh38
rs1011220865 Chr.16: 9118404-9118404 on Build GRCh38
rs192382060 Chr.16: 9118612-9118612 on Build GRCh38
rs758495559 Chr.16: 9118599-9118599 on Build GRCh38
rs563450396 Chr.16: 9118636-9118636 on Build GRCh38
rs1011300385 Chr.16: 9118233-9118233 on Build GRCh38
rs907174884 Chr.16: 9118214-9118218 on Build GRCh38
rs149452835 Chr.16: 9118312-9118312 on Build GRCh38
rs74861452 Chr.16: 9118227-9118227 on Build GRCh38
rs534361441 Chr.16: 9118314-9118314 on Build GRCh38
rs536739128 Chr.16: 9118279-9118279 on Build GRCh38
rs781550749 Chr.16: 9118229-9118230 on Build GRCh38
rs937288305 Chr.16: 9118553-9118553 on Build GRCh38
rs928710081 Chr.16: 9118527-9118530 on Build GRCh38
rs539756949 Chr.16: 9118340-9118340 on Build GRCh38
rs557571023 Chr.16: 9118237-9118237 on Build GRCh38
rs183903688 Chr.16: 9118473-9118473 on Build GRCh38
rs895068892 Chr.16: 9118250-9118250 on Build GRCh38
rs553131738 Chr.16: 9118479-9118479 on Build GRCh38
rs56154228 Chr.16: 9118504-9118504 on Build GRCh38
rs1044490878 Chr.16: 9118410-9118410 on Build GRCh38
rs766993056 Chr.16: 9118228-9118228 on Build GRCh38
rs1045751201 Chr.16: 9118203-9118203 on Build GRCh38
rs747193576 Chr.16: 9118639-9118639 on Build GRCh38
rs750981838 Chr.16: 9118592-9118592 on Build GRCh38
rs773377849 Chr.16: 9118498-9118498 on Build GRCh38
rs974092836 Chr.16: 9118540-9118540 on Build GRCh38
rs188894009 Chr.16: 9118494-9118494 on Build GRCh38
rs759373261 Chr.16: 9118235-9118235 on Build GRCh38
rs1055836659 Chr.16: 9118576-9118576 on Build GRCh38

View Full Product Details