Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27067961-27068469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf82
Entrez Gene ID
162083
Gene Name
chromosome 16 open reading frame 82
Gene Aliases
TNT
UniGene
Hs.120831
Gene Chromosome Location
Chr.16: 27066898-27069166 on Build GRCh38
Transcripts
NM_001145545.1

SNP Detail

SNP ID Chromosome Location
rs374111561 Chr.16: 27068017-27068017 on Build GRCh38
rs186988304 Chr.16: 27068194-27068194 on Build GRCh38
rs148207873 Chr.16: 27068397-27068397 on Build GRCh38
rs927319984 Chr.16: 27068051-27068051 on Build GRCh38
rs778943774 Chr.16: 27068186-27068186 on Build GRCh38
rs891683376 Chr.16: 27068439-27068439 on Build GRCh38
rs545514341 Chr.16: 27068252-27068252 on Build GRCh38
rs891740661 Chr.16: 27068200-27068200 on Build GRCh38
rs909649381 Chr.16: 27068075-27068075 on Build GRCh38
rs543582795 Chr.16: 27068124-27068125 on Build GRCh38
rs1018406334 Chr.16: 27068311-27068311 on Build GRCh38
rs935633916 Chr.16: 27068426-27068426 on Build GRCh38
rs377485885 Chr.16: 27068400-27068400 on Build GRCh38
rs892623248 Chr.16: 27068144-27068144 on Build GRCh38
rs751021437 Chr.16: 27068382-27068382 on Build GRCh38
rs570029802 Chr.16: 27068080-27068080 on Build GRCh38
rs553890986 Chr.16: 27068124-27068124 on Build GRCh38
rs796702033 Chr.16: 27068100-27068100 on Build GRCh38
rs144811712 Chr.16: 27068228-27068228 on Build GRCh38
rs200068485 Chr.16: 27068095-27068096 on Build GRCh38
rs112061367 Chr.16: 27068350-27068350 on Build GRCh38
rs386790114 Chr.16: 27068424-27068425 on Build GRCh38
rs1043882116 Chr.16: 27068322-27068322 on Build GRCh38
rs866466334 Chr.16: 27068210-27068210 on Build GRCh38
rs988781494 Chr.16: 27068253-27068253 on Build GRCh38
rs947671227 Chr.16: 27068079-27068079 on Build GRCh38
rs901829987 Chr.16: 27068421-27068421 on Build GRCh38
rs900183919 Chr.16: 27068123-27068123 on Build GRCh38
rs559363741 Chr.16: 27068270-27068270 on Build GRCh38
rs115622223 Chr.16: 27068424-27068424 on Build GRCh38
rs866933595 Chr.16: 27068096-27068096 on Build GRCh38
rs566121324 Chr.16: 27068431-27068431 on Build GRCh38
rs1039069409 Chr.16: 27068102-27068102 on Build GRCh38
rs547537652 Chr.16: 27068320-27068320 on Build GRCh38
rs1043768372 Chr.16: 27068178-27068179 on Build GRCh38
rs980478040 Chr.16: 27068039-27068039 on Build GRCh38
rs183298388 Chr.16: 27068389-27068389 on Build GRCh38
rs962731741 Chr.16: 27068235-27068235 on Build GRCh38
rs974071020 Chr.16: 27068312-27068312 on Build GRCh38
rs766063244 Chr.16: 27067986-27067986 on Build GRCh38
rs943447069 Chr.16: 27068391-27068391 on Build GRCh38
rs866636392 Chr.16: 27068224-27068224 on Build GRCh38
rs976574392 Chr.16: 27067987-27067987 on Build GRCh38
rs535937440 Chr.16: 27068179-27068179 on Build GRCh38
rs935873405 Chr.16: 27068149-27068149 on Build GRCh38
rs986894337 Chr.16: 27068390-27068390 on Build GRCh38
rs944478400 Chr.16: 27068101-27068101 on Build GRCh38
rs7190095 Chr.16: 27068205-27068205 on Build GRCh38
rs1024149330 Chr.16: 27068206-27068206 on Build GRCh38
rs184084005 Chr.16: 27068095-27068095 on Build GRCh38
rs191967871 Chr.16: 27068219-27068219 on Build GRCh38
rs1011053786 Chr.16: 27068150-27068150 on Build GRCh38
rs963265980 Chr.16: 27068067-27068067 on Build GRCh38
rs918416778 Chr.16: 27067991-27067991 on Build GRCh38
rs918870930 Chr.16: 27068087-27068087 on Build GRCh38
rs572114870 Chr.16: 27068319-27068319 on Build GRCh38
rs549722762 Chr.16: 27068025-27068025 on Build GRCh38
rs561155610 Chr.16: 27068344-27068344 on Build GRCh38
rs115695246 Chr.16: 27068092-27068092 on Build GRCh38
rs116780507 Chr.16: 27068425-27068425 on Build GRCh38
rs914174230 Chr.16: 27068272-27068272 on Build GRCh38
rs114634022 Chr.16: 27068229-27068229 on Build GRCh38
rs540561548 Chr.16: 27068184-27068184 on Build GRCh38
rs550041165 Chr.16: 27068396-27068396 on Build GRCh38
rs748079066 Chr.16: 27068198-27068198 on Build GRCh38

View Full Product Details