Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28822784-28823278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN2L
Entrez Gene ID
11273
Gene Name
ataxin 2 like
Gene Aliases
A2D, A2LG, A2LP, A2RP
UniGene
Hs.460499
Gene Chromosome Location
Chr.16: 28823048-28837237 on Build GRCh38
Transcripts
NM_001308230.1 NM_007245.3 NM_017492.3 NM_145714.2 NM_148414.2 NM_148415.2 NM_148416.2 XM_005255061.1 XM_005255062.1 XM_005255063.3 XM_005255064.1 XM_005255065.1 XM_005255066.1 XM_005255067.1 XM_005255068.1 XM_005255069.1 XM_005255070.1 XM_005255071.1 XM_005255074.1 XM_005255075.1 XM_005255076.1 XM_006721007.1 XM_006721008.1 XM_006721009.1 XM_006721010.2 XM_006721011.1 XM_006721012.3 XM_011545719.2 XM_011545721.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431311 c.16C>A p.P6T Chr.16: 28823275-28823275 on Build GRCh38
4779360 c.15G>A p.Q5Q Chr.16: 28823274-28823274 on Build GRCh38
6431312 c.16C>A p.P6T Chr.16: 28823275-28823275 on Build GRCh38
6431313 c.16C>A p.P6T Chr.16: 28823275-28823275 on Build GRCh38
4779358 c.15G>A p.Q5Q Chr.16: 28823274-28823274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025342813 Chr.16: 28822806-28822806 on Build GRCh38
rs886685365 Chr.16: 28822846-28822848 on Build GRCh38
rs796686806 Chr.16: 28823186-28823186 on Build GRCh38
rs762069653 Chr.16: 28823257-28823257 on Build GRCh38
rs1013186241 Chr.16: 28822882-28822882 on Build GRCh38
rs1002455709 Chr.16: 28822868-28822868 on Build GRCh38
rs1001472493 Chr.16: 28823066-28823066 on Build GRCh38
rs769277884 Chr.16: 28823226-28823226 on Build GRCh38
rs967207214 Chr.16: 28823089-28823089 on Build GRCh38
rs8056390 Chr.16: 28822935-28822935 on Build GRCh38
rs1052363361 Chr.16: 28823017-28823017 on Build GRCh38
rs1026841501 Chr.16: 28822887-28822887 on Build GRCh38
rs372382912 Chr.16: 28822913-28822913 on Build GRCh38
rs1001588505 Chr.16: 28822862-28822863 on Build GRCh38
rs1037928542 Chr.16: 28823037-28823037 on Build GRCh38
rs751326719 Chr.16: 28823215-28823216 on Build GRCh38
rs1042353868 Chr.16: 28822851-28822851 on Build GRCh38
rs555686638 Chr.16: 28822964-28822964 on Build GRCh38
rs1016682217 Chr.16: 28822848-28822848 on Build GRCh38
rs940845740 Chr.16: 28823187-28823187 on Build GRCh38
rs914825665 Chr.16: 28822836-28822836 on Build GRCh38
rs542471701 Chr.16: 28822822-28822822 on Build GRCh38
rs963734394 Chr.16: 28823112-28823112 on Build GRCh38
rs978666300 Chr.16: 28822897-28822897 on Build GRCh38
rs1004925464 Chr.16: 28822846-28822846 on Build GRCh38
rs757925822 Chr.16: 28822936-28822951 on Build GRCh38
rs201131521 Chr.16: 28822934-28822934 on Build GRCh38
rs754101632 Chr.16: 28823246-28823246 on Build GRCh38
rs894201019 Chr.16: 28822871-28822871 on Build GRCh38
rs902898055 Chr.16: 28822865-28822865 on Build GRCh38
rs922387346 Chr.16: 28823153-28823153 on Build GRCh38
rs1034430458 Chr.16: 28823080-28823080 on Build GRCh38
rs777760535 Chr.16: 28822824-28822824 on Build GRCh38
rs540625397 Chr.16: 28823205-28823205 on Build GRCh38
rs762800477 Chr.16: 28823233-28823233 on Build GRCh38
rs933733231 Chr.16: 28823161-28823162 on Build GRCh38
rs772716756 Chr.16: 28823232-28823232 on Build GRCh38
rs900687625 Chr.16: 28822812-28822812 on Build GRCh38
rs75139976 Chr.16: 28823218-28823218 on Build GRCh38
rs754748864 Chr.16: 28823228-28823231 on Build GRCh38
rs553300876 Chr.16: 28822818-28822818 on Build GRCh38
rs886571793 Chr.16: 28823021-28823021 on Build GRCh38
rs908049277 Chr.16: 28823176-28823176 on Build GRCh38
rs768345080 Chr.16: 28823222-28823222 on Build GRCh38
rs895901189 Chr.16: 28823083-28823083 on Build GRCh38
rs1038384273 Chr.16: 28823191-28823191 on Build GRCh38
rs758300889 Chr.16: 28823214-28823215 on Build GRCh38
rs774376478 Chr.16: 28822938-28822951 on Build GRCh38
rs533522970 Chr.16: 28822815-28822815 on Build GRCh38
rs1020544032 Chr.16: 28823087-28823087 on Build GRCh38
rs1030844576 Chr.16: 28823063-28823063 on Build GRCh38
rs78683834 Chr.16: 28822934-28822934 on Build GRCh38
rs777945050 Chr.16: 28823239-28823239 on Build GRCh38
rs932144616 Chr.16: 28822811-28822811 on Build GRCh38
rs1020240219 Chr.16: 28822885-28822885 on Build GRCh38
rs559656630 Chr.16: 28822822-28822823 on Build GRCh38
rs529182697 Chr.16: 28823240-28823240 on Build GRCh38
rs770857185 Chr.16: 28823234-28823234 on Build GRCh38
rs937381106 Chr.16: 28823210-28823210 on Build GRCh38
rs977899227 Chr.16: 28822955-28822955 on Build GRCh38
rs1032961018 Chr.16: 28823241-28823241 on Build GRCh38
rs779858443 Chr.16: 28823214-28823217 on Build GRCh38
rs767354296 Chr.16: 28822939-28822951 on Build GRCh38
rs1037669016 Chr.16: 28822882-28822903 on Build GRCh38
rs1034373442 Chr.16: 28822873-28822873 on Build GRCh38
rs1052055051 Chr.16: 28822841-28822841 on Build GRCh38
rs933512577 Chr.16: 28823012-28823012 on Build GRCh38
rs780312498 Chr.16: 28823220-28823220 on Build GRCh38
rs923310826 Chr.16: 28822810-28822810 on Build GRCh38
rs975069232 Chr.16: 28823136-28823136 on Build GRCh38
rs922331771 Chr.16: 28822967-28822967 on Build GRCh38
rs528999842 Chr.16: 28822884-28822884 on Build GRCh38
rs1014299767 Chr.16: 28823086-28823086 on Build GRCh38
rs747348967 Chr.16: 28823221-28823221 on Build GRCh38
rs564553864 Chr.16: 28823198-28823198 on Build GRCh38
rs1032909182 Chr.16: 28823105-28823105 on Build GRCh38
rs947691694 Chr.16: 28822840-28822840 on Build GRCh38
rs925621029 Chr.16: 28822912-28822912 on Build GRCh38
rs575769191 Chr.16: 28823043-28823043 on Build GRCh38
rs899412626 Chr.16: 28823209-28823209 on Build GRCh38
rs979102742 Chr.16: 28823097-28823097 on Build GRCh38
rs940949330 Chr.16: 28823030-28823030 on Build GRCh38
rs1056819125 Chr.16: 28822870-28822870 on Build GRCh38
rs372126467 Chr.16: 28823235-28823235 on Build GRCh38
rs1050587967 Chr.16: 28822813-28822813 on Build GRCh38
rs1055723879 Chr.16: 28823212-28823212 on Build GRCh38
rs189978904 Chr.16: 28823100-28823100 on Build GRCh38
rs989266107 Chr.16: 28823111-28823111 on Build GRCh38
rs573192299 Chr.16: 28822819-28822819 on Build GRCh38
rs982521387 Chr.16: 28823164-28823164 on Build GRCh38
rs780789806 Chr.16: 28823224-28823224 on Build GRCh38
rs897818760 Chr.16: 28822964-28822977 on Build GRCh38
rs533391270 Chr.16: 28823199-28823199 on Build GRCh38
rs764481046 Chr.16: 28823242-28823242 on Build GRCh38

View Full Product Details