Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30005411-30005904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1 NM_001304562.1 NM_001304563.1 NM_173618.2 NR_130786.1 NR_130787.1 NR_134855.1 XM_011545809.2

Gene Symbol
INO80E
Entrez Gene ID
283899
Gene Name
INO80 complex subunit E
Gene Aliases
CCDC95
UniGene
Hs.434864
Gene Chromosome Location
Chr.16: 29996209-30005794 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1 NM_001304562.1 NM_001304563.1 NM_173618.2 NR_130786.1 NR_130787.1 NR_134855.1 XM_011545809.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4826841 c.706G>A p.D236N Chr.16: 30005413-30005413 on Build GRCh38
3888380 c.720C>T p.I240I Chr.16: 30005427-30005427 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953366984 Chr.16: 30005825-30005825 on Build GRCh38
rs891824186 Chr.16: 30005501-30005501 on Build GRCh38
rs199926778 Chr.16: 30005465-30005465 on Build GRCh38
rs370401881 Chr.16: 30005814-30005814 on Build GRCh38
rs553762569 Chr.16: 30005531-30005531 on Build GRCh38
rs561571360 Chr.16: 30005663-30005663 on Build GRCh38
rs927230527 Chr.16: 30005461-30005461 on Build GRCh38
rs149145925 Chr.16: 30005482-30005482 on Build GRCh38
rs764531081 Chr.16: 30005456-30005456 on Build GRCh38
rs143249661 Chr.16: 30005499-30005499 on Build GRCh38
rs758814328 Chr.16: 30005694-30005694 on Build GRCh38
rs1036125307 Chr.16: 30005805-30005806 on Build GRCh38
rs557746446 Chr.16: 30005634-30005634 on Build GRCh38
rs748186316 Chr.16: 30005483-30005483 on Build GRCh38
rs962640287 Chr.16: 30005577-30005577 on Build GRCh38
rs923700412 Chr.16: 30005646-30005646 on Build GRCh38
rs202176231 Chr.16: 30005468-30005468 on Build GRCh38
rs112388896 Chr.16: 30005489-30005489 on Build GRCh38
rs3186442 Chr.16: 30005718-30005718 on Build GRCh38
rs1005140569 Chr.16: 30005795-30005796 on Build GRCh38
rs751381829 Chr.16: 30005479-30005479 on Build GRCh38
rs750486252 Chr.16: 30005469-30005469 on Build GRCh38
rs138990820 Chr.16: 30005738-30005738 on Build GRCh38
rs143662477 Chr.16: 30005444-30005444 on Build GRCh38
rs944840895 Chr.16: 30005651-30005651 on Build GRCh38
rs776037527 Chr.16: 30005454-30005454 on Build GRCh38
rs995401913 Chr.16: 30005877-30005877 on Build GRCh38
rs966002399 Chr.16: 30005452-30005452 on Build GRCh38
rs545832540 Chr.16: 30005591-30005591 on Build GRCh38
rs375613929 Chr.16: 30005865-30005865 on Build GRCh38
rs769608225 Chr.16: 30005487-30005487 on Build GRCh38
rs748776711 Chr.16: 30005491-30005491 on Build GRCh38
rs978002069 Chr.16: 30005880-30005880 on Build GRCh38
rs371474779 Chr.16: 30005474-30005474 on Build GRCh38
rs370089233 Chr.16: 30005876-30005876 on Build GRCh38
rs752994375 Chr.16: 30005687-30005687 on Build GRCh38
rs1056115680 Chr.16: 30005513-30005513 on Build GRCh38
rs570218227 Chr.16: 30005814-30005814 on Build GRCh38
rs907288962 Chr.16: 30005840-30005840 on Build GRCh38
rs771386740 Chr.16: 30005637-30005637 on Build GRCh38
rs150020641 Chr.16: 30005463-30005463 on Build GRCh38
rs754748173 Chr.16: 30005481-30005481 on Build GRCh38
rs1049438420 Chr.16: 30005739-30005739 on Build GRCh38
rs1011857167 Chr.16: 30005849-30005849 on Build GRCh38
rs770686216 Chr.16: 30005451-30005451 on Build GRCh38
rs12851 Chr.16: 30005656-30005656 on Build GRCh38
rs201230248 Chr.16: 30005445-30005445 on Build GRCh38
rs971280798 Chr.16: 30005453-30005453 on Build GRCh38
rs901939747 Chr.16: 30005806-30005806 on Build GRCh38
rs779478784 Chr.16: 30005629-30005629 on Build GRCh38
rs532516778 Chr.16: 30005705-30005705 on Build GRCh38
rs918232040 Chr.16: 30005585-30005585 on Build GRCh38
rs956321757 Chr.16: 30005442-30005442 on Build GRCh38
rs1034231664 Chr.16: 30005554-30005554 on Build GRCh38
rs540898779 Chr.16: 30005466-30005466 on Build GRCh38
rs1046199344 Chr.16: 30005839-30005839 on Build GRCh38
rs373302757 Chr.16: 30005455-30005455 on Build GRCh38
rs74015003 Chr.16: 30005478-30005478 on Build GRCh38
rs747629099 Chr.16: 30005435-30005435 on Build GRCh38
rs751798869 Chr.16: 30005868-30005868 on Build GRCh38
rs556440077 Chr.16: 30005504-30005504 on Build GRCh38
rs576322517 Chr.16: 30005641-30005641 on Build GRCh38
rs990800138 Chr.16: 30005597-30005597 on Build GRCh38
rs1001378835 Chr.16: 30005553-30005553 on Build GRCh38
rs537229802 Chr.16: 30005490-30005490 on Build GRCh38
rs887599585 Chr.16: 30005540-30005540 on Build GRCh38
rs373830911 Chr.16: 30005443-30005443 on Build GRCh38
rs3186441 Chr.16: 30005685-30005685 on Build GRCh38
rs188366782 Chr.16: 30005722-30005722 on Build GRCh38
rs529303540 Chr.16: 30005699-30005699 on Build GRCh38
rs560196995 Chr.16: 30005578-30005578 on Build GRCh38
rs891789387 Chr.16: 30005772-30005772 on Build GRCh38
rs890599570 Chr.16: 30005475-30005475 on Build GRCh38
rs1028912323 Chr.16: 30005579-30005579 on Build GRCh38
rs111261004 Chr.16: 30005619-30005619 on Build GRCh38
rs183654095 Chr.16: 30005519-30005519 on Build GRCh38
rs777928473 Chr.16: 30005759-30005759 on Build GRCh38
rs867653136 Chr.16: 30005806-30005806 on Build GRCh38
rs545014761 Chr.16: 30005523-30005523 on Build GRCh38
rs551784891 Chr.16: 30005804-30005804 on Build GRCh38
rs1034199275 Chr.16: 30005873-30005873 on Build GRCh38
rs200708746 Chr.16: 30005436-30005436 on Build GRCh38

View Full Product Details