Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30008918-30009418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968575 c.347T>A p.L116H Chr.16: 30009272-30009272 on Build GRCh38
3111623 c.513G>A p.T171T Chr.16: 30009010-30009010 on Build GRCh38
7304777 c.418-3C>G p.? Chr.16: 30009108-30009108 on Build GRCh38
4986121 c.497C>G p.T166R Chr.16: 30009026-30009026 on Build GRCh38
6144435 c.463G>C p.V155L Chr.16: 30009060-30009060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979698389 Chr.16: 30009007-30009007 on Build GRCh38
rs968659592 Chr.16: 30008980-30008980 on Build GRCh38
rs760238459 Chr.16: 30009141-30009141 on Build GRCh38
rs1021826397 Chr.16: 30009381-30009381 on Build GRCh38
rs766487878 Chr.16: 30009235-30009240 on Build GRCh38
rs200874282 Chr.16: 30009034-30009034 on Build GRCh38
rs759227165 Chr.16: 30009043-30009043 on Build GRCh38
rs754200718 Chr.16: 30008959-30008959 on Build GRCh38
rs770040011 Chr.16: 30008982-30008983 on Build GRCh38
rs780869467 Chr.16: 30009384-30009384 on Build GRCh38
rs766949223 Chr.16: 30009173-30009173 on Build GRCh38
rs763358306 Chr.16: 30009268-30009268 on Build GRCh38
rs765691034 Chr.16: 30009144-30009144 on Build GRCh38
rs1007306841 Chr.16: 30009330-30009330 on Build GRCh38
rs780276872 Chr.16: 30008995-30008995 on Build GRCh38
rs371916491 Chr.16: 30009319-30009319 on Build GRCh38
rs756390032 Chr.16: 30009311-30009311 on Build GRCh38
rs1003812826 Chr.16: 30009361-30009363 on Build GRCh38
rs141230451 Chr.16: 30009077-30009077 on Build GRCh38
rs951035320 Chr.16: 30009352-30009352 on Build GRCh38
rs562821261 Chr.16: 30008956-30008956 on Build GRCh38
rs759950090 Chr.16: 30009299-30009299 on Build GRCh38
rs750795745 Chr.16: 30009149-30009149 on Build GRCh38
rs1002388455 Chr.16: 30009293-30009293 on Build GRCh38
rs769699664 Chr.16: 30009101-30009101 on Build GRCh38
rs1026637467 Chr.16: 30009333-30009333 on Build GRCh38
rs765665192 Chr.16: 30009063-30009063 on Build GRCh38
rs747292511 Chr.16: 30009002-30009002 on Build GRCh38
rs751374412 Chr.16: 30009186-30009186 on Build GRCh38
rs779428518 Chr.16: 30008977-30008977 on Build GRCh38
rs777535448 Chr.16: 30009105-30009105 on Build GRCh38
rs140999204 Chr.16: 30009176-30009176 on Build GRCh38
rs745371931 Chr.16: 30009193-30009193 on Build GRCh38
rs747932681 Chr.16: 30009011-30009011 on Build GRCh38
rs370585487 Chr.16: 30009178-30009178 on Build GRCh38
rs200640037 Chr.16: 30009048-30009048 on Build GRCh38
rs912409908 Chr.16: 30009064-30009064 on Build GRCh38
rs773739403 Chr.16: 30009247-30009247 on Build GRCh38
rs756981220 Chr.16: 30009316-30009316 on Build GRCh38
rs763013871 Chr.16: 30009195-30009196 on Build GRCh38
rs781424313 Chr.16: 30009099-30009099 on Build GRCh38
rs150061510 Chr.16: 30009016-30009016 on Build GRCh38
rs368430709 Chr.16: 30009314-30009314 on Build GRCh38
rs747051337 Chr.16: 30009089-30009089 on Build GRCh38
rs755183524 Chr.16: 30009098-30009098 on Build GRCh38
rs867280784 Chr.16: 30009251-30009251 on Build GRCh38
rs888805591 Chr.16: 30009254-30009254 on Build GRCh38
rs762875731 Chr.16: 30009033-30009033 on Build GRCh38
rs372290822 Chr.16: 30009243-30009243 on Build GRCh38
rs143554943 Chr.16: 30009217-30009217 on Build GRCh38
rs921364526 Chr.16: 30009148-30009148 on Build GRCh38
rs535619781 Chr.16: 30009235-30009235 on Build GRCh38
rs201992999 Chr.16: 30009010-30009010 on Build GRCh38
rs752182130 Chr.16: 30009295-30009295 on Build GRCh38
rs780326177 Chr.16: 30009088-30009088 on Build GRCh38
rs948108194 Chr.16: 30009092-30009092 on Build GRCh38
rs982652670 Chr.16: 30009061-30009061 on Build GRCh38
rs745957156 Chr.16: 30008978-30008978 on Build GRCh38
rs775140393 Chr.16: 30009138-30009138 on Build GRCh38
rs374545340 Chr.16: 30009100-30009100 on Build GRCh38
rs377448847 Chr.16: 30009055-30009055 on Build GRCh38
rs369857979 Chr.16: 30009019-30009019 on Build GRCh38
rs768972930 Chr.16: 30009005-30009005 on Build GRCh38
rs758484437 Chr.16: 30008989-30008989 on Build GRCh38
rs773333448 Chr.16: 30008989-30008989 on Build GRCh38
rs770823790 Chr.16: 30009117-30009117 on Build GRCh38
rs749277235 Chr.16: 30009107-30009107 on Build GRCh38
rs771639462 Chr.16: 30009130-30009130 on Build GRCh38
rs767687842 Chr.16: 30009302-30009302 on Build GRCh38
rs757736610 Chr.16: 30008968-30008968 on Build GRCh38
rs201008541 Chr.16: 30009125-30009125 on Build GRCh38
rs201364446 Chr.16: 30009118-30009118 on Build GRCh38
rs767004365 Chr.16: 30009276-30009276 on Build GRCh38
rs752691978 Chr.16: 30009307-30009307 on Build GRCh38
rs763591269 Chr.16: 30009167-30009167 on Build GRCh38
rs778766962 Chr.16: 30009191-30009191 on Build GRCh38
rs751283499 Chr.16: 30008951-30008951 on Build GRCh38
rs753025242 Chr.16: 30009177-30009177 on Build GRCh38
rs754178826 Chr.16: 30009052-30009052 on Build GRCh38
rs377241271 Chr.16: 30009244-30009244 on Build GRCh38
rs556418417 Chr.16: 30009181-30009181 on Build GRCh38
rs565034123 Chr.16: 30009385-30009385 on Build GRCh38
rs894264953 Chr.16: 30009317-30009317 on Build GRCh38
rs200073865 Chr.16: 30009081-30009081 on Build GRCh38
rs985871482 Chr.16: 30008960-30008960 on Build GRCh38

View Full Product Details