Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30009116-30009610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2A
Entrez Gene ID
8448
Gene Name
double C2 domain alpha
Gene Aliases
Doc2
UniGene
Hs.355281
Gene Chromosome Location
Chr.16: 30005514-30023280 on Build GRCh38
Transcripts
NM_001282062.1 NM_001282063.1 NM_001282068.1 NM_003586.2 NR_104089.1 NR_104090.1 XM_011545975.1 XM_017023776.1 XM_017023777.1 XM_017023778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968575 c.347T>A p.L116H Chr.16: 30009272-30009272 on Build GRCh38
7365198 c.316C>G p.L106V Chr.16: 30009504-30009504 on Build GRCh38
6522751 c.302G>T p.R101L Chr.16: 30009518-30009518 on Build GRCh38
4917605 c.301C>A p.R101R Chr.16: 30009519-30009519 on Build GRCh38
5992011 c.263-5_263-4delTC p.? Chr.16: 30009561-30009562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760238459 Chr.16: 30009141-30009141 on Build GRCh38
rs1021826397 Chr.16: 30009381-30009381 on Build GRCh38
rs374928975 Chr.16: 30009452-30009452 on Build GRCh38
rs766487878 Chr.16: 30009235-30009240 on Build GRCh38
rs200020237 Chr.16: 30009518-30009518 on Build GRCh38
rs527517415 Chr.16: 30009526-30009526 on Build GRCh38
rs369549862 Chr.16: 30009514-30009514 on Build GRCh38
rs779655524 Chr.16: 30009445-30009445 on Build GRCh38
rs562734616 Chr.16: 30009474-30009474 on Build GRCh38
rs865830567 Chr.16: 30009448-30009448 on Build GRCh38
rs780869467 Chr.16: 30009384-30009384 on Build GRCh38
rs1140240 Chr.16: 30009545-30009545 on Build GRCh38
rs766949223 Chr.16: 30009173-30009173 on Build GRCh38
rs763358306 Chr.16: 30009268-30009268 on Build GRCh38
rs748703201 Chr.16: 30009561-30009561 on Build GRCh38
rs765691034 Chr.16: 30009144-30009144 on Build GRCh38
rs1007306841 Chr.16: 30009330-30009330 on Build GRCh38
rs371916491 Chr.16: 30009319-30009319 on Build GRCh38
rs756390032 Chr.16: 30009311-30009311 on Build GRCh38
rs1003812826 Chr.16: 30009361-30009363 on Build GRCh38
rs372830998 Chr.16: 30009567-30009567 on Build GRCh38
rs202175354 Chr.16: 30009481-30009481 on Build GRCh38
rs951035320 Chr.16: 30009352-30009352 on Build GRCh38
rs759950090 Chr.16: 30009299-30009299 on Build GRCh38
rs1048151795 Chr.16: 30009561-30009564 on Build GRCh38
rs1001010704 Chr.16: 30009587-30009587 on Build GRCh38
rs750795745 Chr.16: 30009149-30009149 on Build GRCh38
rs775662234 Chr.16: 30009524-30009524 on Build GRCh38
rs745348941 Chr.16: 30009439-30009439 on Build GRCh38
rs1002388455 Chr.16: 30009293-30009293 on Build GRCh38
rs1026637467 Chr.16: 30009333-30009333 on Build GRCh38
rs919119708 Chr.16: 30009519-30009519 on Build GRCh38
rs751374412 Chr.16: 30009186-30009186 on Build GRCh38
rs552369466 Chr.16: 30009544-30009544 on Build GRCh38
rs140999204 Chr.16: 30009176-30009176 on Build GRCh38
rs745371931 Chr.16: 30009193-30009193 on Build GRCh38
rs370585487 Chr.16: 30009178-30009178 on Build GRCh38
rs1007643898 Chr.16: 30009550-30009550 on Build GRCh38
rs773739403 Chr.16: 30009247-30009247 on Build GRCh38
rs1031226931 Chr.16: 30009418-30009418 on Build GRCh38
rs756981220 Chr.16: 30009316-30009316 on Build GRCh38
rs763013871 Chr.16: 30009195-30009196 on Build GRCh38
rs746459111 Chr.16: 30009451-30009451 on Build GRCh38
rs368430709 Chr.16: 30009314-30009314 on Build GRCh38
rs867280784 Chr.16: 30009251-30009251 on Build GRCh38
rs888805591 Chr.16: 30009254-30009254 on Build GRCh38
rs372290822 Chr.16: 30009243-30009243 on Build GRCh38
rs143554943 Chr.16: 30009217-30009217 on Build GRCh38
rs933220946 Chr.16: 30009522-30009522 on Build GRCh38
rs921364526 Chr.16: 30009148-30009148 on Build GRCh38
rs375995826 Chr.16: 30009560-30009560 on Build GRCh38
rs576762890 Chr.16: 30009427-30009427 on Build GRCh38
rs535619781 Chr.16: 30009235-30009235 on Build GRCh38
rs752182130 Chr.16: 30009295-30009295 on Build GRCh38
rs958141060 Chr.16: 30009438-30009438 on Build GRCh38
rs1001441936 Chr.16: 30009410-30009410 on Build GRCh38
rs560611003 Chr.16: 30009556-30009556 on Build GRCh38
rs754124502 Chr.16: 30009555-30009555 on Build GRCh38
rs767687842 Chr.16: 30009302-30009302 on Build GRCh38
rs767004365 Chr.16: 30009276-30009276 on Build GRCh38
rs771483290 Chr.16: 30009511-30009511 on Build GRCh38
rs752691978 Chr.16: 30009307-30009307 on Build GRCh38
rs763591269 Chr.16: 30009167-30009167 on Build GRCh38
rs778766962 Chr.16: 30009191-30009191 on Build GRCh38
rs111254879 Chr.16: 30009553-30009553 on Build GRCh38
rs753025242 Chr.16: 30009177-30009177 on Build GRCh38
rs377241271 Chr.16: 30009244-30009244 on Build GRCh38
rs556418417 Chr.16: 30009181-30009181 on Build GRCh38
rs1028569697 Chr.16: 30009423-30009423 on Build GRCh38
rs565034123 Chr.16: 30009385-30009385 on Build GRCh38
rs767876453 Chr.16: 30009523-30009523 on Build GRCh38
rs894264953 Chr.16: 30009317-30009317 on Build GRCh38
rs544239082 Chr.16: 30009456-30009456 on Build GRCh38
rs201359056 Chr.16: 30009487-30009487 on Build GRCh38
rs917908736 Chr.16: 30009446-30009446 on Build GRCh38

View Full Product Details