Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30989051-30989560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD3B7
Entrez Gene ID
80270
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases
CBAS1, PFIC4, SDR11E3
UniGene
Hs.460618
Gene Chromosome Location
Chr.16: 30985189-30989152 on Build GRCh38
Transcripts
NM_001142777.1 NM_001142778.1 NM_025193.3 XM_005255601.3 XM_011545960.2 XM_011545961.1 XM_011545962.2 XM_017023732.1 NM_052874.4 XM_017022893.1

Gene Symbol
STX1B
Entrez Gene ID
112755
Gene Name
syntaxin 1B
Gene Aliases
GEFSP9, STX1B1, STX1B2
UniGene
Hs.542230
Gene Chromosome Location
Chr.16: 30989256-31010638 on Build GRCh38
Transcripts
NM_001142777.1 NM_001142778.1 NM_025193.3 XM_005255601.3 XM_011545960.2 XM_011545961.1 XM_011545962.2 XM_017023732.1 NM_052874.4 XM_017022893.1

SNP Detail

SNP ID Chromosome Location
rs922905335 Chr.16: 30989328-30989328 on Build GRCh38
rs952896993 Chr.16: 30989503-30989503 on Build GRCh38
rs143151167 Chr.16: 30989406-30989429 on Build GRCh38
rs992375387 Chr.16: 30989493-30989493 on Build GRCh38
rs905698420 Chr.16: 30989454-30989454 on Build GRCh38
rs1025682766 Chr.16: 30989490-30989490 on Build GRCh38
rs558594252 Chr.16: 30989077-30989077 on Build GRCh38
rs960048638 Chr.16: 30989178-30989178 on Build GRCh38
rs3833843 Chr.16: 30989432-30989455 on Build GRCh38
rs953978639 Chr.16: 30989158-30989158 on Build GRCh38
rs961318568 Chr.16: 30989353-30989354 on Build GRCh38
rs895766748 Chr.16: 30989487-30989487 on Build GRCh38
rs576911808 Chr.16: 30989355-30989355 on Build GRCh38
rs1029545219 Chr.16: 30989354-30989354 on Build GRCh38
rs1052870856 Chr.16: 30989076-30989076 on Build GRCh38
rs977766614 Chr.16: 30989442-30989443 on Build GRCh38
rs911427472 Chr.16: 30989250-30989250 on Build GRCh38
rs553756111 Chr.16: 30989195-30989195 on Build GRCh38
rs1018817160 Chr.16: 30989528-30989528 on Build GRCh38
rs998110370 Chr.16: 30989353-30989353 on Build GRCh38
rs980850295 Chr.16: 30989429-30989429 on Build GRCh38
rs918653114 Chr.16: 30989212-30989212 on Build GRCh38
rs1035616529 Chr.16: 30989169-30989169 on Build GRCh38
rs560440933 Chr.16: 30989234-30989234 on Build GRCh38
rs13708 Chr.16: 30989488-30989488 on Build GRCh38
rs984228695 Chr.16: 30989508-30989508 on Build GRCh38
rs531518746 Chr.16: 30989105-30989105 on Build GRCh38
rs914950863 Chr.16: 30989199-30989199 on Build GRCh38
rs895327697 Chr.16: 30989252-30989252 on Build GRCh38
rs976618753 Chr.16: 30989150-30989150 on Build GRCh38
rs557817158 Chr.16: 30989356-30989356 on Build GRCh38
rs985228837 Chr.16: 30989196-30989196 on Build GRCh38
rs932962840 Chr.16: 30989331-30989331 on Build GRCh38
rs948274277 Chr.16: 30989516-30989516 on Build GRCh38
rs942893234 Chr.16: 30989269-30989269 on Build GRCh38
rs887843501 Chr.16: 30989318-30989318 on Build GRCh38
rs947192427 Chr.16: 30989357-30989357 on Build GRCh38
rs1042434677 Chr.16: 30989210-30989210 on Build GRCh38
rs747264647 Chr.16: 30989349-30989349 on Build GRCh38
rs1041319817 Chr.16: 30989322-30989322 on Build GRCh38
rs1012790886 Chr.16: 30989284-30989284 on Build GRCh38
rs924218702 Chr.16: 30989213-30989213 on Build GRCh38
rs915026604 Chr.16: 30989368-30989368 on Build GRCh38
rs1035584276 Chr.16: 30989480-30989480 on Build GRCh38
rs923603198 Chr.16: 30989464-30989464 on Build GRCh38
rs867059850 Chr.16: 30989146-30989146 on Build GRCh38
rs535757515 Chr.16: 30989155-30989155 on Build GRCh38
rs576082177 Chr.16: 30989359-30989359 on Build GRCh38
rs548070215 Chr.16: 30989126-30989126 on Build GRCh38
rs968184780 Chr.16: 30989438-30989438 on Build GRCh38
rs550022266 Chr.16: 30989125-30989125 on Build GRCh38
rs970373221 Chr.16: 30989149-30989149 on Build GRCh38
rs1049674592 Chr.16: 30989519-30989519 on Build GRCh38
rs984691581 Chr.16: 30989247-30989247 on Build GRCh38
rs543583485 Chr.16: 30989463-30989463 on Build GRCh38
rs751623756 Chr.16: 30989293-30989293 on Build GRCh38
rs961264848 Chr.16: 30989145-30989145 on Build GRCh38
rs916808800 Chr.16: 30989494-30989494 on Build GRCh38
rs972763470 Chr.16: 30989200-30989200 on Build GRCh38

View Full Product Details