Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31109591-31110098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
UniGene
Hs.513520
Gene Chromosome Location
Chr.16: 31108294-31117651 on Build GRCh38
Transcripts
NM_001122957.2 NM_001271926.1 NM_005881.3 XM_017022859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6610579 c.283C>T p.Q95* Chr.16: 31109691-31109691 on Build GRCh38
5023659 c.376-8C>T p.? Chr.16: 31110069-31110069 on Build GRCh38
4665377 c.392G>A p.R131H Chr.16: 31110093-31110093 on Build GRCh38
5451388 c.375+4A>C p.? Chr.16: 31109787-31109787 on Build GRCh38
3969545 c.328C>T p.R110C Chr.16: 31109736-31109736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371158752 Chr.16: 31109830-31109830 on Build GRCh38
rs556255123 Chr.16: 31109713-31109713 on Build GRCh38
rs891760155 Chr.16: 31109811-31109811 on Build GRCh38
rs138595718 Chr.16: 31109873-31109873 on Build GRCh38
rs144675159 Chr.16: 31109653-31109653 on Build GRCh38
rs764266445 Chr.16: 31110069-31110069 on Build GRCh38
rs200230790 Chr.16: 31110034-31110034 on Build GRCh38
rs887271429 Chr.16: 31109745-31109745 on Build GRCh38
rs1002056919 Chr.16: 31109863-31109863 on Build GRCh38
rs746683923 Chr.16: 31109829-31109829 on Build GRCh38
rs574589732 Chr.16: 31109809-31109809 on Build GRCh38
rs772489972 Chr.16: 31110043-31110043 on Build GRCh38
rs749243097 Chr.16: 31109696-31109696 on Build GRCh38
rs967610602 Chr.16: 31109871-31109871 on Build GRCh38
rs903717685 Chr.16: 31109691-31109691 on Build GRCh38
rs374518693 Chr.16: 31109646-31109646 on Build GRCh38
rs910902195 Chr.16: 31109927-31109927 on Build GRCh38
rs957251871 Chr.16: 31109901-31109901 on Build GRCh38
rs144747578 Chr.16: 31109680-31109680 on Build GRCh38
rs750237108 Chr.16: 31109825-31109825 on Build GRCh38
rs571884389 Chr.16: 31109853-31109853 on Build GRCh38
rs572628932 Chr.16: 31109950-31109950 on Build GRCh38
rs745531188 Chr.16: 31109716-31109716 on Build GRCh38
rs757332822 Chr.16: 31109819-31109819 on Build GRCh38
rs942452459 Chr.16: 31109938-31109938 on Build GRCh38
rs925103138 Chr.16: 31109943-31109943 on Build GRCh38
rs779596120 Chr.16: 31109754-31109754 on Build GRCh38
rs148112682 Chr.16: 31109780-31109780 on Build GRCh38
rs970151601 Chr.16: 31109846-31109846 on Build GRCh38
rs778821974 Chr.16: 31109823-31109823 on Build GRCh38
rs910988235 Chr.16: 31109626-31109629 on Build GRCh38
rs918848108 Chr.16: 31109898-31109898 on Build GRCh38
rs751764506 Chr.16: 31109625-31109625 on Build GRCh38
rs746246063 Chr.16: 31109687-31109688 on Build GRCh38
rs767584077 Chr.16: 31109799-31109799 on Build GRCh38
rs775517661 Chr.16: 31110045-31110045 on Build GRCh38
rs1055766387 Chr.16: 31109645-31109645 on Build GRCh38
rs776991732 Chr.16: 31109971-31109971 on Build GRCh38
rs771798261 Chr.16: 31109791-31109792 on Build GRCh38
rs184927873 Chr.16: 31109957-31109957 on Build GRCh38
rs564838932 Chr.16: 31110029-31110029 on Build GRCh38
rs925780476 Chr.16: 31109891-31109891 on Build GRCh38
rs202110382 Chr.16: 31110052-31110052 on Build GRCh38
rs375986099 Chr.16: 31109870-31109870 on Build GRCh38
rs1009047605 Chr.16: 31109864-31109864 on Build GRCh38
rs370133758 Chr.16: 31109642-31109642 on Build GRCh38
rs1052316770 Chr.16: 31109954-31109954 on Build GRCh38
rs267604516 Chr.16: 31109710-31109710 on Build GRCh38
rs991418085 Chr.16: 31109903-31109903 on Build GRCh38
rs753814671 Chr.16: 31109818-31109818 on Build GRCh38
rs368623088 Chr.16: 31109810-31109810 on Build GRCh38
rs760604265 Chr.16: 31109811-31109811 on Build GRCh38
rs757061262 Chr.16: 31109704-31109704 on Build GRCh38
rs915900895 Chr.16: 31109939-31109939 on Build GRCh38
rs371628815 Chr.16: 31109812-31109812 on Build GRCh38
rs141282419 Chr.16: 31109781-31109781 on Build GRCh38
rs746459006 Chr.16: 31109755-31109755 on Build GRCh38
rs932889417 Chr.16: 31110073-31110073 on Build GRCh38
rs771666142 Chr.16: 31109737-31109737 on Build GRCh38
rs771279207 Chr.16: 31109793-31109793 on Build GRCh38
rs201870679 Chr.16: 31109681-31109681 on Build GRCh38
rs777812892 Chr.16: 31109683-31109683 on Build GRCh38
rs181894154 Chr.16: 31109947-31109947 on Build GRCh38
rs940102319 Chr.16: 31110037-31110037 on Build GRCh38
rs757688773 Chr.16: 31109682-31109684 on Build GRCh38
rs986500680 Chr.16: 31109926-31109926 on Build GRCh38
rs768248141 Chr.16: 31109775-31109775 on Build GRCh38
rs200118163 Chr.16: 31109826-31109826 on Build GRCh38
rs759779446 Chr.16: 31109797-31109797 on Build GRCh38
rs948383378 Chr.16: 31109913-31109933 on Build GRCh38
rs774735276 Chr.16: 31109796-31109796 on Build GRCh38
rs905896427 Chr.16: 31109984-31109984 on Build GRCh38
rs935912612 Chr.16: 31109613-31109613 on Build GRCh38
rs1004418237 Chr.16: 31109747-31109747 on Build GRCh38
rs749761316 Chr.16: 31110032-31110032 on Build GRCh38
rs772770984 Chr.16: 31109914-31109914 on Build GRCh38
rs779349318 Chr.16: 31109685-31109686 on Build GRCh38
rs768136995 Chr.16: 31109831-31109831 on Build GRCh38
rs780744605 Chr.16: 31109834-31109834 on Build GRCh38
rs949353223 Chr.16: 31109960-31109960 on Build GRCh38
rs997071412 Chr.16: 31109800-31109800 on Build GRCh38
rs775238667 Chr.16: 31109826-31109826 on Build GRCh38
rs779537233 Chr.16: 31109827-31109827 on Build GRCh38
rs759778061 Chr.16: 31110038-31110038 on Build GRCh38

View Full Product Details