Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48096507-48097009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC12
Entrez Gene ID
94160
Gene Name
ATP binding cassette subfamily C member 12
Gene Aliases
MRP9
UniGene
Hs.410111
Gene Chromosome Location
Chr.16: 48082973-48146770 on Build GRCh38
Transcripts
NM_033226.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5384912 c.3161C>T p.S1054L Chr.16: 48096780-48096780 on Build GRCh38
2832869 c.3094C>A p.L1032I Chr.16: 48096847-48096847 on Build GRCh38
3509808 c.3049T>C p.Y1017H Chr.16: 48096892-48096892 on Build GRCh38
3509807 c.3072G>A p.W1024* Chr.16: 48096869-48096869 on Build GRCh38
1301992 c.3066C>T p.L1022L Chr.16: 48096875-48096875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866003769 Chr.16: 48096743-48096743 on Build GRCh38
rs777055206 Chr.16: 48096780-48096780 on Build GRCh38
rs2019240 Chr.16: 48096923-48096923 on Build GRCh38
rs534666690 Chr.16: 48096538-48096538 on Build GRCh38
rs553803646 Chr.16: 48096627-48096627 on Build GRCh38
rs760867676 Chr.16: 48096886-48096887 on Build GRCh38
rs192674180 Chr.16: 48096954-48096954 on Build GRCh38
rs1036483788 Chr.16: 48096691-48096691 on Build GRCh38
rs565920027 Chr.16: 48096644-48096644 on Build GRCh38
rs532273327 Chr.16: 48096835-48096835 on Build GRCh38
rs754774686 Chr.16: 48096981-48096981 on Build GRCh38
rs530897385 Chr.16: 48096696-48096696 on Build GRCh38
rs377482004 Chr.16: 48096938-48096938 on Build GRCh38
rs369953644 Chr.16: 48096700-48096700 on Build GRCh38
rs1052220289 Chr.16: 48096655-48096655 on Build GRCh38
rs773415702 Chr.16: 48096765-48096765 on Build GRCh38
rs1006994736 Chr.16: 48096584-48096584 on Build GRCh38
rs528428460 Chr.16: 48096614-48096614 on Build GRCh38
rs770322404 Chr.16: 48096920-48096920 on Build GRCh38
rs571436457 Chr.16: 48096688-48096688 on Build GRCh38
rs567163495 Chr.16: 48096713-48096713 on Build GRCh38
rs760207717 Chr.16: 48096977-48096977 on Build GRCh38
rs551350688 Chr.16: 48096664-48096664 on Build GRCh38
rs768646653 Chr.16: 48096728-48096728 on Build GRCh38
rs768769755 Chr.16: 48096724-48096724 on Build GRCh38
rs942253370 Chr.16: 48096892-48096892 on Build GRCh38
rs756918926 Chr.16: 48096698-48096698 on Build GRCh38
rs771140837 Chr.16: 48096773-48096773 on Build GRCh38
rs536065631 Chr.16: 48096650-48096650 on Build GRCh38
rs375570056 Chr.16: 48096723-48096723 on Build GRCh38
rs748127550 Chr.16: 48096899-48096899 on Build GRCh38
rs763443563 Chr.16: 48096695-48096695 on Build GRCh38
rs36102575 Chr.16: 48096870-48096870 on Build GRCh38
rs888178356 Chr.16: 48096860-48096861 on Build GRCh38
rs1010230017 Chr.16: 48096687-48096687 on Build GRCh38
rs550910113 Chr.16: 48096697-48096697 on Build GRCh38
rs769382230 Chr.16: 48096669-48096669 on Build GRCh38
rs767204530 Chr.16: 48096830-48096830 on Build GRCh38
rs79688031 Chr.16: 48096913-48096913 on Build GRCh38
rs948009863 Chr.16: 48096663-48096663 on Build GRCh38
rs764222617 Chr.16: 48096836-48096836 on Build GRCh38
rs142376716 Chr.16: 48096793-48096793 on Build GRCh38
rs200089399 Chr.16: 48096858-48096858 on Build GRCh38
rs541834952 Chr.16: 48096908-48096908 on Build GRCh38
rs576249592 Chr.16: 48096914-48096914 on Build GRCh38
rs778804010 Chr.16: 48096891-48096891 on Build GRCh38
rs929625616 Chr.16: 48096585-48096585 on Build GRCh38
rs761011793 Chr.16: 48096771-48096771 on Build GRCh38
rs760457854 Chr.16: 48096649-48096649 on Build GRCh38
rs890487345 Chr.16: 48096686-48096686 on Build GRCh38
rs778926587 Chr.16: 48096740-48096740 on Build GRCh38
rs150779342 Chr.16: 48096863-48096863 on Build GRCh38
rs759920573 Chr.16: 48096917-48096917 on Build GRCh38
rs982062051 Chr.16: 48096886-48096886 on Build GRCh38
rs764137323 Chr.16: 48096959-48096961 on Build GRCh38
rs748254080 Chr.16: 48096747-48096747 on Build GRCh38
rs139138303 Chr.16: 48096864-48096864 on Build GRCh38
rs776810597 Chr.16: 48096916-48096916 on Build GRCh38
rs559490689 Chr.16: 48096604-48096604 on Build GRCh38
rs376472401 Chr.16: 48096648-48096648 on Build GRCh38
rs544586060 Chr.16: 48096861-48096861 on Build GRCh38
rs372317501 Chr.16: 48096706-48096706 on Build GRCh38
rs369089648 Chr.16: 48096831-48096831 on Build GRCh38
rs951084163 Chr.16: 48096859-48096859 on Build GRCh38
rs753304736 Chr.16: 48096795-48096795 on Build GRCh38
rs942243540 Chr.16: 48096628-48096628 on Build GRCh38
rs766080498 Chr.16: 48096978-48096978 on Build GRCh38
rs199935431 Chr.16: 48096705-48096705 on Build GRCh38
rs747130827 Chr.16: 48096910-48096910 on Build GRCh38
rs148538340 Chr.16: 48096758-48096758 on Build GRCh38
rs916531292 Chr.16: 48096657-48096657 on Build GRCh38
rs777840016 Chr.16: 48096907-48096907 on Build GRCh38
rs267604557 Chr.16: 48096807-48096807 on Build GRCh38
rs187842439 Chr.16: 48096904-48096904 on Build GRCh38
rs927864107 Chr.16: 48096906-48096906 on Build GRCh38
rs761374827 Chr.16: 48096815-48096815 on Build GRCh38
rs1000443174 Chr.16: 48096712-48096712 on Build GRCh38
rs573782879 Chr.16: 48096794-48096794 on Build GRCh38
rs763613929 Chr.16: 48096924-48096924 on Build GRCh38
rs982414332 Chr.16: 48096683-48096683 on Build GRCh38
rs1047994383 Chr.16: 48096610-48096610 on Build GRCh38

View Full Product Details